Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92110998-92111510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB3
Entrez Gene ID
22849
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
UniGene
Hs.131683
Gene Chromosome Location
Chr.10: 92046692-92291118 on Build GRCh38
Transcripts
NM_001178137.1 NM_014912.4 XM_005269630.4 XM_006717714.3 XM_006717715.3 XM_011539514.2 XM_011539515.2 XM_011539517.2 XM_011539518.2 XM_011539519.2 XM_011539520.2 XM_017015927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807984 c.1428G>T p.L476L Chr.10: 92111178-92111178 on Build GRCh38
6569211 c.1518C>G p.I506M Chr.10: 92111088-92111088 on Build GRCh38
4865869 c.1488A>C p.K496N Chr.10: 92111118-92111118 on Build GRCh38
6634540 c.1466C>T p.A489V Chr.10: 92111140-92111140 on Build GRCh38
145261 c.1504G>A p.D502N Chr.10: 92111144-92111144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028897158 Chr.10: 92111343-92111343 on Build GRCh38
rs201037277 Chr.10: 92111217-92111217 on Build GRCh38
rs192431236 Chr.10: 92111281-92111281 on Build GRCh38
rs145051728 Chr.10: 92111310-92111310 on Build GRCh38
rs753487614 Chr.10: 92111467-92111467 on Build GRCh38
rs371269451 Chr.10: 92111118-92111118 on Build GRCh38
rs766452754 Chr.10: 92111175-92111175 on Build GRCh38
rs773591106 Chr.10: 92111159-92111159 on Build GRCh38
rs1033734029 Chr.10: 92111474-92111474 on Build GRCh38
rs191121892 Chr.10: 92111088-92111088 on Build GRCh38
rs747527259 Chr.10: 92111244-92111244 on Build GRCh38
rs11186829 Chr.10: 92111461-92111461 on Build GRCh38
rs701846 Chr.10: 92111309-92111309 on Build GRCh38
rs1014138572 Chr.10: 92111253-92111253 on Build GRCh38
rs764783295 Chr.10: 92111475-92111475 on Build GRCh38
rs554468609 Chr.10: 92111271-92111271 on Build GRCh38
rs932415259 Chr.10: 92111202-92111202 on Build GRCh38
rs752573875 Chr.10: 92111084-92111085 on Build GRCh38
rs112326498 Chr.10: 92111028-92111028 on Build GRCh38
rs141004546 Chr.10: 92111218-92111218 on Build GRCh38
rs763518472 Chr.10: 92111264-92111264 on Build GRCh38
rs759134140 Chr.10: 92111468-92111468 on Build GRCh38
rs200566432 Chr.10: 92111082-92111082 on Build GRCh38
rs762008054 Chr.10: 92111116-92111116 on Build GRCh38
rs568985716 Chr.10: 92111029-92111029 on Build GRCh38
rs1010018784 Chr.10: 92111350-92111350 on Build GRCh38
rs987067418 Chr.10: 92111462-92111462 on Build GRCh38
rs542819457 Chr.10: 92111283-92111283 on Build GRCh38
rs922378372 Chr.10: 92111091-92111091 on Build GRCh38
rs541945091 Chr.10: 92111348-92111348 on Build GRCh38
rs892707295 Chr.10: 92111329-92111329 on Build GRCh38
rs34268975 Chr.10: 92111458-92111459 on Build GRCh38
rs1053989679 Chr.10: 92111326-92111326 on Build GRCh38
rs1024973456 Chr.10: 92111453-92111453 on Build GRCh38
rs775478869 Chr.10: 92111234-92111234 on Build GRCh38
rs376947453 Chr.10: 92111237-92111237 on Build GRCh38
rs750294986 Chr.10: 92111125-92111125 on Build GRCh38
rs369944745 Chr.10: 92111115-92111115 on Build GRCh38
rs768559309 Chr.10: 92111113-92111113 on Build GRCh38
rs760812609 Chr.10: 92111089-92111089 on Build GRCh38
rs769427027 Chr.10: 92111079-92111079 on Build GRCh38
rs889297257 Chr.10: 92111225-92111225 on Build GRCh38
rs754533657 Chr.10: 92111205-92111205 on Build GRCh38
rs183447446 Chr.10: 92111463-92111463 on Build GRCh38
rs774842917 Chr.10: 92111306-92111306 on Build GRCh38
rs747702289 Chr.10: 92111067-92111067 on Build GRCh38
rs751388716 Chr.10: 92111193-92111193 on Build GRCh38
rs780621735 Chr.10: 92111240-92111240 on Build GRCh38
rs772588360 Chr.10: 92111085-92111085 on Build GRCh38
rs754409665 Chr.10: 92111038-92111038 on Build GRCh38
rs780905458 Chr.10: 92111047-92111047 on Build GRCh38
rs557785608 Chr.10: 92111084-92111084 on Build GRCh38
rs765532394 Chr.10: 92111066-92111067 on Build GRCh38
rs1056018815 Chr.10: 92111178-92111178 on Build GRCh38
rs1032795769 Chr.10: 92111319-92111319 on Build GRCh38
rs762788690 Chr.10: 92111164-92111164 on Build GRCh38

View Full Product Details