Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94941731-94942238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C9
Entrez Gene ID
1559
Gene Name
cytochrome P450 family 2 subfamily C member 9
Gene Aliases
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
UniGene
Hs.282624
Gene Chromosome Location
Chr.10: 94905867-94989395 on Build GRCh38
Transcripts
NM_000771.3 XM_017015758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4990023 c.265G>A p.D89N Chr.10: 94941954-94941954 on Build GRCh38
4486168 c.301C>T p.P101S Chr.10: 94941990-94941990 on Build GRCh38
5913026 c.331+5G>C p.? Chr.10: 94942025-94942025 on Build GRCh38
5919836 c.332G>A p.G111E Chr.10: 94942192-94942192 on Build GRCh38
5919838 c.169-5C>T p.? Chr.10: 94941853-94941853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762239445 Chr.10: 94941982-94941982 on Build GRCh38
rs374133592 Chr.10: 94942153-94942153 on Build GRCh38
rs111553080 Chr.10: 94942190-94942190 on Build GRCh38
rs772658219 Chr.10: 94941889-94941889 on Build GRCh38
rs781137080 Chr.10: 94941993-94941993 on Build GRCh38
rs757861807 Chr.10: 94942064-94942066 on Build GRCh38
rs750662900 Chr.10: 94941984-94941984 on Build GRCh38
rs113044007 Chr.10: 94942097-94942097 on Build GRCh38
rs770279317 Chr.10: 94942052-94942052 on Build GRCh38
rs374466581 Chr.10: 94942115-94942115 on Build GRCh38
rs139656048 Chr.10: 94941952-94941952 on Build GRCh38
rs756823317 Chr.10: 94942179-94942179 on Build GRCh38
rs9332121 Chr.10: 94942182-94942182 on Build GRCh38
rs780906222 Chr.10: 94941946-94941946 on Build GRCh38
rs747732331 Chr.10: 94941867-94941867 on Build GRCh38
rs752668480 Chr.10: 94941995-94941995 on Build GRCh38
rs111808915 Chr.10: 94941909-94941909 on Build GRCh38
rs773973620 Chr.10: 94941968-94941968 on Build GRCh38
rs976841079 Chr.10: 94941811-94941811 on Build GRCh38
rs756921618 Chr.10: 94941834-94941836 on Build GRCh38
rs559536673 Chr.10: 94942189-94942189 on Build GRCh38
rs757562041 Chr.10: 94941828-94941828 on Build GRCh38
rs767942291 Chr.10: 94941817-94941817 on Build GRCh38
rs948296481 Chr.10: 94942083-94942083 on Build GRCh38
rs867193471 Chr.10: 94941874-94941874 on Build GRCh38
rs896332784 Chr.10: 94942046-94942046 on Build GRCh38
rs1023441738 Chr.10: 94941777-94941777 on Build GRCh38
rs779778121 Chr.10: 94942184-94942184 on Build GRCh38
rs754095470 Chr.10: 94941827-94941827 on Build GRCh38
rs202113950 Chr.10: 94941918-94941918 on Build GRCh38
rs777257186 Chr.10: 94942045-94942045 on Build GRCh38
rs372078207 Chr.10: 94942050-94942050 on Build GRCh38
rs201050615 Chr.10: 94941893-94941893 on Build GRCh38
rs9332119 Chr.10: 94941844-94941844 on Build GRCh38
rs951007542 Chr.10: 94941837-94941837 on Build GRCh38
rs779113278 Chr.10: 94942166-94942166 on Build GRCh38
rs773630199 Chr.10: 94942068-94942068 on Build GRCh38
rs199560417 Chr.10: 94942186-94942186 on Build GRCh38
rs758317601 Chr.10: 94941843-94941843 on Build GRCh38
rs761033063 Chr.10: 94941976-94941976 on Build GRCh38
rs528415233 Chr.10: 94941790-94941790 on Build GRCh38
rs201491545 Chr.10: 94941906-94941906 on Build GRCh38
rs762081829 Chr.10: 94941907-94941907 on Build GRCh38
rs550654438 Chr.10: 94941849-94941849 on Build GRCh38
rs371055887 Chr.10: 94941897-94941897 on Build GRCh38
rs775834634 Chr.10: 94941898-94941898 on Build GRCh38
rs888697150 Chr.10: 94941830-94941830 on Build GRCh38
rs928126083 Chr.10: 94942095-94942095 on Build GRCh38
rs200794294 Chr.10: 94941929-94941929 on Build GRCh38
rs759580344 Chr.10: 94942149-94942149 on Build GRCh38
rs754271461 Chr.10: 94941902-94941902 on Build GRCh38
rs905976904 Chr.10: 94942075-94942075 on Build GRCh38
rs868456968 Chr.10: 94941786-94941786 on Build GRCh38
rs761298103 Chr.10: 94941899-94941899 on Build GRCh38
rs1005106504 Chr.10: 94942100-94942100 on Build GRCh38
rs770329611 Chr.10: 94941965-94941965 on Build GRCh38
rs72558187 Chr.10: 94941958-94941958 on Build GRCh38
rs1056361080 Chr.10: 94941983-94941983 on Build GRCh38
rs774607211 Chr.10: 94942216-94942216 on Build GRCh38
rs1012470155 Chr.10: 94942061-94942061 on Build GRCh38
rs1045243646 Chr.10: 94941862-94941862 on Build GRCh38
rs769449338 Chr.10: 94941879-94941879 on Build GRCh38
rs767631303 Chr.10: 94942152-94942152 on Build GRCh38
rs377542177 Chr.10: 94941960-94941960 on Build GRCh38
rs559387905 Chr.10: 94941787-94941787 on Build GRCh38
rs371806531 Chr.10: 94942175-94942175 on Build GRCh38
rs755877934 Chr.10: 94941999-94941999 on Build GRCh38
rs745417361 Chr.10: 94942014-94942014 on Build GRCh38
rs771750993 Chr.10: 94942043-94942043 on Build GRCh38
rs17847036 Chr.10: 94941917-94941917 on Build GRCh38
rs144043022 Chr.10: 94941953-94941953 on Build GRCh38
rs1009885743 Chr.10: 94942120-94942120 on Build GRCh38
rs776477272 Chr.10: 94941850-94941850 on Build GRCh38
rs764565287 Chr.10: 94941823-94941823 on Build GRCh38
rs770634494 Chr.10: 94942006-94942006 on Build GRCh38
rs1009856123 Chr.10: 94941920-94941920 on Build GRCh38
rs766645332 Chr.10: 94942141-94942141 on Build GRCh38
rs750075832 Chr.10: 94941962-94941977 on Build GRCh38
rs370315268 Chr.10: 94941892-94941892 on Build GRCh38
rs778600416 Chr.10: 94942180-94942180 on Build GRCh38
rs201856860 Chr.10: 94942196-94942196 on Build GRCh38
rs374201833 Chr.10: 94941979-94941979 on Build GRCh38
rs754947384 Chr.10: 94941936-94941936 on Build GRCh38
rs538852786 Chr.10: 94941901-94941901 on Build GRCh38
rs200363544 Chr.10: 94942174-94942174 on Build GRCh38
rs77811631 Chr.10: 94941818-94941818 on Build GRCh38
rs778240430 Chr.10: 94942011-94942011 on Build GRCh38
rs1006763207 Chr.10: 94942169-94942169 on Build GRCh38
rs756107008 Chr.10: 94942170-94942170 on Build GRCh38
rs375198934 Chr.10: 94942181-94942181 on Build GRCh38
rs748884303 Chr.10: 94942005-94942005 on Build GRCh38
rs189151736 Chr.10: 94941836-94941836 on Build GRCh38
rs918588278 Chr.10: 94941813-94941813 on Build GRCh38
rs771237265 Chr.10: 94942203-94942203 on Build GRCh38
rs543422060 Chr.10: 94942098-94942098 on Build GRCh38
rs1019133201 Chr.10: 94942123-94942123 on Build GRCh38
rs552391533 Chr.10: 94941766-94941766 on Build GRCh38
rs760633019 Chr.10: 94942070-94942070 on Build GRCh38
rs371138960 Chr.10: 94941923-94941923 on Build GRCh38
rs766327823 Chr.10: 94941888-94941888 on Build GRCh38
rs966304252 Chr.10: 94942131-94942131 on Build GRCh38
rs754820878 Chr.10: 94941991-94941991 on Build GRCh38
rs563432894 Chr.10: 94942142-94942142 on Build GRCh38
rs199569641 Chr.10: 94942065-94942065 on Build GRCh38
rs267602639 Chr.10: 94941989-94941989 on Build GRCh38
rs9332120 Chr.10: 94942093-94942093 on Build GRCh38
rs767093588 Chr.10: 94941862-94941863 on Build GRCh38
rs975346974 Chr.10: 94942166-94942166 on Build GRCh38
rs371997605 Chr.10: 94941954-94941954 on Build GRCh38
rs532137731 Chr.10: 94941764-94941764 on Build GRCh38
rs367848139 Chr.10: 94942001-94942001 on Build GRCh38
rs750716359 Chr.10: 94941910-94941910 on Build GRCh38

View Full Product Details