Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96532973-96533486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM9SF3
Entrez Gene ID
56889
Gene Name
transmembrane 9 superfamily member 3
Gene Aliases
EP70-P-iso, SMBP
UniGene
Hs.500674
Gene Chromosome Location
Chr.10: 96518110-96587052 on Build GRCh38
Transcripts
NM_020123.3 XM_011539976.2 XM_011539977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5566025 c.1285C>A p.R429S Chr.10: 96533091-96533091 on Build GRCh38
7221580 c.1253G>A p.R418Q Chr.10: 96533123-96533123 on Build GRCh38
4016989 c.1261T>C p.S421P Chr.10: 96533115-96533115 on Build GRCh38
921918 c.1239T>C p.G413G Chr.10: 96533137-96533137 on Build GRCh38
1350122 c.1323_1324insA p.W442fs*23 Chr.10: 96533052-96533053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042562399 Chr.10: 96533293-96533293 on Build GRCh38
rs372088218 Chr.10: 96533007-96533007 on Build GRCh38
rs886347819 Chr.10: 96533426-96533426 on Build GRCh38
rs751623216 Chr.10: 96533127-96533127 on Build GRCh38
rs776147486 Chr.10: 96533029-96533029 on Build GRCh38
rs780052460 Chr.10: 96533009-96533009 on Build GRCh38
rs145349156 Chr.10: 96533098-96533098 on Build GRCh38
rs775979542 Chr.10: 96533305-96533306 on Build GRCh38
rs752524997 Chr.10: 96533160-96533160 on Build GRCh38
rs754953961 Chr.10: 96533131-96533131 on Build GRCh38
rs962790459 Chr.10: 96533110-96533110 on Build GRCh38
rs372458773 Chr.10: 96533026-96533026 on Build GRCh38
rs762249987 Chr.10: 96533221-96533221 on Build GRCh38
rs199584414 Chr.10: 96533089-96533089 on Build GRCh38
rs769273178 Chr.10: 96533068-96533068 on Build GRCh38
rs763096470 Chr.10: 96533101-96533101 on Build GRCh38
rs781079968 Chr.10: 96533142-96533142 on Build GRCh38
rs369043118 Chr.10: 96533015-96533015 on Build GRCh38
rs575134059 Chr.10: 96533270-96533270 on Build GRCh38
rs963136734 Chr.10: 96533461-96533461 on Build GRCh38
rs759033037 Chr.10: 96533237-96533249 on Build GRCh38
rs758359677 Chr.10: 96533004-96533004 on Build GRCh38
rs1042554425 Chr.10: 96533272-96533272 on Build GRCh38
rs755819136 Chr.10: 96533171-96533171 on Build GRCh38
rs902922913 Chr.10: 96533312-96533312 on Build GRCh38
rs573002896 Chr.10: 96533403-96533403 on Build GRCh38
rs762042642 Chr.10: 96533072-96533072 on Build GRCh38
rs778623686 Chr.10: 96533188-96533188 on Build GRCh38
rs374784977 Chr.10: 96533011-96533011 on Build GRCh38
rs1005129080 Chr.10: 96533437-96533437 on Build GRCh38
rs746878794 Chr.10: 96533010-96533010 on Build GRCh38
rs760514449 Chr.10: 96533455-96533455 on Build GRCh38
rs776152880 Chr.10: 96533210-96533211 on Build GRCh38
rs1038696458 Chr.10: 96533353-96533353 on Build GRCh38
rs761333719 Chr.10: 96533030-96533030 on Build GRCh38
rs777228514 Chr.10: 96533217-96533217 on Build GRCh38
rs766509352 Chr.10: 96533118-96533118 on Build GRCh38
rs900150514 Chr.10: 96533360-96533360 on Build GRCh38
rs919328053 Chr.10: 96533067-96533067 on Build GRCh38
rs918156031 Chr.10: 96533247-96533251 on Build GRCh38
rs747912378 Chr.10: 96533012-96533012 on Build GRCh38
rs745350224 Chr.10: 96533201-96533201 on Build GRCh38
rs748854744 Chr.10: 96533025-96533025 on Build GRCh38
rs556875070 Chr.10: 96533210-96533210 on Build GRCh38
rs1051669910 Chr.10: 96533394-96533394 on Build GRCh38
rs376285496 Chr.10: 96533084-96533084 on Build GRCh38
rs368077941 Chr.10: 96533279-96533281 on Build GRCh38
rs867212091 Chr.10: 96533063-96533063 on Build GRCh38
rs997367306 Chr.10: 96533001-96533001 on Build GRCh38
rs540426398 Chr.10: 96533430-96533430 on Build GRCh38
rs770614241 Chr.10: 96533186-96533186 on Build GRCh38
rs777254722 Chr.10: 96533017-96533017 on Build GRCh38
rs777497827 Chr.10: 96533184-96533184 on Build GRCh38
rs538195039 Chr.10: 96533185-96533185 on Build GRCh38
rs777185537 Chr.10: 96533069-96533069 on Build GRCh38
rs950884960 Chr.10: 96533090-96533090 on Build GRCh38
rs770355878 Chr.10: 96533004-96533005 on Build GRCh38
rs116111327 Chr.10: 96533369-96533369 on Build GRCh38
rs769985417 Chr.10: 96533230-96533230 on Build GRCh38
rs41291634 Chr.10: 96533283-96533283 on Build GRCh38
rs547161349 Chr.10: 96533246-96533246 on Build GRCh38
rs1016057171 Chr.10: 96533440-96533440 on Build GRCh38

View Full Product Details