Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1841355-1841847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNI2
Entrez Gene ID
7136
Gene Name
troponin I2, fast skeletal type
Gene Aliases
AMCD2B, DA2B, FSSV, fsTnI
UniGene
Hs.523403
Gene Chromosome Location
Chr.11: 1839003-1841680 on Build GRCh38
Transcripts
NM_001145829.1 NM_001145841.1 NM_003282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1353018 c.467G>A p.R156Q Chr.11: 1841469-1841469 on Build GRCh38
6392787 c.470A>G p.D157G Chr.11: 1841472-1841472 on Build GRCh38
3446020 c.486G>A p.R162R Chr.11: 1841488-1841488 on Build GRCh38
4972580 c.541G>C p.E181Q Chr.11: 1841543-1841543 on Build GRCh38
2092801 c.471C>T p.D157D Chr.11: 1841473-1841473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780448902 Chr.11: 1841569-1841569 on Build GRCh38
rs775683835 Chr.11: 1841459-1841459 on Build GRCh38
rs761449899 Chr.11: 1841472-1841472 on Build GRCh38
rs104894312 Chr.11: 1841468-1841468 on Build GRCh38
rs770212007 Chr.11: 1841451-1841451 on Build GRCh38
rs139487112 Chr.11: 1841608-1841609 on Build GRCh38
rs767358691 Chr.11: 1841473-1841473 on Build GRCh38
rs34434911 Chr.11: 1841388-1841388 on Build GRCh38
rs531041924 Chr.11: 1841778-1841778 on Build GRCh38
rs1038968266 Chr.11: 1841651-1841651 on Build GRCh38
rs995362649 Chr.11: 1841613-1841613 on Build GRCh38
rs747284144 Chr.11: 1841426-1841426 on Build GRCh38
rs375200601 Chr.11: 1841542-1841542 on Build GRCh38
rs767038297 Chr.11: 1841408-1841408 on Build GRCh38
rs979543179 Chr.11: 1841726-1841726 on Build GRCh38
rs117830156 Chr.11: 1841645-1841645 on Build GRCh38
rs778099397 Chr.11: 1841507-1841507 on Build GRCh38
rs905581654 Chr.11: 1841633-1841633 on Build GRCh38
rs200732729 Chr.11: 1841580-1841580 on Build GRCh38
rs954117636 Chr.11: 1841737-1841737 on Build GRCh38
rs1002165169 Chr.11: 1841791-1841791 on Build GRCh38
rs1051612816 Chr.11: 1841396-1841396 on Build GRCh38
rs945588746 Chr.11: 1841511-1841511 on Build GRCh38
rs374542803 Chr.11: 1841425-1841425 on Build GRCh38
rs371341053 Chr.11: 1841474-1841474 on Build GRCh38
rs1014022018 Chr.11: 1841768-1841768 on Build GRCh38
rs555004172 Chr.11: 1841626-1841626 on Build GRCh38
rs760708666 Chr.11: 1841588-1841588 on Build GRCh38
rs527859351 Chr.11: 1841792-1841792 on Build GRCh38
rs1001708492 Chr.11: 1841635-1841635 on Build GRCh38
rs113313839 Chr.11: 1841453-1841453 on Build GRCh38
rs104894311 Chr.11: 1841523-1841523 on Build GRCh38
rs753322831 Chr.11: 1841497-1841497 on Build GRCh38
rs1013835015 Chr.11: 1841678-1841678 on Build GRCh38
rs373113911 Chr.11: 1841571-1841571 on Build GRCh38
rs536326619 Chr.11: 1841434-1841434 on Build GRCh38
rs548385691 Chr.11: 1841731-1841731 on Build GRCh38
rs374362300 Chr.11: 1841732-1841732 on Build GRCh38
rs957752971 Chr.11: 1841821-1841821 on Build GRCh38
rs913343222 Chr.11: 1841605-1841605 on Build GRCh38
rs755564422 Chr.11: 1841416-1841416 on Build GRCh38
rs911231735 Chr.11: 1841625-1841625 on Build GRCh38
rs771354182 Chr.11: 1841433-1841433 on Build GRCh38
rs559105717 Chr.11: 1841659-1841659 on Build GRCh38
rs774378823 Chr.11: 1841469-1841469 on Build GRCh38
rs781603963 Chr.11: 1841551-1841551 on Build GRCh38
rs763177134 Chr.11: 1841464-1841464 on Build GRCh38
rs571941719 Chr.11: 1841615-1841615 on Build GRCh38
rs369756656 Chr.11: 1841695-1841695 on Build GRCh38
rs751292587 Chr.11: 1841405-1841405 on Build GRCh38
rs117519669 Chr.11: 1841435-1841435 on Build GRCh38
rs765964715 Chr.11: 1841583-1841583 on Build GRCh38
rs199474800 Chr.11: 1841501-1841503 on Build GRCh38
rs966839264 Chr.11: 1841709-1841709 on Build GRCh38
rs61452009 Chr.11: 1841612-1841613 on Build GRCh38
rs771089985 Chr.11: 1841388-1841388 on Build GRCh38
rs746278905 Chr.11: 1841558-1841558 on Build GRCh38
rs767480843 Chr.11: 1841776-1841776 on Build GRCh38
rs768432453 Chr.11: 1841465-1841465 on Build GRCh38
rs199474801 Chr.11: 1841529-1841531 on Build GRCh38
rs758869533 Chr.11: 1841418-1841418 on Build GRCh38
rs113791474 Chr.11: 1841679-1841679 on Build GRCh38
rs753412639 Chr.11: 1841417-1841417 on Build GRCh38
rs761692575 Chr.11: 1841407-1841407 on Build GRCh38
rs367690402 Chr.11: 1841584-1841584 on Build GRCh38
rs960009132 Chr.11: 1841655-1841655 on Build GRCh38
rs970683318 Chr.11: 1841543-1841543 on Build GRCh38
rs758900679 Chr.11: 1841585-1841585 on Build GRCh38
rs752107981 Chr.11: 1841533-1841533 on Build GRCh38
rs1023939718 Chr.11: 1841781-1841781 on Build GRCh38
rs113592805 Chr.11: 1841786-1841786 on Build GRCh38
rs770122157 Chr.11: 1841559-1841559 on Build GRCh38
rs985955087 Chr.11: 1841762-1841762 on Build GRCh38
rs988977237 Chr.11: 1841823-1841823 on Build GRCh38
rs1036929551 Chr.11: 1841815-1841815 on Build GRCh38
rs201612866 Chr.11: 1841579-1841579 on Build GRCh38
rs797046046 Chr.11: 1841527-1841527 on Build GRCh38
rs750081987 Chr.11: 1841412-1841412 on Build GRCh38
rs543012338 Chr.11: 1841578-1841578 on Build GRCh38
rs898999134 Chr.11: 1841429-1841429 on Build GRCh38
rs766058720 Chr.11: 1841480-1841480 on Build GRCh38
rs529876148 Chr.11: 1841703-1841703 on Build GRCh38
rs761865454 Chr.11: 1841562-1841563 on Build GRCh38
rs199691669 Chr.11: 1841570-1841570 on Build GRCh38
rs979820791 Chr.11: 1841400-1841400 on Build GRCh38

View Full Product Details