Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2130143-2130650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

SNP Detail

SNP ID Chromosome Location
rs990963311 Chr.11: 2130223-2130223 on Build GRCh38
rs3180700 Chr.11: 2130589-2130589 on Build GRCh38
rs1053276420 Chr.11: 2130265-2130265 on Build GRCh38
rs964822584 Chr.11: 2130338-2130338 on Build GRCh38
rs148117489 Chr.11: 2130539-2130539 on Build GRCh38
rs1003092128 Chr.11: 2130261-2130261 on Build GRCh38
rs933206944 Chr.11: 2130175-2130175 on Build GRCh38
rs973256797 Chr.11: 2130446-2130446 on Build GRCh38
rs1053787428 Chr.11: 2130491-2130491 on Build GRCh38
rs907748176 Chr.11: 2130435-2130435 on Build GRCh38
rs538570399 Chr.11: 2130534-2130534 on Build GRCh38
rs566593592 Chr.11: 2130495-2130495 on Build GRCh38
rs35818489 Chr.11: 2130482-2130482 on Build GRCh38
rs1050713640 Chr.11: 2130268-2130268 on Build GRCh38
rs968247008 Chr.11: 2130514-2130514 on Build GRCh38
rs927650983 Chr.11: 2130462-2130462 on Build GRCh38
rs1040422996 Chr.11: 2130529-2130530 on Build GRCh38
rs1011208490 Chr.11: 2130624-2130624 on Build GRCh38
rs117403190 Chr.11: 2130611-2130611 on Build GRCh38
rs529224313 Chr.11: 2130266-2130266 on Build GRCh38
rs934958222 Chr.11: 2130484-2130484 on Build GRCh38
rs551471446 Chr.11: 2130436-2130436 on Build GRCh38
rs892040446 Chr.11: 2130492-2130492 on Build GRCh38
rs946918562 Chr.11: 2130260-2130260 on Build GRCh38
rs1046028032 Chr.11: 2130255-2130255 on Build GRCh38
rs887613391 Chr.11: 2130360-2130360 on Build GRCh38
rs1022162911 Chr.11: 2130322-2130322 on Build GRCh38
rs915373881 Chr.11: 2130469-2130469 on Build GRCh38
rs1030326132 Chr.11: 2130367-2130367 on Build GRCh38
rs556573980 Chr.11: 2130558-2130558 on Build GRCh38
rs555759341 Chr.11: 2130531-2130531 on Build GRCh38
rs539200153 Chr.11: 2130380-2130380 on Build GRCh38
rs1002805849 Chr.11: 2130599-2130599 on Build GRCh38
rs11825733 Chr.11: 2130165-2130165 on Build GRCh38
rs1011712909 Chr.11: 2130318-2130318 on Build GRCh38
rs774865092 Chr.11: 2130424-2130424 on Build GRCh38
rs1004752801 Chr.11: 2130363-2130363 on Build GRCh38
rs1043213824 Chr.11: 2130508-2130508 on Build GRCh38
rs916338622 Chr.11: 2130447-2130447 on Build GRCh38
rs543215705 Chr.11: 2130520-2130520 on Build GRCh38
rs923224306 Chr.11: 2130169-2130169 on Build GRCh38
rs954666960 Chr.11: 2130378-2130378 on Build GRCh38
rs894885170 Chr.11: 2130231-2130231 on Build GRCh38
rs924218360 Chr.11: 2130516-2130516 on Build GRCh38
rs947792248 Chr.11: 2130456-2130456 on Build GRCh38
rs907492564 Chr.11: 2130257-2130257 on Build GRCh38
rs889261883 Chr.11: 2130290-2130290 on Build GRCh38
rs72851054 Chr.11: 2130588-2130588 on Build GRCh38
rs944371986 Chr.11: 2130520-2130521 on Build GRCh38
rs143773245 Chr.11: 2130582-2130583 on Build GRCh38
rs947761387 Chr.11: 2130237-2130237 on Build GRCh38
rs900275939 Chr.11: 2130292-2130292 on Build GRCh38
rs57423851 Chr.11: 2130538-2130539 on Build GRCh38
rs550594230 Chr.11: 2130271-2130271 on Build GRCh38
rs868076786 Chr.11: 2130590-2130590 on Build GRCh38
rs770477972 Chr.11: 2130589-2130599 on Build GRCh38
rs978361513 Chr.11: 2130514-2130515 on Build GRCh38
rs368927765 Chr.11: 2130528-2130529 on Build GRCh38
rs921608086 Chr.11: 2130530-2130530 on Build GRCh38
rs113798050 Chr.11: 2130539-2130539 on Build GRCh38
rs1037861952 Chr.11: 2130203-2130203 on Build GRCh38
rs370546130 Chr.11: 2130588-2130588 on Build GRCh38
rs375769044 Chr.11: 2130496-2130496 on Build GRCh38
rs561506861 Chr.11: 2130233-2130233 on Build GRCh38
rs907715975 Chr.11: 2130185-2130185 on Build GRCh38
rs751847850 Chr.11: 2130400-2130400 on Build GRCh38
rs941852364 Chr.11: 2130191-2130191 on Build GRCh38
rs990911051 Chr.11: 2130468-2130468 on Build GRCh38
rs915426313 Chr.11: 2130250-2130250 on Build GRCh38
rs578217783 Chr.11: 2130567-2130567 on Build GRCh38
rs531894998 Chr.11: 2130399-2130399 on Build GRCh38
rs573936129 Chr.11: 2130533-2130533 on Build GRCh38
rs749385462 Chr.11: 2130487-2130487 on Build GRCh38
rs907090633 Chr.11: 2130594-2130594 on Build GRCh38
rs545576517 Chr.11: 2130592-2130592 on Build GRCh38
rs766504615 Chr.11: 2130182-2130182 on Build GRCh38
rs1036263002 Chr.11: 2130593-2130594 on Build GRCh38
rs1046101177 Chr.11: 2130457-2130457 on Build GRCh38
rs903269675 Chr.11: 2130577-2130577 on Build GRCh38
rs928066949 Chr.11: 2130181-2130181 on Build GRCh38
rs759815269 Chr.11: 2130405-2130405 on Build GRCh38
rs930878836 Chr.11: 2130259-2130259 on Build GRCh38
rs569080826 Chr.11: 2130342-2130342 on Build GRCh38
rs893162553 Chr.11: 2130620-2130620 on Build GRCh38

View Full Product Details