Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2130990-2131524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

SNP Detail

SNP ID Chromosome Location
rs527480230 Chr.11: 2131446-2131446 on Build GRCh38
rs561607562 Chr.11: 2131176-2131176 on Build GRCh38
rs924582068 Chr.11: 2131497-2131497 on Build GRCh38
rs905099875 Chr.11: 2131199-2131199 on Build GRCh38
rs1056270582 Chr.11: 2131376-2131376 on Build GRCh38
rs922220746 Chr.11: 2131253-2131253 on Build GRCh38
rs1023950326 Chr.11: 2131060-2131060 on Build GRCh38
rs967750910 Chr.11: 2131241-2131241 on Build GRCh38
rs59196953 Chr.11: 2131037-2131037 on Build GRCh38
rs930713965 Chr.11: 2131486-2131486 on Build GRCh38
rs975469460 Chr.11: 2131250-2131250 on Build GRCh38
rs369965474 Chr.11: 2131091-2131091 on Build GRCh38
rs981897718 Chr.11: 2131069-2131069 on Build GRCh38
rs930684778 Chr.11: 2131348-2131348 on Build GRCh38
rs1034807054 Chr.11: 2131217-2131217 on Build GRCh38
rs3168310 Chr.11: 2131311-2131311 on Build GRCh38
rs143209706 Chr.11: 2131461-2131461 on Build GRCh38
rs1065687 Chr.11: 2131409-2131409 on Build GRCh38
rs866654379 Chr.11: 2131428-2131428 on Build GRCh38
rs566307265 Chr.11: 2131072-2131072 on Build GRCh38
rs969206996 Chr.11: 2131067-2131067 on Build GRCh38
rs920663008 Chr.11: 2131315-2131315 on Build GRCh38
rs947306380 Chr.11: 2131102-2131102 on Build GRCh38
rs996550336 Chr.11: 2131425-2131425 on Build GRCh38
rs890580693 Chr.11: 2131154-2131154 on Build GRCh38
rs770810653 Chr.11: 2131198-2131198 on Build GRCh38
rs79275529 Chr.11: 2131029-2131029 on Build GRCh38
rs559955724 Chr.11: 2131438-2131438 on Build GRCh38
rs892316096 Chr.11: 2131377-2131377 on Build GRCh38
rs778335820 Chr.11: 2131283-2131283 on Build GRCh38
rs997661616 Chr.11: 2131187-2131187 on Build GRCh38
rs972217226 Chr.11: 2131284-2131284 on Build GRCh38
rs979053892 Chr.11: 2131104-2131104 on Build GRCh38
rs537801892 Chr.11: 2131115-2131115 on Build GRCh38
rs905152105 Chr.11: 2131362-2131362 on Build GRCh38
rs987689429 Chr.11: 2131292-2131292 on Build GRCh38
rs117486875 Chr.11: 2131347-2131347 on Build GRCh38
rs915763184 Chr.11: 2131310-2131310 on Build GRCh38
rs988246121 Chr.11: 2131092-2131092 on Build GRCh38
rs901899726 Chr.11: 2131385-2131385 on Build GRCh38
rs1031117398 Chr.11: 2131143-2131143 on Build GRCh38
rs912068745 Chr.11: 2131306-2131306 on Build GRCh38
rs79531557 Chr.11: 2131414-2131414 on Build GRCh38
rs1022438932 Chr.11: 2131236-2131236 on Build GRCh38
rs1003421527 Chr.11: 2131372-2131372 on Build GRCh38
rs972623695 Chr.11: 2131453-2131453 on Build GRCh38
rs1045041690 Chr.11: 2131356-2131356 on Build GRCh38
rs554442693 Chr.11: 2131237-2131237 on Build GRCh38
rs567611973 Chr.11: 2131499-2131499 on Build GRCh38
rs928434473 Chr.11: 2131289-2131289 on Build GRCh38
rs1003370611 Chr.11: 2131216-2131216 on Build GRCh38
rs3208122 Chr.11: 2131361-2131361 on Build GRCh38
rs58527086 Chr.11: 2131105-2131105 on Build GRCh38
rs762205555 Chr.11: 2131498-2131499 on Build GRCh38
rs749731671 Chr.11: 2131056-2131056 on Build GRCh38
rs555123166 Chr.11: 2131322-2131322 on Build GRCh38
rs1045093214 Chr.11: 2131488-2131488 on Build GRCh38
rs533083978 Chr.11: 2131422-2131422 on Build GRCh38
rs914918792 Chr.11: 2131093-2131093 on Build GRCh38
rs1035775664 Chr.11: 2131444-2131453 on Build GRCh38
rs572669571 Chr.11: 2131270-2131270 on Build GRCh38
rs915742054 Chr.11: 2131466-2131466 on Build GRCh38
rs567696295 Chr.11: 2131109-2131109 on Build GRCh38
rs149099319 Chr.11: 2131188-2131188 on Build GRCh38
rs762624032 Chr.11: 2131332-2131333 on Build GRCh38
rs997575331 Chr.11: 2131401-2131401 on Build GRCh38
rs961776820 Chr.11: 2131251-2131251 on Build GRCh38
rs932164190 Chr.11: 2131114-2131114 on Build GRCh38
rs181654351 Chr.11: 2131096-2131096 on Build GRCh38
rs940795018 Chr.11: 2131155-2131155 on Build GRCh38
rs374392622 Chr.11: 2131427-2131427 on Build GRCh38
rs529166030 Chr.11: 2131220-2131220 on Build GRCh38
rs987909416 Chr.11: 2131223-2131223 on Build GRCh38
rs544886527 Chr.11: 2131437-2131437 on Build GRCh38
rs925275681 Chr.11: 2131360-2131360 on Build GRCh38
rs920730056 Chr.11: 2131475-2131475 on Build GRCh38
rs940831512 Chr.11: 2131308-2131308 on Build GRCh38
rs200159614 Chr.11: 2131440-2131441 on Build GRCh38
rs1043032212 Chr.11: 2131333-2131333 on Build GRCh38
rs991237943 Chr.11: 2131455-2131455 on Build GRCh38
rs746965737 Chr.11: 2131175-2131175 on Build GRCh38
rs985015689 Chr.11: 2131219-2131219 on Build GRCh38
rs1016555143 Chr.11: 2131428-2131429 on Build GRCh38
rs556823779 Chr.11: 2131140-2131140 on Build GRCh38

View Full Product Details