Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64341507-64342008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88B
Entrez Gene ID
283234
Gene Name
coiled-coil domain containing 88B
Gene Aliases
BRLZ, CCDC88, HKRP3, gipie
UniGene
Hs.98564
Gene Chromosome Location
Chr.11: 64340203-64357534 on Build GRCh38
Transcripts
NM_032251.5 XM_006718519.3 XM_011544957.2 XM_011544959.2 XM_011544960.2 XM_017017601.1 NR_106977.1

Gene Symbol
MIR7155
Entrez Gene ID
102466815
Gene Name
microRNA 7155
Gene Aliases
hsa-mir-7155
UniGene
Gene Chromosome Location
Chr.11: 64341849-64341904 on Build GRCh38
Transcripts
NM_032251.5 XM_006718519.3 XM_011544957.2 XM_011544959.2 XM_011544960.2 XM_017017601.1 NR_106977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035114 c.609G>A p.E203E Chr.11: 64341676-64341676 on Build GRCh38
1628148 c.680T>G p.L227R Chr.11: 64341998-64341998 on Build GRCh38
2163477 c.659G>A p.R220H Chr.11: 64341726-64341726 on Build GRCh38
7113513 c.655G>C p.E219Q Chr.11: 64341722-64341722 on Build GRCh38
429535 c.579T>G p.D193E Chr.11: 64341646-64341646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777511119 Chr.11: 64341938-64341938 on Build GRCh38
rs779596149 Chr.11: 64341759-64341759 on Build GRCh38
rs375096255 Chr.11: 64341582-64341582 on Build GRCh38
rs201049666 Chr.11: 64341923-64341923 on Build GRCh38
rs748064027 Chr.11: 64341912-64341912 on Build GRCh38
rs745889074 Chr.11: 64341958-64341958 on Build GRCh38
rs753340392 Chr.11: 64341897-64341897 on Build GRCh38
rs913331223 Chr.11: 64341882-64341882 on Build GRCh38
rs761193202 Chr.11: 64341618-64341618 on Build GRCh38
rs141970879 Chr.11: 64341686-64341686 on Build GRCh38
rs944321117 Chr.11: 64341945-64341945 on Build GRCh38
rs773481138 Chr.11: 64341787-64341787 on Build GRCh38
rs868434614 Chr.11: 64341935-64341935 on Build GRCh38
rs200216096 Chr.11: 64341594-64341594 on Build GRCh38
rs986501412 Chr.11: 64341768-64341768 on Build GRCh38
rs777354647 Chr.11: 64341557-64341557 on Build GRCh38
rs766467406 Chr.11: 64341719-64341719 on Build GRCh38
rs569639786 Chr.11: 64341821-64341821 on Build GRCh38
rs749216813 Chr.11: 64341677-64341680 on Build GRCh38
rs532027601 Chr.11: 64341670-64341670 on Build GRCh38
rs191336693 Chr.11: 64341715-64341715 on Build GRCh38
rs763481810 Chr.11: 64341846-64341846 on Build GRCh38
rs375252275 Chr.11: 64341962-64341962 on Build GRCh38
rs768438535 Chr.11: 64341848-64341855 on Build GRCh38
rs774166525 Chr.11: 64341690-64341690 on Build GRCh38
rs770612437 Chr.11: 64341565-64341565 on Build GRCh38
rs563193264 Chr.11: 64341659-64341659 on Build GRCh38
rs138265263 Chr.11: 64341626-64341626 on Build GRCh38
rs747260415 Chr.11: 64341940-64341940 on Build GRCh38
rs375914174 Chr.11: 64341785-64341785 on Build GRCh38
rs367964927 Chr.11: 64341610-64341610 on Build GRCh38
rs930139730 Chr.11: 64341796-64341796 on Build GRCh38
rs1036019748 Chr.11: 64341696-64341696 on Build GRCh38
rs143094813 Chr.11: 64341644-64341644 on Build GRCh38
rs142786551 Chr.11: 64341599-64341599 on Build GRCh38
rs756927022 Chr.11: 64341651-64341651 on Build GRCh38
rs184592238 Chr.11: 64341833-64341833 on Build GRCh38
rs529852829 Chr.11: 64341757-64341757 on Build GRCh38
rs755763233 Chr.11: 64341853-64341853 on Build GRCh38
rs750152063 Chr.11: 64341852-64341852 on Build GRCh38
rs647152 Chr.11: 64341646-64341646 on Build GRCh38
rs775998314 Chr.11: 64341844-64341844 on Build GRCh38
rs201623170 Chr.11: 64341949-64341949 on Build GRCh38
rs779925965 Chr.11: 64341682-64341682 on Build GRCh38
rs370989286 Chr.11: 64341552-64341552 on Build GRCh38
rs371949923 Chr.11: 64341960-64341960 on Build GRCh38
rs758236358 Chr.11: 64341924-64341924 on Build GRCh38
rs758073998 Chr.11: 64341556-64341556 on Build GRCh38
rs765548160 Chr.11: 64341725-64341725 on Build GRCh38
rs200711256 Chr.11: 64341588-64341588 on Build GRCh38
rs558172037 Chr.11: 64341850-64341850 on Build GRCh38
rs201011301 Chr.11: 64341591-64341591 on Build GRCh38
rs367832758 Chr.11: 64341720-64341720 on Build GRCh38
rs199673065 Chr.11: 64341986-64341986 on Build GRCh38
rs756583196 Chr.11: 64341532-64341532 on Build GRCh38
rs903048027 Chr.11: 64341685-64341685 on Build GRCh38
rs139000172 Chr.11: 64341676-64341676 on Build GRCh38
rs549548519 Chr.11: 64341809-64341809 on Build GRCh38
rs61735121 Chr.11: 64341736-64341736 on Build GRCh38
rs750462536 Chr.11: 64341613-64341613 on Build GRCh38
rs766805380 Chr.11: 64341621-64341621 on Build GRCh38
rs780428012 Chr.11: 64341550-64341550 on Build GRCh38
rs774818594 Chr.11: 64341973-64341973 on Build GRCh38
rs779903561 Chr.11: 64341564-64341589 on Build GRCh38
rs1057402966 Chr.11: 64341911-64341911 on Build GRCh38
rs747881574 Chr.11: 64341784-64341784 on Build GRCh38
rs774217650 Chr.11: 64341851-64341859 on Build GRCh38
rs746657628 Chr.11: 64341563-64341563 on Build GRCh38
rs762002647 Chr.11: 64341609-64341609 on Build GRCh38
rs754884236 Chr.11: 64341898-64341898 on Build GRCh38
rs770954831 Chr.11: 64341800-64341800 on Build GRCh38
rs765838859 Chr.11: 64341855-64341855 on Build GRCh38
rs556067934 Chr.11: 64341564-64341566 on Build GRCh38
rs762055626 Chr.11: 64341698-64341698 on Build GRCh38
rs763534779 Chr.11: 64341964-64341964 on Build GRCh38
rs372654482 Chr.11: 64341755-64341755 on Build GRCh38
rs745316100 Chr.11: 64341661-64341661 on Build GRCh38
rs778974800 Chr.11: 64341900-64341900 on Build GRCh38
rs776583606 Chr.11: 64341952-64341952 on Build GRCh38
rs755746224 Chr.11: 64341758-64341758 on Build GRCh38
rs767696733 Chr.11: 64341574-64341574 on Build GRCh38
rs750535053 Chr.11: 64341575-64341575 on Build GRCh38
rs747881598 Chr.11: 64341852-64341852 on Build GRCh38
rs79183699 Chr.11: 64341688-64341688 on Build GRCh38
rs747209129 Chr.11: 64341790-64341790 on Build GRCh38
rs749225752 Chr.11: 64341765-64341765 on Build GRCh38
rs768599756 Chr.11: 64341766-64341766 on Build GRCh38
rs778596045 Chr.11: 64341772-64341772 on Build GRCh38
rs764595680 Chr.11: 64341967-64341967 on Build GRCh38
rs372217739 Chr.11: 64341708-64341708 on Build GRCh38

View Full Product Details