Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 72792814-72793189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHQ1
Entrez Gene ID
55164
Gene Name
SHQ1, H/ACA ribonucleoprotein assembly factor
Gene Aliases
GRIM-1, Shq1p
UniGene
Hs.606584
Gene Chromosome Location
Chr.3: 72725703-72848475 on Build GRCh38
Transcripts
NM_018130.2 XM_011533895.2 XM_011533896.2 XM_011533897.2 XM_011533898.2 XM_017006724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6778012 c.1091A>G p.D364G Chr.3: 72793006-72793006 on Build GRCh38
6926991 c.1124C>T p.A375V Chr.3: 72792973-72792973 on Build GRCh38
6980622 c.1138G>T p.D380Y Chr.3: 72792959-72792959 on Build GRCh38
6970947 c.1096C>G p.H366D Chr.3: 72793001-72793001 on Build GRCh38
6972790 c.1157A>G p.Y386C Chr.3: 72792940-72792940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767279001 Chr.3: 72792961-72792961 on Build GRCh38
rs753606311 Chr.3: 72792881-72792881 on Build GRCh38
rs941586324 Chr.3: 72793061-72793061 on Build GRCh38
rs773093678 Chr.3: 72792963-72792963 on Build GRCh38
rs778666293 Chr.3: 72792890-72792890 on Build GRCh38
rs776668832 Chr.3: 72792935-72792935 on Build GRCh38
rs750601853 Chr.3: 72792892-72792895 on Build GRCh38
rs111329127 Chr.3: 72793143-72793143 on Build GRCh38
rs758942456 Chr.3: 72793049-72793050 on Build GRCh38
rs765349388 Chr.3: 72792892-72792892 on Build GRCh38
rs936881136 Chr.3: 72793119-72793119 on Build GRCh38
rs778144069 Chr.3: 72792896-72792896 on Build GRCh38
rs71300549 Chr.3: 72793120-72793120 on Build GRCh38
rs541820371 Chr.3: 72793114-72793114 on Build GRCh38
rs35869395 Chr.3: 72792863-72792863 on Build GRCh38
rs75548717 Chr.3: 72792892-72792892 on Build GRCh38
rs887278085 Chr.3: 72793049-72793049 on Build GRCh38
rs184053800 Chr.3: 72792893-72792893 on Build GRCh38
rs775643341 Chr.3: 72792954-72792954 on Build GRCh38
rs905424154 Chr.3: 72793112-72793112 on Build GRCh38
rs377683457 Chr.3: 72793067-72793067 on Build GRCh38
rs770973555 Chr.3: 72792915-72792915 on Build GRCh38
rs772298278 Chr.3: 72793121-72793121 on Build GRCh38
rs534973484 Chr.3: 72792972-72792972 on Build GRCh38
rs577488660 Chr.3: 72792964-72792964 on Build GRCh38
rs759793149 Chr.3: 72792940-72792940 on Build GRCh38
rs191813552 Chr.3: 72792885-72792885 on Build GRCh38
rs369246048 Chr.3: 72793047-72793047 on Build GRCh38
rs778185754 Chr.3: 72793048-72793048 on Build GRCh38
rs553343230 Chr.3: 72792987-72792987 on Build GRCh38
rs754485995 Chr.3: 72792997-72792997 on Build GRCh38
rs111287698 Chr.3: 72793141-72793141 on Build GRCh38
rs747546466 Chr.3: 72792904-72792904 on Build GRCh38
rs574698015 Chr.3: 72793084-72793084 on Build GRCh38
rs916882248 Chr.3: 72792950-72792950 on Build GRCh38
rs764990247 Chr.3: 72793016-72793016 on Build GRCh38
rs929036333 Chr.3: 72792891-72792891 on Build GRCh38
rs754326459 Chr.3: 72792952-72792952 on Build GRCh38
rs752327401 Chr.3: 72793027-72793027 on Build GRCh38
rs187502298 Chr.3: 72792942-72792942 on Build GRCh38
rs189902165 Chr.3: 72793154-72793154 on Build GRCh38
rs563483454 Chr.3: 72793133-72793133 on Build GRCh38
rs761610326 Chr.3: 72792959-72792959 on Build GRCh38

View Full Product Details