Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114678391-114678902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE2
Entrez Gene ID
120406
Gene Name
neurexophilin and PC-esterase domain family member 2
Gene Aliases
FAM55B
UniGene
Hs.446760
Gene Chromosome Location
Chr.11: 114464276-114813702 on Build GRCh38
Transcripts
NM_182495.5 XM_011542604.2 XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 XM_017017211.1 XM_017017212.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5590894 c.21C>T p.I7I Chr.11: 114678596-114678596 on Build GRCh38
7259590 c.19A>G p.I7V Chr.11: 114678594-114678594 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757388748 Chr.11: 114678852-114678853 on Build GRCh38
rs970609099 Chr.11: 114678440-114678440 on Build GRCh38
rs529609139 Chr.11: 114678637-114678637 on Build GRCh38
rs944779159 Chr.11: 114678575-114678575 on Build GRCh38
rs932506854 Chr.11: 114678598-114678598 on Build GRCh38
rs747530357 Chr.11: 114678636-114678636 on Build GRCh38
rs986954538 Chr.11: 114678571-114678571 on Build GRCh38
rs976096585 Chr.11: 114678858-114678858 on Build GRCh38
rs777304576 Chr.11: 114678602-114678602 on Build GRCh38
rs774991633 Chr.11: 114678569-114678569 on Build GRCh38
rs564599466 Chr.11: 114678719-114678719 on Build GRCh38
rs987457709 Chr.11: 114678751-114678751 on Build GRCh38
rs543768451 Chr.11: 114678850-114678851 on Build GRCh38
rs778550381 Chr.11: 114678607-114678607 on Build GRCh38
rs969818201 Chr.11: 114678684-114678684 on Build GRCh38
rs917429797 Chr.11: 114678438-114678438 on Build GRCh38
rs756052709 Chr.11: 114678595-114678595 on Build GRCh38
rs564976285 Chr.11: 114678421-114678421 on Build GRCh38
rs527377127 Chr.11: 114678439-114678439 on Build GRCh38
rs930862064 Chr.11: 114678528-114678528 on Build GRCh38
rs920871490 Chr.11: 114678520-114678520 on Build GRCh38
rs1040821831 Chr.11: 114678585-114678585 on Build GRCh38
rs1047797294 Chr.11: 114678553-114678553 on Build GRCh38
rs1031295726 Chr.11: 114678415-114678415 on Build GRCh38
rs1049563246 Chr.11: 114678603-114678603 on Build GRCh38
rs867909566 Chr.11: 114678497-114678497 on Build GRCh38
rs935307485 Chr.11: 114678606-114678606 on Build GRCh38
rs769500808 Chr.11: 114678521-114678521 on Build GRCh38
rs547118600 Chr.11: 114678441-114678441 on Build GRCh38
rs137967033 Chr.11: 114678851-114678852 on Build GRCh38
rs955824964 Chr.11: 114678416-114678416 on Build GRCh38
rs559098184 Chr.11: 114678846-114678846 on Build GRCh38
rs993809453 Chr.11: 114678741-114678741 on Build GRCh38
rs767537650 Chr.11: 114678741-114678741 on Build GRCh38
rs1016551523 Chr.11: 114678542-114678542 on Build GRCh38
rs77578036 Chr.11: 114678694-114678694 on Build GRCh38
rs972420546 Chr.11: 114678495-114678495 on Build GRCh38
rs918812737 Chr.11: 114678508-114678508 on Build GRCh38
rs756854265 Chr.11: 114678604-114678604 on Build GRCh38
rs140539487 Chr.11: 114678618-114678618 on Build GRCh38
rs746210513 Chr.11: 114678480-114678480 on Build GRCh38
rs964546883 Chr.11: 114678796-114678797 on Build GRCh38
rs901024454 Chr.11: 114678588-114678588 on Build GRCh38
rs746165207 Chr.11: 114678612-114678612 on Build GRCh38
rs777601575 Chr.11: 114678644-114678644 on Build GRCh38
rs1001396608 Chr.11: 114678667-114678667 on Build GRCh38
rs184364159 Chr.11: 114678653-114678653 on Build GRCh38
rs1027820616 Chr.11: 114678717-114678717 on Build GRCh38
rs944665971 Chr.11: 114678845-114678845 on Build GRCh38
rs935423338 Chr.11: 114678849-114678850 on Build GRCh38
rs773178893 Chr.11: 114678791-114678791 on Build GRCh38
rs980391076 Chr.11: 114678476-114678476 on Build GRCh38
rs77452963 Chr.11: 114678747-114678747 on Build GRCh38
rs1044181326 Chr.11: 114678601-114678601 on Build GRCh38
rs1014025789 Chr.11: 114678661-114678661 on Build GRCh38
rs942921466 Chr.11: 114678589-114678589 on Build GRCh38
rs891425689 Chr.11: 114678660-114678660 on Build GRCh38
rs767279182 Chr.11: 114678552-114678552 on Build GRCh38
rs907878467 Chr.11: 114678762-114678762 on Build GRCh38
rs1024443999 Chr.11: 114678682-114678682 on Build GRCh38
rs1045681497 Chr.11: 114678655-114678655 on Build GRCh38
rs533178680 Chr.11: 114678868-114678868 on Build GRCh38
rs748912037 Chr.11: 114678614-114678614 on Build GRCh38
rs781559945 Chr.11: 114678418-114678418 on Build GRCh38

View Full Product Details