Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118532063-118532553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM25
Entrez Gene ID
84866
Gene Name
transmembrane protein 25
Gene Aliases
-
UniGene
Hs.564188
Gene Chromosome Location
Chr.11: 118531088-118546670 on Build GRCh38
Transcripts
NM_001144034.1 NM_001144035.1 NM_001144036.1 NM_001144037.1 NM_001144038.1 NM_001318755.1 NM_001318757.1 NM_032780.3 NR_134852.1 XM_005271703.1 XM_006718926.2 XM_006718927.2 XM_011543037.2 XM_011543038.2 XM_017018422.1 XM_017018423.1 XM_017018424.1 XM_017018425.1 XM_017018426.1 XM_017018427.1 XM_017018428.1 XM_017018429.1 XM_017018430.1 XM_017018431.1 XM_017018432.1 XM_017018433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4333853 c.166delG p.P58fs*23 Chr.11: 118532245-118532245 on Build GRCh38
6803938 c.243C>T p.S81S Chr.11: 118532322-118532322 on Build GRCh38
5499119 c.358G>A p.A120T Chr.11: 118532437-118532437 on Build GRCh38
1604174 c.92A>G p.Q31R Chr.11: 118532171-118532171 on Build GRCh38
4018360 c.211C>T p.L71L Chr.11: 118532290-118532290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781809397 Chr.11: 118532174-118532174 on Build GRCh38
rs782188288 Chr.11: 118532224-118532224 on Build GRCh38
rs782611195 Chr.11: 118532352-118532352 on Build GRCh38
rs782472733 Chr.11: 118532265-118532265 on Build GRCh38
rs367682925 Chr.11: 118532179-118532179 on Build GRCh38
rs782579566 Chr.11: 118532476-118532476 on Build GRCh38
rs781958177 Chr.11: 118532367-118532368 on Build GRCh38
rs782182534 Chr.11: 118532481-118532481 on Build GRCh38
rs782687208 Chr.11: 118532178-118532178 on Build GRCh38
rs781919370 Chr.11: 118532290-118532290 on Build GRCh38
rs377348449 Chr.11: 118532206-118532206 on Build GRCh38
rs570450435 Chr.11: 118532198-118532198 on Build GRCh38
rs781965949 Chr.11: 118532375-118532375 on Build GRCh38
rs200390061 Chr.11: 118532218-118532218 on Build GRCh38
rs936857118 Chr.11: 118532532-118532532 on Build GRCh38
rs572377333 Chr.11: 118532237-118532237 on Build GRCh38
rs374703242 Chr.11: 118532363-118532363 on Build GRCh38
rs146752919 Chr.11: 118532354-118532354 on Build GRCh38
rs576036583 Chr.11: 118532506-118532506 on Build GRCh38
rs781944339 Chr.11: 118532311-118532311 on Build GRCh38
rs551854493 Chr.11: 118532157-118532157 on Build GRCh38
rs931963683 Chr.11: 118532500-118532500 on Build GRCh38
rs781900792 Chr.11: 118532258-118532258 on Build GRCh38
rs781827618 Chr.11: 118532399-118532399 on Build GRCh38
rs1024681771 Chr.11: 118532104-118532104 on Build GRCh38
rs781911808 Chr.11: 118532329-118532329 on Build GRCh38
rs137861034 Chr.11: 118532340-118532340 on Build GRCh38
rs782482328 Chr.11: 118532349-118532349 on Build GRCh38
rs371160569 Chr.11: 118532336-118532336 on Build GRCh38
rs782662360 Chr.11: 118532483-118532483 on Build GRCh38
rs782420838 Chr.11: 118532225-118532225 on Build GRCh38
rs782754802 Chr.11: 118532299-118532299 on Build GRCh38
rs782227773 Chr.11: 118532443-118532443 on Build GRCh38
rs782540962 Chr.11: 118532332-118532332 on Build GRCh38
rs782802854 Chr.11: 118532426-118532426 on Build GRCh38
rs969074927 Chr.11: 118532137-118532137 on Build GRCh38
rs782789593 Chr.11: 118532509-118532509 on Build GRCh38
rs368395608 Chr.11: 118532323-118532323 on Build GRCh38
rs782617413 Chr.11: 118532271-118532271 on Build GRCh38
rs781848450 Chr.11: 118532261-118532261 on Build GRCh38
rs781862145 Chr.11: 118532343-118532343 on Build GRCh38
rs781925006 Chr.11: 118532093-118532093 on Build GRCh38
rs377709210 Chr.11: 118532492-118532492 on Build GRCh38
rs782164816 Chr.11: 118532381-118532381 on Build GRCh38
rs782448032 Chr.11: 118532425-118532425 on Build GRCh38
rs11553524 Chr.11: 118532236-118532236 on Build GRCh38
rs782045503 Chr.11: 118532283-118532283 on Build GRCh38
rs782628004 Chr.11: 118532281-118532281 on Build GRCh38
rs368759722 Chr.11: 118532373-118532373 on Build GRCh38
rs965324328 Chr.11: 118532457-118532457 on Build GRCh38
rs782697443 Chr.11: 118532250-118532250 on Build GRCh38
rs782032713 Chr.11: 118532502-118532502 on Build GRCh38
rs781968336 Chr.11: 118532242-118532242 on Build GRCh38
rs976049417 Chr.11: 118532223-118532223 on Build GRCh38
rs150904224 Chr.11: 118532377-118532377 on Build GRCh38
rs782198287 Chr.11: 118532522-118532522 on Build GRCh38
rs782764269 Chr.11: 118532260-118532260 on Build GRCh38
rs201041295 Chr.11: 118532357-118532357 on Build GRCh38
rs138350313 Chr.11: 118532436-118532436 on Build GRCh38
rs782295341 Chr.11: 118532293-118532293 on Build GRCh38
rs781881619 Chr.11: 118532184-118532184 on Build GRCh38
rs543452387 Chr.11: 118532510-118532510 on Build GRCh38
rs782165117 Chr.11: 118532112-118532112 on Build GRCh38
rs370682545 Chr.11: 118532278-118532278 on Build GRCh38
rs782405715 Chr.11: 118532294-118532294 on Build GRCh38
rs782332976 Chr.11: 118532287-118532288 on Build GRCh38
rs375061841 Chr.11: 118532284-118532284 on Build GRCh38
rs35915434 Chr.11: 118532154-118532154 on Build GRCh38
rs782370463 Chr.11: 118532374-118532374 on Build GRCh38
rs561840081 Chr.11: 118532511-118532511 on Build GRCh38
rs782112229 Chr.11: 118532297-118532297 on Build GRCh38
rs367564776 Chr.11: 118532461-118532461 on Build GRCh38
rs908427255 Chr.11: 118532209-118532209 on Build GRCh38
rs782619881 Chr.11: 118532200-118532200 on Build GRCh38
rs952634150 Chr.11: 118532172-118532172 on Build GRCh38
rs782689375 Chr.11: 118532274-118532274 on Build GRCh38
rs782802438 Chr.11: 118532385-118532385 on Build GRCh38
rs782003975 Chr.11: 118532388-118532388 on Build GRCh38
rs782264747 Chr.11: 118532484-118532484 on Build GRCh38
rs782441594 Chr.11: 118532482-118532482 on Build GRCh38
rs782436805 Chr.11: 118532177-118532177 on Build GRCh38
rs149608668 Chr.11: 118532437-118532437 on Build GRCh38
rs782729825 Chr.11: 118532162-118532162 on Build GRCh38
rs782404272 Chr.11: 118532152-118532153 on Build GRCh38
rs925336323 Chr.11: 118532527-118532527 on Build GRCh38
rs782029967 Chr.11: 118532281-118532282 on Build GRCh38
rs782351469 Chr.11: 118532207-118532207 on Build GRCh38
rs371764377 Chr.11: 118532322-118532322 on Build GRCh38
rs374774013 Chr.11: 118532438-118532438 on Build GRCh38
rs782321954 Chr.11: 118532285-118532285 on Build GRCh38
rs782282864 Chr.11: 118532276-118532276 on Build GRCh38
rs782118346 Chr.11: 118532158-118532158 on Build GRCh38

View Full Product Details