Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119342518-119343020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF5
Entrez Gene ID
114902
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
CTRP5
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_015645.4 NM_031433.3

Gene Symbol
MFRP
Entrez Gene ID
83552
Gene Name
membrane frizzled-related protein
Gene Aliases
MCOP5, NNO2, RD6
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_015645.4 NM_031433.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346294 c.1257C>A p.N419K Chr.11: 119342726-119342726 on Build GRCh38
6874293 c.1211G>A p.S404N Chr.11: 119342917-119342917 on Build GRCh38
4186963 c.1333G>T p.D445Y Chr.11: 119342650-119342650 on Build GRCh38
5926775 c.1384C>T p.P462S Chr.11: 119342599-119342599 on Build GRCh38
1561578 c.1365C>T p.S455S Chr.11: 119342618-119342618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761110898 Chr.11: 119342799-119342799 on Build GRCh38
rs562312824 Chr.11: 119342943-119342943 on Build GRCh38
rs760263410 Chr.11: 119342990-119342990 on Build GRCh38
rs779992611 Chr.11: 119342911-119342911 on Build GRCh38
rs587776595 Chr.11: 119342978-119342979 on Build GRCh38
rs781215218 Chr.11: 119342823-119342823 on Build GRCh38
rs748551210 Chr.11: 119342545-119342545 on Build GRCh38
rs750070481 Chr.11: 119342972-119342972 on Build GRCh38
rs754780011 Chr.11: 119342994-119342994 on Build GRCh38
rs369531002 Chr.11: 119342617-119342617 on Build GRCh38
rs758398734 Chr.11: 119342980-119342980 on Build GRCh38
rs908938174 Chr.11: 119342919-119342919 on Build GRCh38
rs574601214 Chr.11: 119342642-119342642 on Build GRCh38
rs891121482 Chr.11: 119342798-119342798 on Build GRCh38
rs927210885 Chr.11: 119342960-119342960 on Build GRCh38
rs371250038 Chr.11: 119342777-119342777 on Build GRCh38
rs764467122 Chr.11: 119342843-119342844 on Build GRCh38
rs754348823 Chr.11: 119342710-119342710 on Build GRCh38
rs1051134777 Chr.11: 119342784-119342784 on Build GRCh38
rs371946990 Chr.11: 119342898-119342898 on Build GRCh38
rs199608492 Chr.11: 119342756-119342756 on Build GRCh38
rs139725672 Chr.11: 119342638-119342638 on Build GRCh38
rs764203618 Chr.11: 119342978-119342978 on Build GRCh38
rs757712621 Chr.11: 119342978-119342978 on Build GRCh38
rs937108029 Chr.11: 119342686-119342686 on Build GRCh38
rs191660790 Chr.11: 119342690-119342690 on Build GRCh38
rs772644068 Chr.11: 119342952-119342952 on Build GRCh38
rs958347798 Chr.11: 119342897-119342897 on Build GRCh38
rs369692906 Chr.11: 119342751-119342751 on Build GRCh38
rs767672258 Chr.11: 119342878-119342878 on Build GRCh38
rs367859225 Chr.11: 119342868-119342868 on Build GRCh38
rs775425747 Chr.11: 119342639-119342639 on Build GRCh38
rs769780473 Chr.11: 119342839-119342839 on Build GRCh38
rs1006954954 Chr.11: 119342813-119342813 on Build GRCh38
rs762676918 Chr.11: 119342610-119342610 on Build GRCh38
rs548201441 Chr.11: 119342560-119342560 on Build GRCh38
rs905681288 Chr.11: 119342737-119342737 on Build GRCh38
rs926007264 Chr.11: 119342829-119342829 on Build GRCh38
rs770467284 Chr.11: 119342551-119342551 on Build GRCh38
rs759830697 Chr.11: 119342965-119342965 on Build GRCh38
rs139684307 Chr.11: 119342910-119342910 on Build GRCh38
rs145881139 Chr.11: 119342609-119342609 on Build GRCh38
rs377600522 Chr.11: 119342778-119342778 on Build GRCh38
rs759321899 Chr.11: 119342755-119342755 on Build GRCh38
rs368782877 Chr.11: 119342935-119342935 on Build GRCh38
rs747235836 Chr.11: 119342733-119342733 on Build GRCh38
rs755623213 Chr.11: 119342699-119342699 on Build GRCh38
rs767917702 Chr.11: 119342725-119342725 on Build GRCh38
rs759109552 Chr.11: 119342669-119342669 on Build GRCh38
rs764605750 Chr.11: 119342964-119342964 on Build GRCh38
rs754311603 Chr.11: 119342976-119342977 on Build GRCh38
rs745827185 Chr.11: 119342846-119342846 on Build GRCh38
rs752415500 Chr.11: 119342695-119342695 on Build GRCh38
rs142846614 Chr.11: 119342641-119342641 on Build GRCh38
rs752871438 Chr.11: 119342889-119342889 on Build GRCh38
rs991249582 Chr.11: 119342917-119342917 on Build GRCh38
rs775475230 Chr.11: 119342851-119342851 on Build GRCh38
rs561701695 Chr.11: 119342566-119342566 on Build GRCh38
rs766909293 Chr.11: 119342618-119342618 on Build GRCh38
rs765828336 Chr.11: 119342772-119342772 on Build GRCh38
rs377761571 Chr.11: 119342776-119342776 on Build GRCh38
rs774247976 Chr.11: 119342668-119342668 on Build GRCh38
rs988161323 Chr.11: 119342539-119342539 on Build GRCh38
rs766936271 Chr.11: 119342961-119342961 on Build GRCh38
rs529813598 Chr.11: 119342959-119342959 on Build GRCh38
rs149762583 Chr.11: 119342988-119342988 on Build GRCh38
rs746309815 Chr.11: 119342726-119342726 on Build GRCh38
rs556189272 Chr.11: 119342619-119342619 on Build GRCh38
rs749732936 Chr.11: 119342570-119342570 on Build GRCh38
rs201420631 Chr.11: 119342672-119342672 on Build GRCh38
rs11217241 Chr.11: 119342593-119342593 on Build GRCh38
rs669462 Chr.11: 119342795-119342795 on Build GRCh38
rs374018417 Chr.11: 119342758-119342758 on Build GRCh38
rs771945249 Chr.11: 119342850-119342850 on Build GRCh38
rs762905998 Chr.11: 119342645-119342645 on Build GRCh38
rs1017825378 Chr.11: 119342801-119342801 on Build GRCh38
rs766197375 Chr.11: 119342612-119342612 on Build GRCh38
rs771999204 Chr.11: 119342584-119342584 on Build GRCh38
rs142533439 Chr.11: 119342731-119342731 on Build GRCh38
rs756339504 Chr.11: 119342973-119342973 on Build GRCh38
rs763730179 Chr.11: 119342703-119342703 on Build GRCh38
rs756183471 Chr.11: 119342890-119342890 on Build GRCh38
rs774312463 Chr.11: 119342631-119342631 on Build GRCh38
rs764657694 Chr.11: 119342863-119342863 on Build GRCh38
rs749155264 Chr.11: 119342744-119342744 on Build GRCh38
rs376122111 Chr.11: 119342844-119342844 on Build GRCh38
rs765223589 Chr.11: 119342608-119342608 on Build GRCh38
rs775243867 Chr.11: 119342757-119342757 on Build GRCh38
rs370286787 Chr.11: 119342857-119342857 on Build GRCh38
rs777738320 Chr.11: 119342891-119342891 on Build GRCh38
rs564824660 Chr.11: 119342789-119342789 on Build GRCh38
rs771438559 Chr.11: 119342571-119342571 on Build GRCh38
rs769294778 Chr.11: 119342948-119342948 on Build GRCh38
rs1011833093 Chr.11: 119342996-119342996 on Build GRCh38
rs763388417 Chr.11: 119342866-119342866 on Build GRCh38
rs748321183 Chr.11: 119342825-119342825 on Build GRCh38
rs776266547 Chr.11: 119342860-119342860 on Build GRCh38
rs372817789 Chr.11: 119342632-119342632 on Build GRCh38
rs974233404 Chr.11: 119342951-119342951 on Build GRCh38
rs757833183 Chr.11: 119342716-119342716 on Build GRCh38
rs374259317 Chr.11: 119342923-119342923 on Build GRCh38
rs950121355 Chr.11: 119342902-119342902 on Build GRCh38
rs750907785 Chr.11: 119342775-119342775 on Build GRCh38
rs767903765 Chr.11: 119342966-119342966 on Build GRCh38
rs752919228 Chr.11: 119342968-119342968 on Build GRCh38
rs752936411 Chr.11: 119342827-119342827 on Build GRCh38
rs767900062 Chr.11: 119342649-119342650 on Build GRCh38
rs199473711 Chr.11: 119342839-119342840 on Build GRCh38
rs770971634 Chr.11: 119342745-119342745 on Build GRCh38
rs572431639 Chr.11: 119342770-119342770 on Build GRCh38
rs751958579 Chr.11: 119342876-119342876 on Build GRCh38
rs560280998 Chr.11: 119342743-119342743 on Build GRCh38
rs371437024 Chr.11: 119342576-119342576 on Build GRCh38
rs377135056 Chr.11: 119342637-119342637 on Build GRCh38
rs373984029 Chr.11: 119342858-119342858 on Build GRCh38
rs910703854 Chr.11: 119342575-119342575 on Build GRCh38
rs766037107 Chr.11: 119342655-119342655 on Build GRCh38
rs779359811 Chr.11: 119342633-119342633 on Build GRCh38
rs755240112 Chr.11: 119342822-119342822 on Build GRCh38
rs778370220 Chr.11: 119342621-119342621 on Build GRCh38
rs148399243 Chr.11: 119342613-119342613 on Build GRCh38
rs142248522 Chr.11: 119342601-119342601 on Build GRCh38
rs761326934 Chr.11: 119342862-119342862 on Build GRCh38
rs150648414 Chr.11: 119342877-119342877 on Build GRCh38
rs780080675 Chr.11: 119342984-119342984 on Build GRCh38

View Full Product Details