Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45035911-45036372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBX2
Entrez Gene ID
440097
Gene Name
developing brain homeobox 2
Gene Aliases
-
UniGene
Hs.302764
Gene Chromosome Location
Chr.12: 45014756-45051099 on Build GRCh38
Transcripts
NM_001004329.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072672 c.479C>A p.A160E Chr.12: 45036039-45036039 on Build GRCh38
2067567 c.435G>A p.P145P Chr.12: 45036083-45036083 on Build GRCh38
5564583 c.473G>A p.R158H Chr.12: 45036045-45036045 on Build GRCh38
3460942 c.497C>T p.P166L Chr.12: 45036021-45036021 on Build GRCh38
693932 c.404-2A>C p.? Chr.12: 45036116-45036116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761883356 Chr.12: 45036087-45036087 on Build GRCh38
rs369603405 Chr.12: 45036089-45036089 on Build GRCh38
rs767644541 Chr.12: 45036088-45036088 on Build GRCh38
rs746687891 Chr.12: 45036115-45036115 on Build GRCh38
rs144046195 Chr.12: 45036072-45036072 on Build GRCh38
rs777518379 Chr.12: 45036057-45036057 on Build GRCh38
rs963195263 Chr.12: 45036307-45036307 on Build GRCh38
rs769399201 Chr.12: 45036325-45036325 on Build GRCh38
rs892499723 Chr.12: 45036202-45036202 on Build GRCh38
rs753790501 Chr.12: 45036048-45036048 on Build GRCh38
rs749763879 Chr.12: 45036146-45036146 on Build GRCh38
rs777222663 Chr.12: 45035998-45035998 on Build GRCh38
rs906753230 Chr.12: 45036173-45036173 on Build GRCh38
rs1003760331 Chr.12: 45036213-45036213 on Build GRCh38
rs569379129 Chr.12: 45036317-45036317 on Build GRCh38
rs772724002 Chr.12: 45035970-45035970 on Build GRCh38
rs756053743 Chr.12: 45036073-45036075 on Build GRCh38
rs757390727 Chr.12: 45036045-45036045 on Build GRCh38
rs138216785 Chr.12: 45036049-45036049 on Build GRCh38
rs1035292142 Chr.12: 45036218-45036218 on Build GRCh38
rs61739294 Chr.12: 45036038-45036038 on Build GRCh38
rs1051730537 Chr.12: 45036084-45036084 on Build GRCh38
rs754250328 Chr.12: 45036084-45036084 on Build GRCh38
rs554666946 Chr.12: 45036162-45036162 on Build GRCh38
rs933265525 Chr.12: 45036083-45036083 on Build GRCh38
rs368571974 Chr.12: 45035973-45035973 on Build GRCh38
rs758239934 Chr.12: 45036112-45036112 on Build GRCh38
rs1045274366 Chr.12: 45036172-45036172 on Build GRCh38
rs757268338 Chr.12: 45036122-45036122 on Build GRCh38
rs202178921 Chr.12: 45036070-45036070 on Build GRCh38
rs377142325 Chr.12: 45036069-45036069 on Build GRCh38
rs1015198857 Chr.12: 45036329-45036329 on Build GRCh38
rs1012437894 Chr.12: 45035945-45035945 on Build GRCh38
rs778034743 Chr.12: 45036000-45036000 on Build GRCh38
rs1019562883 Chr.12: 45036273-45036273 on Build GRCh38
rs770312977 Chr.12: 45035994-45035994 on Build GRCh38
rs781083157 Chr.12: 45036046-45036046 on Build GRCh38
rs771776443 Chr.12: 45036201-45036201 on Build GRCh38
rs201941605 Chr.12: 45036061-45036061 on Build GRCh38
rs770421711 Chr.12: 45036116-45036116 on Build GRCh38
rs756176101 Chr.12: 45036091-45036091 on Build GRCh38
rs764428228 Chr.12: 45036042-45036042 on Build GRCh38
rs1028607394 Chr.12: 45036316-45036316 on Build GRCh38
rs2160987 Chr.12: 45036128-45036128 on Build GRCh38
rs756342390 Chr.12: 45035988-45035988 on Build GRCh38
rs778867516 Chr.12: 45036097-45036097 on Build GRCh38
rs767692350 Chr.12: 45036161-45036161 on Build GRCh38
rs764326947 Chr.12: 45036071-45036071 on Build GRCh38
rs900968851 Chr.12: 45036297-45036297 on Build GRCh38
rs1024291827 Chr.12: 45035965-45035965 on Build GRCh38
rs368914694 Chr.12: 45036062-45036062 on Build GRCh38
rs149527014 Chr.12: 45036051-45036051 on Build GRCh38
rs376901737 Chr.12: 45036129-45036129 on Build GRCh38
rs762237857 Chr.12: 45036158-45036158 on Build GRCh38
rs746599102 Chr.12: 45036058-45036058 on Build GRCh38
rs776174073 Chr.12: 45036138-45036138 on Build GRCh38
rs983621435 Chr.12: 45036015-45036015 on Build GRCh38
rs746694801 Chr.12: 45036245-45036245 on Build GRCh38
rs542219068 Chr.12: 45036132-45036132 on Build GRCh38
rs534833867 Chr.12: 45035936-45035936 on Build GRCh38
rs777378990 Chr.12: 45036113-45036113 on Build GRCh38
rs771739848 Chr.12: 45036052-45036052 on Build GRCh38
rs372280888 Chr.12: 45036068-45036068 on Build GRCh38
rs867367451 Chr.12: 45036253-45036253 on Build GRCh38
rs774515819 Chr.12: 45036152-45036152 on Build GRCh38
rs996698449 Chr.12: 45036304-45036304 on Build GRCh38
rs954580848 Chr.12: 45036323-45036323 on Build GRCh38
rs1032792204 Chr.12: 45035959-45035959 on Build GRCh38
rs11182802 Chr.12: 45036322-45036322 on Build GRCh38
rs572578273 Chr.12: 45036238-45036238 on Build GRCh38
rs905269768 Chr.12: 45036131-45036131 on Build GRCh38
rs374885519 Chr.12: 45036013-45036013 on Build GRCh38
rs768880295 Chr.12: 45036203-45036203 on Build GRCh38
rs369195739 Chr.12: 45036151-45036151 on Build GRCh38
rs753042794 Chr.12: 45036031-45036031 on Build GRCh38
rs1053776584 Chr.12: 45036192-45036192 on Build GRCh38
rs752596730 Chr.12: 45036101-45036101 on Build GRCh38
rs751701932 Chr.12: 45036043-45036043 on Build GRCh38
rs1009458720 Chr.12: 45036220-45036220 on Build GRCh38

View Full Product Details