Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75413409-75413911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1L2
Entrez Gene ID
144321
Gene Name
GLI pathogenesis-related 1 like 2
Gene Aliases
-
UniGene
Hs.406728
Gene Chromosome Location
Chr.12: 75391070-75432688 on Build GRCh38
Transcripts
NM_001270396.1 NM_152436.2 NR_072995.1 XM_011537949.2 XM_011537950.2 XM_017018848.1 XM_017018849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7136149 c.487T>C p.W163R Chr.12: 75413604-75413604 on Build GRCh38
1364216 c.578C>T p.A193V Chr.12: 75413695-75413695 on Build GRCh38
3417144 c.540A>G p.G180G Chr.12: 75413657-75413657 on Build GRCh38
6520465 c.580C>A p.P194T Chr.12: 75413697-75413697 on Build GRCh38
325405 c.484G>T p.V162F Chr.12: 75413601-75413601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772665038 Chr.12: 75413671-75413671 on Build GRCh38
rs916327347 Chr.12: 75413540-75413540 on Build GRCh38
rs754080228 Chr.12: 75413592-75413592 on Build GRCh38
rs147266448 Chr.12: 75413612-75413612 on Build GRCh38
rs768147552 Chr.12: 75413562-75413562 on Build GRCh38
rs995757243 Chr.12: 75413855-75413855 on Build GRCh38
rs1027133696 Chr.12: 75413577-75413577 on Build GRCh38
rs36018677 Chr.12: 75413503-75413503 on Build GRCh38
rs868769855 Chr.12: 75413693-75413693 on Build GRCh38
rs781012569 Chr.12: 75413731-75413731 on Build GRCh38
rs983852322 Chr.12: 75413515-75413515 on Build GRCh38
rs769269866 Chr.12: 75413638-75413638 on Build GRCh38
rs764132569 Chr.12: 75413580-75413580 on Build GRCh38
rs140863628 Chr.12: 75413659-75413659 on Build GRCh38
rs199637012 Chr.12: 75413730-75413730 on Build GRCh38
rs144196161 Chr.12: 75413742-75413742 on Build GRCh38
rs770493107 Chr.12: 75413650-75413650 on Build GRCh38
rs1039671266 Chr.12: 75413820-75413820 on Build GRCh38
rs781141091 Chr.12: 75413618-75413618 on Build GRCh38
rs370180556 Chr.12: 75413436-75413436 on Build GRCh38
rs900040252 Chr.12: 75413510-75413510 on Build GRCh38
rs891111753 Chr.12: 75413452-75413452 on Build GRCh38
rs191983256 Chr.12: 75413704-75413704 on Build GRCh38
rs535625371 Chr.12: 75413487-75413487 on Build GRCh38
rs752615296 Chr.12: 75413737-75413737 on Build GRCh38
rs757492523 Chr.12: 75413595-75413595 on Build GRCh38
rs761325625 Chr.12: 75413692-75413692 on Build GRCh38
rs753871327 Chr.12: 75413696-75413696 on Build GRCh38
rs555634895 Chr.12: 75413491-75413491 on Build GRCh38
rs527372697 Chr.12: 75413435-75413435 on Build GRCh38
rs376456819 Chr.12: 75413675-75413675 on Build GRCh38
rs777269956 Chr.12: 75413751-75413751 on Build GRCh38
rs900104463 Chr.12: 75413844-75413844 on Build GRCh38
rs931188350 Chr.12: 75413497-75413497 on Build GRCh38
rs751318067 Chr.12: 75413724-75413724 on Build GRCh38
rs925098663 Chr.12: 75413726-75413726 on Build GRCh38
rs962055677 Chr.12: 75413862-75413862 on Build GRCh38
rs867774073 Chr.12: 75413634-75413634 on Build GRCh38
rs1009007518 Chr.12: 75413837-75413837 on Build GRCh38
rs762038083 Chr.12: 75413702-75413702 on Build GRCh38
rs540140473 Chr.12: 75413818-75413818 on Build GRCh38
rs748965111 Chr.12: 75413646-75413646 on Build GRCh38
rs748125465 Chr.12: 75413629-75413629 on Build GRCh38
rs550463995 Chr.12: 75413695-75413695 on Build GRCh38
rs759996677 Chr.12: 75413669-75413669 on Build GRCh38
rs996093925 Chr.12: 75413528-75413528 on Build GRCh38
rs149571330 Chr.12: 75413606-75413606 on Build GRCh38
rs912600460 Chr.12: 75413780-75413780 on Build GRCh38
rs777203704 Chr.12: 75413642-75413642 on Build GRCh38
rs185289565 Chr.12: 75413886-75413886 on Build GRCh38
rs965249180 Chr.12: 75413448-75413448 on Build GRCh38
rs991800038 Chr.12: 75413668-75413668 on Build GRCh38
rs752773142 Chr.12: 75413573-75413573 on Build GRCh38
rs200975261 Chr.12: 75413614-75413614 on Build GRCh38
rs760865435 Chr.12: 75413579-75413579 on Build GRCh38
rs751369509 Chr.12: 75413608-75413608 on Build GRCh38

View Full Product Details