Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121243858-121244392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKK2
Entrez Gene ID
10645
Gene Name
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases
CAMKK, CAMKKB
UniGene
Hs.297343
Gene Chromosome Location
Chr.12: 121237692-121298308 on Build GRCh38
Transcripts
NM_001270485.1 NM_001270486.1 NM_006549.3 NM_153499.2 NM_153500.1 NM_172214.2 NM_172215.2 NM_172216.1 NM_172226.2 XM_005253822.3 XM_005253823.1 XM_005253824.3 XM_011537763.1 XM_011537764.2 XM_017018694.1 XM_017018695.1 XM_017018696.1 XM_017018697.1 XM_017018698.1 XM_017018699.1 XM_017018700.1 XM_017018701.1 XM_017018702.1 XM_017018703.1 XM_017018704.1 XM_017018705.1 XM_017018706.1 XM_017018707.1 XM_017018708.1 XM_017018709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
936003 c.1602G>T p.*534Y Chr.12: 121244124-121244124 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200977605 Chr.12: 121243997-121243997 on Build GRCh38
rs1001056206 Chr.12: 121244147-121244147 on Build GRCh38
rs1063841 Chr.12: 121243937-121243937 on Build GRCh38
rs201761462 Chr.12: 121244023-121244023 on Build GRCh38
rs200682617 Chr.12: 121243907-121243907 on Build GRCh38
rs991039470 Chr.12: 121243981-121243981 on Build GRCh38
rs201804162 Chr.12: 121244175-121244175 on Build GRCh38
rs941204732 Chr.12: 121244209-121244209 on Build GRCh38
rs372478031 Chr.12: 121244176-121244176 on Build GRCh38
rs750494167 Chr.12: 121244088-121244088 on Build GRCh38
rs748751234 Chr.12: 121244170-121244170 on Build GRCh38
rs566911597 Chr.12: 121244246-121244246 on Build GRCh38
rs1023221279 Chr.12: 121244319-121244319 on Build GRCh38
rs201723499 Chr.12: 121243909-121243909 on Build GRCh38
rs1013627325 Chr.12: 121244316-121244316 on Build GRCh38
rs918046044 Chr.12: 121243982-121243982 on Build GRCh38
rs201392268 Chr.12: 121244167-121244167 on Build GRCh38
rs772237108 Chr.12: 121244086-121244087 on Build GRCh38
rs200491578 Chr.12: 121244052-121244052 on Build GRCh38
rs753043765 Chr.12: 121243938-121243938 on Build GRCh38
rs1036797890 Chr.12: 121244212-121244212 on Build GRCh38
rs757298101 Chr.12: 121244057-121244057 on Build GRCh38
rs1013157717 Chr.12: 121244002-121244002 on Build GRCh38
rs1063836 Chr.12: 121243999-121243999 on Build GRCh38
rs1063842 Chr.12: 121243902-121243902 on Build GRCh38
rs1063839 Chr.12: 121243963-121243963 on Build GRCh38
rs201017691 Chr.12: 121244063-121244063 on Build GRCh38
rs200181191 Chr.12: 121244070-121244070 on Build GRCh38
rs1020728476 Chr.12: 121243928-121243928 on Build GRCh38
rs1063840 Chr.12: 121243950-121243950 on Build GRCh38
rs201084726 Chr.12: 121244071-121244071 on Build GRCh38
rs73403886 Chr.12: 121244135-121244135 on Build GRCh38
rs552747828 Chr.12: 121244260-121244260 on Build GRCh38
rs186730517 Chr.12: 121244358-121244358 on Build GRCh38
rs908077361 Chr.12: 121244230-121244230 on Build GRCh38
rs547237658 Chr.12: 121244203-121244203 on Build GRCh38
rs528522523 Chr.12: 121244048-121244048 on Build GRCh38
rs752839438 Chr.12: 121244124-121244124 on Build GRCh38
rs199769751 Chr.12: 121244089-121244089 on Build GRCh38
rs544844889 Chr.12: 121244314-121244314 on Build GRCh38
rs75456247 Chr.12: 121243942-121243942 on Build GRCh38
rs181714613 Chr.12: 121244274-121244274 on Build GRCh38
rs199868940 Chr.12: 121243968-121243968 on Build GRCh38
rs201125813 Chr.12: 121244045-121244045 on Build GRCh38
rs973907068 Chr.12: 121244269-121244269 on Build GRCh38
rs758280650 Chr.12: 121244227-121244227 on Build GRCh38
rs747307994 Chr.12: 121244098-121244098 on Build GRCh38
rs959640965 Chr.12: 121243972-121243972 on Build GRCh38
rs543018965 Chr.12: 121243954-121243954 on Build GRCh38
rs1063835 Chr.12: 121244056-121244056 on Build GRCh38
rs890964773 Chr.12: 121243885-121243885 on Build GRCh38
rs199747256 Chr.12: 121243911-121243911 on Build GRCh38
rs202053663 Chr.12: 121244047-121244047 on Build GRCh38
rs545193151 Chr.12: 121243926-121243926 on Build GRCh38
rs200126033 Chr.12: 121244026-121244026 on Build GRCh38
rs1063843 Chr.12: 121243884-121243884 on Build GRCh38
rs761913966 Chr.12: 121244082-121244082 on Build GRCh38
rs200582298 Chr.12: 121244102-121244102 on Build GRCh38
rs373935013 Chr.12: 121244221-121244221 on Build GRCh38
rs201878496 Chr.12: 121244064-121244064 on Build GRCh38
rs373323104 Chr.12: 121244228-121244228 on Build GRCh38
rs2686340 Chr.12: 121244248-121244248 on Build GRCh38
rs755743601 Chr.12: 121244141-121244141 on Build GRCh38
rs200790309 Chr.12: 121244144-121244144 on Build GRCh38
rs922277430 Chr.12: 121244121-121244121 on Build GRCh38
rs751481957 Chr.12: 121244114-121244114 on Build GRCh38
rs201435474 Chr.12: 121244116-121244116 on Build GRCh38
rs1025194262 Chr.12: 121244222-121244222 on Build GRCh38
rs983472035 Chr.12: 121244009-121244009 on Build GRCh38
rs377717128 Chr.12: 121244085-121244085 on Build GRCh38

View Full Product Details