Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122935193-122935691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
UniGene
Hs.511951
Gene Chromosome Location
Chr.12: 122918708-122975160 on Build GRCh38
Transcripts
NM_001243013.1 NM_001243014.1 NM_019624.3 NM_019625.3 NM_203444.3 XM_005253558.2 XM_011538095.2 XM_011538096.2 XM_011538098.2 XM_011538099.2 XM_017019103.1 XM_017019104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692546 c.1859A>G p.N620S Chr.12: 122935316-122935316 on Build GRCh38
3811342 c.1771C>T p.L591L Chr.12: 122935404-122935404 on Build GRCh38
5584331 c.1776C>T p.F592F Chr.12: 122935399-122935399 on Build GRCh38
6850723 c.1762G>T p.E588* Chr.12: 122935413-122935413 on Build GRCh38
5954854 c.1872C>T p.F624F Chr.12: 122935303-122935303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369578606 Chr.12: 122935440-122935440 on Build GRCh38
rs939374753 Chr.12: 122935441-122935441 on Build GRCh38
rs779782244 Chr.12: 122935258-122935258 on Build GRCh38
rs776321756 Chr.12: 122935434-122935434 on Build GRCh38
rs757258308 Chr.12: 122935329-122935329 on Build GRCh38
rs540720609 Chr.12: 122935337-122935337 on Build GRCh38
rs370354968 Chr.12: 122935593-122935593 on Build GRCh38
rs750274038 Chr.12: 122935389-122935389 on Build GRCh38
rs778564699 Chr.12: 122935437-122935437 on Build GRCh38
rs142162332 Chr.12: 122935429-122935429 on Build GRCh38
rs759840557 Chr.12: 122935443-122935449 on Build GRCh38
rs372342420 Chr.12: 122935239-122935239 on Build GRCh38
rs373846386 Chr.12: 122935408-122935408 on Build GRCh38
rs758201231 Chr.12: 122935444-122935444 on Build GRCh38
rs752120982 Chr.12: 122935413-122935413 on Build GRCh38
rs561296678 Chr.12: 122935243-122935243 on Build GRCh38
rs1021771545 Chr.12: 122935492-122935492 on Build GRCh38
rs185108816 Chr.12: 122935385-122935385 on Build GRCh38
rs746801159 Chr.12: 122935462-122935462 on Build GRCh38
rs773100436 Chr.12: 122935299-122935299 on Build GRCh38
rs60920158 Chr.12: 122935567-122935567 on Build GRCh38
rs567208712 Chr.12: 122935496-122935496 on Build GRCh38
rs771047411 Chr.12: 122935394-122935394 on Build GRCh38
rs61747213 Chr.12: 122935423-122935423 on Build GRCh38
rs147328787 Chr.12: 122935308-122935308 on Build GRCh38
rs571037447 Chr.12: 122935601-122935601 on Build GRCh38
rs759638966 Chr.12: 122935396-122935396 on Build GRCh38
rs190003896 Chr.12: 122935399-122935399 on Build GRCh38
rs774428844 Chr.12: 122935395-122935395 on Build GRCh38
rs777478165 Chr.12: 122935445-122935445 on Build GRCh38
rs757191236 Chr.12: 122935466-122935466 on Build GRCh38
rs201983318 Chr.12: 122935285-122935285 on Build GRCh38
rs144937544 Chr.12: 122935384-122935384 on Build GRCh38
rs1030673914 Chr.12: 122935402-122935402 on Build GRCh38
rs866807727 Chr.12: 122935373-122935373 on Build GRCh38
rs755648175 Chr.12: 122935249-122935249 on Build GRCh38
rs369456007 Chr.12: 122935284-122935284 on Build GRCh38
rs754640896 Chr.12: 122935378-122935378 on Build GRCh38
rs149319315 Chr.12: 122935369-122935369 on Build GRCh38
rs897100201 Chr.12: 122935261-122935261 on Build GRCh38
rs772502697 Chr.12: 122935393-122935393 on Build GRCh38
rs370215646 Chr.12: 122935395-122935395 on Build GRCh38
rs781153198 Chr.12: 122935473-122935473 on Build GRCh38
rs377056518 Chr.12: 122935475-122935475 on Build GRCh38
rs779368981 Chr.12: 122935366-122935366 on Build GRCh38
rs538367668 Chr.12: 122935602-122935602 on Build GRCh38
rs765484313 Chr.12: 122935316-122935316 on Build GRCh38
rs770865113 Chr.12: 122935309-122935309 on Build GRCh38
rs549018731 Chr.12: 122935493-122935493 on Build GRCh38
rs970240682 Chr.12: 122935507-122935507 on Build GRCh38
rs778404765 Chr.12: 122935232-122935232 on Build GRCh38
rs776924654 Chr.12: 122935310-122935310 on Build GRCh38
rs1052993034 Chr.12: 122935634-122935634 on Build GRCh38
rs372875439 Chr.12: 122935345-122935345 on Build GRCh38
rs368354303 Chr.12: 122935226-122935226 on Build GRCh38
rs755701170 Chr.12: 122935359-122935359 on Build GRCh38
rs779755014 Chr.12: 122935557-122935557 on Build GRCh38
rs375317325 Chr.12: 122935269-122935269 on Build GRCh38
rs748839191 Chr.12: 122935263-122935263 on Build GRCh38
rs934568080 Chr.12: 122935632-122935632 on Build GRCh38
rs375875165 Chr.12: 122935348-122935348 on Build GRCh38
rs115582875 Chr.12: 122935382-122935382 on Build GRCh38
rs759608941 Chr.12: 122935312-122935312 on Build GRCh38
rs753333607 Chr.12: 122935432-122935432 on Build GRCh38
rs928752481 Chr.12: 122935562-122935562 on Build GRCh38
rs763436577 Chr.12: 122935407-122935407 on Build GRCh38

View Full Product Details