Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16253394-16253940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO42
Entrez Gene ID
54455
Gene Name
F-box protein 42
Gene Aliases
Fbx42, JFK
UniGene
Hs.522384
Gene Chromosome Location
Chr.1: 16246840-16352491 on Build GRCh38
Transcripts
NM_018994.2 XM_006710698.3 XM_017001502.1 XM_017001503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229972 c.770G>T p.S257I Chr.1: 16253729-16253729 on Build GRCh38
1335945 c.803_804insCAGTG p.W268fs*26 Chr.1: 16253695-16253696 on Build GRCh38
6479406 c.827C>G p.S276C Chr.1: 16253672-16253672 on Build GRCh38
5599543 c.819G>A p.P273P Chr.1: 16253680-16253680 on Build GRCh38
6506960 c.841C>T p.H281Y Chr.1: 16253658-16253658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778488633 Chr.1: 16253600-16253600 on Build GRCh38
rs758355341 Chr.1: 16253566-16253566 on Build GRCh38
rs982389703 Chr.1: 16253786-16253786 on Build GRCh38
rs200270786 Chr.1: 16253692-16253692 on Build GRCh38
rs546529339 Chr.1: 16253771-16253771 on Build GRCh38
rs536164340 Chr.1: 16253549-16253549 on Build GRCh38
rs751621831 Chr.1: 16253787-16253787 on Build GRCh38
rs764135682 Chr.1: 16253591-16253591 on Build GRCh38
rs566284629 Chr.1: 16253772-16253772 on Build GRCh38
rs925946758 Chr.1: 16253437-16253437 on Build GRCh38
rs752670963 Chr.1: 16253522-16253522 on Build GRCh38
rs370922326 Chr.1: 16253474-16253474 on Build GRCh38
rs151049038 Chr.1: 16253539-16253539 on Build GRCh38
rs749390223 Chr.1: 16253681-16253681 on Build GRCh38
rs528708026 Chr.1: 16253543-16253543 on Build GRCh38
rs755678090 Chr.1: 16253594-16253594 on Build GRCh38
rs754539255 Chr.1: 16253735-16253735 on Build GRCh38
rs937763281 Chr.1: 16253797-16253797 on Build GRCh38
rs868750617 Chr.1: 16253505-16253505 on Build GRCh38
rs749463542 Chr.1: 16253783-16253783 on Build GRCh38
rs772917474 Chr.1: 16253719-16253719 on Build GRCh38
rs767328940 Chr.1: 16253715-16253715 on Build GRCh38
rs757799210 Chr.1: 16253764-16253764 on Build GRCh38
rs768720065 Chr.1: 16253684-16253684 on Build GRCh38
rs557112350 Chr.1: 16253488-16253488 on Build GRCh38
rs546570439 Chr.1: 16253537-16253537 on Build GRCh38
rs989318938 Chr.1: 16253496-16253496 on Build GRCh38
rs945172909 Chr.1: 16253503-16253503 on Build GRCh38
rs1054805687 Chr.1: 16253561-16253561 on Build GRCh38
rs372879342 Chr.1: 16253654-16253654 on Build GRCh38
rs926575262 Chr.1: 16253445-16253445 on Build GRCh38
rs959267621 Chr.1: 16253484-16253484 on Build GRCh38
rs181648207 Chr.1: 16253508-16253508 on Build GRCh38
rs764781543 Chr.1: 16253751-16253751 on Build GRCh38
rs753337516 Chr.1: 16253598-16253598 on Build GRCh38
rs898795597 Chr.1: 16253572-16253572 on Build GRCh38
rs769950759 Chr.1: 16253658-16253658 on Build GRCh38
rs185176747 Chr.1: 16253578-16253578 on Build GRCh38
rs373398613 Chr.1: 16253726-16253726 on Build GRCh38
rs377314532 Chr.1: 16253713-16253713 on Build GRCh38
rs752135309 Chr.1: 16253760-16253760 on Build GRCh38
rs747452132 Chr.1: 16253607-16253607 on Build GRCh38
rs913831746 Chr.1: 16253773-16253773 on Build GRCh38
rs995495219 Chr.1: 16253744-16253744 on Build GRCh38
rs190454215 Chr.1: 16253593-16253593 on Build GRCh38
rs781449051 Chr.1: 16253889-16253889 on Build GRCh38
rs767238972 Chr.1: 16253585-16253585 on Build GRCh38
rs368997762 Chr.1: 16253686-16253686 on Build GRCh38
rs376503139 Chr.1: 16253734-16253734 on Build GRCh38
rs1025276164 Chr.1: 16253839-16253839 on Build GRCh38
rs991976159 Chr.1: 16253487-16253487 on Build GRCh38
rs780210121 Chr.1: 16253775-16253775 on Build GRCh38
rs762779149 Chr.1: 16253597-16253598 on Build GRCh38
rs1046212199 Chr.1: 16253517-16253517 on Build GRCh38
rs561840962 Chr.1: 16253547-16253547 on Build GRCh38
rs759346511 Chr.1: 16253433-16253433 on Build GRCh38
rs374686297 Chr.1: 16253657-16253657 on Build GRCh38
rs1054859054 Chr.1: 16253824-16253824 on Build GRCh38
rs930322161 Chr.1: 16253896-16253896 on Build GRCh38
rs775762526 Chr.1: 16253680-16253680 on Build GRCh38
rs776196585 Chr.1: 16253420-16253420 on Build GRCh38
rs865832476 Chr.1: 16253672-16253672 on Build GRCh38
rs757303378 Chr.1: 16253855-16253855 on Build GRCh38
rs766019404 Chr.1: 16253595-16253595 on Build GRCh38
rs781756251 Chr.1: 16253766-16253766 on Build GRCh38
rs992102224 Chr.1: 16253801-16253801 on Build GRCh38
rs541511767 Chr.1: 16253544-16253544 on Build GRCh38
rs760492886 Chr.1: 16253723-16253723 on Build GRCh38
rs200092981 Chr.1: 16253641-16253641 on Build GRCh38

View Full Product Details