Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20652773-20653279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641369 c.1005C>T p.I335I Chr.1: 20652960-20652960 on Build GRCh38
4676707 c.1109A>T p.K370M Chr.1: 20652856-20652856 on Build GRCh38
678811 c.1018C>A p.L340I Chr.1: 20652947-20652947 on Build GRCh38
7363935 c.1110G>C p.K370N Chr.1: 20652855-20652855 on Build GRCh38
6285700 c.1090G>T p.V364L Chr.1: 20652875-20652875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756958668 Chr.1: 20652839-20652839 on Build GRCh38
rs370326677 Chr.1: 20652846-20652846 on Build GRCh38
rs373520642 Chr.1: 20652965-20652965 on Build GRCh38
rs776647294 Chr.1: 20652853-20652855 on Build GRCh38
rs398102562 Chr.1: 20652817-20652818 on Build GRCh38
rs924285226 Chr.1: 20653137-20653137 on Build GRCh38
rs778674191 Chr.1: 20652979-20652979 on Build GRCh38
rs773138274 Chr.1: 20652914-20652914 on Build GRCh38
rs746740290 Chr.1: 20652866-20652866 on Build GRCh38
rs1013543878 Chr.1: 20653255-20653255 on Build GRCh38
rs745423124 Chr.1: 20652848-20652848 on Build GRCh38
rs190780604 Chr.1: 20652915-20652915 on Build GRCh38
rs374150866 Chr.1: 20653017-20653017 on Build GRCh38
rs567466993 Chr.1: 20653012-20653012 on Build GRCh38
rs753498493 Chr.1: 20652838-20652838 on Build GRCh38
rs770209876 Chr.1: 20652898-20652898 on Build GRCh38
rs914518661 Chr.1: 20653100-20653100 on Build GRCh38
rs1057289119 Chr.1: 20653243-20653243 on Build GRCh38
rs534878707 Chr.1: 20653018-20653018 on Build GRCh38
rs753124108 Chr.1: 20652974-20652974 on Build GRCh38
rs965461154 Chr.1: 20653045-20653045 on Build GRCh38
rs953262698 Chr.1: 20652807-20652807 on Build GRCh38
rs766751039 Chr.1: 20652918-20652918 on Build GRCh38
rs757955819 Chr.1: 20653007-20653007 on Build GRCh38
rs923107922 Chr.1: 20652799-20652799 on Build GRCh38
rs746980641 Chr.1: 20653010-20653010 on Build GRCh38
rs770639102 Chr.1: 20653178-20653178 on Build GRCh38
rs776936181 Chr.1: 20652889-20652889 on Build GRCh38
rs762114838 Chr.1: 20652890-20652890 on Build GRCh38
rs577217583 Chr.1: 20653051-20653051 on Build GRCh38
rs569901169 Chr.1: 20652794-20652794 on Build GRCh38
rs765422891 Chr.1: 20652809-20652809 on Build GRCh38
rs916082191 Chr.1: 20653258-20653258 on Build GRCh38
rs370620552 Chr.1: 20653152-20653152 on Build GRCh38
rs998682903 Chr.1: 20653059-20653059 on Build GRCh38
rs1042154999 Chr.1: 20653061-20653061 on Build GRCh38
rs959002030 Chr.1: 20653172-20653172 on Build GRCh38
rs370821391 Chr.1: 20653016-20653016 on Build GRCh38
rs1048717980 Chr.1: 20653013-20653013 on Build GRCh38
rs748072642 Chr.1: 20652984-20652984 on Build GRCh38
rs760581266 Chr.1: 20652813-20652814 on Build GRCh38
rs750576584 Chr.1: 20652814-20652814 on Build GRCh38
rs751853015 Chr.1: 20652831-20652831 on Build GRCh38
rs768576471 Chr.1: 20652881-20652881 on Build GRCh38
rs36110575 Chr.1: 20653080-20653080 on Build GRCh38
rs921210147 Chr.1: 20652959-20652959 on Build GRCh38
rs778884507 Chr.1: 20652937-20652937 on Build GRCh38
rs373275420 Chr.1: 20653117-20653117 on Build GRCh38
rs200247842 Chr.1: 20652966-20652966 on Build GRCh38
rs953252004 Chr.1: 20653081-20653081 on Build GRCh38
rs761918335 Chr.1: 20652800-20652800 on Build GRCh38
rs12410193 Chr.1: 20653087-20653087 on Build GRCh38
rs552700525 Chr.1: 20653027-20653027 on Build GRCh38
rs766952000 Chr.1: 20652825-20652825 on Build GRCh38
rs968337196 Chr.1: 20653104-20653104 on Build GRCh38
rs977091840 Chr.1: 20653115-20653115 on Build GRCh38
rs771880722 Chr.1: 20652861-20652861 on Build GRCh38
rs759832138 Chr.1: 20652928-20652928 on Build GRCh38
rs752142561 Chr.1: 20652927-20652927 on Build GRCh38
rs780062607 Chr.1: 20652864-20652864 on Build GRCh38
rs755396887 Chr.1: 20652832-20652832 on Build GRCh38
rs1057271999 Chr.1: 20652891-20652891 on Build GRCh38
rs761733911 Chr.1: 20652983-20652984 on Build GRCh38
rs746812261 Chr.1: 20653146-20653146 on Build GRCh38
rs749852803 Chr.1: 20652983-20652983 on Build GRCh38
rs924409280 Chr.1: 20652885-20652885 on Build GRCh38
rs779789282 Chr.1: 20653008-20653008 on Build GRCh38
rs376370824 Chr.1: 20653006-20653006 on Build GRCh38
rs756870725 Chr.1: 20652978-20652978 on Build GRCh38
rs11428148 Chr.1: 20652814-20652815 on Build GRCh38

View Full Product Details