Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20655351-20655855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1929149 c.513G>A p.T171T Chr.1: 20655529-20655529 on Build GRCh38
1929150 c.508-1G>T p.? Chr.1: 20655535-20655535 on Build GRCh38
118659 c.582C>T p.P194P Chr.1: 20655460-20655460 on Build GRCh38
3803756 c.424G>A p.G142S Chr.1: 20655759-20655759 on Build GRCh38
3731976 c.436G>A p.G146R Chr.1: 20655747-20655747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111862732 Chr.1: 20655675-20655675 on Build GRCh38
rs747589765 Chr.1: 20655450-20655450 on Build GRCh38
rs981233757 Chr.1: 20655390-20655394 on Build GRCh38
rs779877980 Chr.1: 20655641-20655641 on Build GRCh38
rs924948918 Chr.1: 20655533-20655533 on Build GRCh38
rs752665292 Chr.1: 20655405-20655405 on Build GRCh38
rs369049692 Chr.1: 20655544-20655544 on Build GRCh38
rs769322830 Chr.1: 20655457-20655457 on Build GRCh38
rs1046715723 Chr.1: 20655390-20655390 on Build GRCh38
rs758308800 Chr.1: 20655742-20655742 on Build GRCh38
rs757889180 Chr.1: 20655418-20655418 on Build GRCh38
rs1007990405 Chr.1: 20655765-20655765 on Build GRCh38
rs754011834 Chr.1: 20655486-20655486 on Build GRCh38
rs1019417198 Chr.1: 20655768-20655768 on Build GRCh38
rs199859745 Chr.1: 20655733-20655733 on Build GRCh38
rs750695197 Chr.1: 20655496-20655496 on Build GRCh38
rs374078485 Chr.1: 20655806-20655806 on Build GRCh38
rs376317784 Chr.1: 20655713-20655713 on Build GRCh38
rs376445992 Chr.1: 20655654-20655654 on Build GRCh38
rs769066839 Chr.1: 20655657-20655657 on Build GRCh38
rs765830229 Chr.1: 20655494-20655494 on Build GRCh38
rs373404531 Chr.1: 20655693-20655693 on Build GRCh38
rs369573026 Chr.1: 20655429-20655429 on Build GRCh38
rs761964650 Chr.1: 20655663-20655663 on Build GRCh38
rs1013765860 Chr.1: 20655703-20655703 on Build GRCh38
rs759393357 Chr.1: 20655481-20655481 on Build GRCh38
rs1043222730 Chr.1: 20655566-20655566 on Build GRCh38
rs768942021 Chr.1: 20655750-20655750 on Build GRCh38
rs557808453 Chr.1: 20655756-20655756 on Build GRCh38
rs756270668 Chr.1: 20655413-20655413 on Build GRCh38
rs780666099 Chr.1: 20655526-20655526 on Build GRCh38
rs766705326 Chr.1: 20655684-20655684 on Build GRCh38
rs11547742 Chr.1: 20655530-20655530 on Build GRCh38
rs554184374 Chr.1: 20655672-20655672 on Build GRCh38
rs376070047 Chr.1: 20655523-20655523 on Build GRCh38
rs763699436 Chr.1: 20655774-20655774 on Build GRCh38
rs745612594 Chr.1: 20655570-20655570 on Build GRCh38
rs201594599 Chr.1: 20655716-20655716 on Build GRCh38
rs904279388 Chr.1: 20655576-20655577 on Build GRCh38
rs910452022 Chr.1: 20655817-20655818 on Build GRCh38
rs749969715 Chr.1: 20655807-20655807 on Build GRCh38
rs149487745 Chr.1: 20655484-20655484 on Build GRCh38
rs184384404 Chr.1: 20655762-20655762 on Build GRCh38
rs969517205 Chr.1: 20655388-20655388 on Build GRCh38
rs992126555 Chr.1: 20655385-20655385 on Build GRCh38
rs756911269 Chr.1: 20655799-20655799 on Build GRCh38
rs913666293 Chr.1: 20655830-20655830 on Build GRCh38
rs756635483 Chr.1: 20655393-20655395 on Build GRCh38
rs936394586 Chr.1: 20655543-20655543 on Build GRCh38
rs1037673565 Chr.1: 20655724-20655724 on Build GRCh38
rs749197629 Chr.1: 20655468-20655468 on Build GRCh38
rs572098042 Chr.1: 20655719-20655719 on Build GRCh38
rs546737829 Chr.1: 20655834-20655834 on Build GRCh38
rs767499385 Chr.1: 20655789-20655789 on Build GRCh38
rs754849715 Chr.1: 20655822-20655822 on Build GRCh38
rs780152653 Chr.1: 20655819-20655819 on Build GRCh38
rs949134639 Chr.1: 20655833-20655833 on Build GRCh38
rs1013834937 Chr.1: 20655386-20655386 on Build GRCh38
rs774264202 Chr.1: 20655393-20655393 on Build GRCh38
rs748859395 Chr.1: 20655545-20655545 on Build GRCh38
rs750968899 Chr.1: 20655666-20655666 on Build GRCh38
rs776997435 Chr.1: 20655754-20655754 on Build GRCh38
rs751958293 Chr.1: 20655685-20655685 on Build GRCh38
rs770937124 Chr.1: 20655470-20655470 on Build GRCh38
rs765595117 Chr.1: 20655664-20655664 on Build GRCh38
rs1042720788 Chr.1: 20655824-20655824 on Build GRCh38
rs372393674 Chr.1: 20655392-20655392 on Build GRCh38
rs146767054 Chr.1: 20655482-20655482 on Build GRCh38
rs773211968 Chr.1: 20655763-20655763 on Build GRCh38
rs758396731 Chr.1: 20655812-20655812 on Build GRCh38
rs778459576 Chr.1: 20655394-20655395 on Build GRCh38
rs755998781 Chr.1: 20655810-20655810 on Build GRCh38
rs746732719 Chr.1: 20655821-20655821 on Build GRCh38
rs755809942 Chr.1: 20655538-20655538 on Build GRCh38
rs773390340 Chr.1: 20655399-20655399 on Build GRCh38
rs202107268 Chr.1: 20655790-20655790 on Build GRCh38
rs749381938 Chr.1: 20655828-20655828 on Build GRCh38
rs933186948 Chr.1: 20655460-20655460 on Build GRCh38
rs771926276 Chr.1: 20655573-20655573 on Build GRCh38
rs148134377 Chr.1: 20655748-20655748 on Build GRCh38
rs975020316 Chr.1: 20655529-20655529 on Build GRCh38
rs189359933 Chr.1: 20655595-20655595 on Build GRCh38
rs754573232 Chr.1: 20655439-20655439 on Build GRCh38
rs938259502 Chr.1: 20655722-20655722 on Build GRCh38
rs759678026 Chr.1: 20655396-20655396 on Build GRCh38
rs3738140 Chr.1: 20655658-20655658 on Build GRCh38
rs373001976 Chr.1: 20655548-20655548 on Build GRCh38
rs767826481 Chr.1: 20655404-20655404 on Build GRCh38
rs372948054 Chr.1: 20655489-20655489 on Build GRCh38
rs914065741 Chr.1: 20655599-20655599 on Build GRCh38
rs774007750 Chr.1: 20655480-20655480 on Build GRCh38
rs192228415 Chr.1: 20655638-20655638 on Build GRCh38
rs111334879 Chr.1: 20655483-20655483 on Build GRCh38
rs745797101 Chr.1: 20655739-20655739 on Build GRCh38
rs749731950 Chr.1: 20655803-20655803 on Build GRCh38
rs1001022882 Chr.1: 20655596-20655596 on Build GRCh38
rs745406431 Chr.1: 20655547-20655548 on Build GRCh38
rs886045871 Chr.1: 20655532-20655532 on Build GRCh38
rs748376592 Chr.1: 20655823-20655823 on Build GRCh38
rs777635772 Chr.1: 20655459-20655459 on Build GRCh38
rs778917497 Chr.1: 20655556-20655556 on Build GRCh38
rs759903353 Chr.1: 20655773-20655773 on Build GRCh38
rs763482311 Chr.1: 20655767-20655767 on Build GRCh38
rs779117025 Chr.1: 20655423-20655423 on Build GRCh38
rs771814545 Chr.1: 20655580-20655581 on Build GRCh38
rs933239309 Chr.1: 20655791-20655791 on Build GRCh38
rs746746682 Chr.1: 20655758-20655758 on Build GRCh38
rs766934491 Chr.1: 20655771-20655771 on Build GRCh38
rs780909764 Chr.1: 20655449-20655449 on Build GRCh38
rs147322620 Chr.1: 20655772-20655772 on Build GRCh38
rs768578151 Chr.1: 20655818-20655818 on Build GRCh38
rs754420505 Chr.1: 20655416-20655416 on Build GRCh38
rs756826240 Chr.1: 20655721-20655721 on Build GRCh38
rs568742601 Chr.1: 20655641-20655641 on Build GRCh38
rs773333649 Chr.1: 20655825-20655825 on Build GRCh38
rs993998618 Chr.1: 20655454-20655454 on Build GRCh38
rs76266717 Chr.1: 20655415-20655415 on Build GRCh38
rs759658987 Chr.1: 20655549-20655549 on Build GRCh38
rs779992404 Chr.1: 20655747-20655747 on Build GRCh38

View Full Product Details