Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20660803-20661309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481854 c.254C>T p.S85F Chr.1: 20660943-20660943 on Build GRCh38
1667214 c.175A>G p.T59A Chr.1: 20661227-20661227 on Build GRCh38
903317 c.220C>A p.L74I Chr.1: 20660977-20660977 on Build GRCh38
1338559 c.102G>T p.L34L Chr.1: 20661300-20661300 on Build GRCh38
4481317 c.249C>T p.S83S Chr.1: 20660948-20660948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761796965 Chr.1: 20660925-20660926 on Build GRCh38
rs761600319 Chr.1: 20661149-20661149 on Build GRCh38
rs747098798 Chr.1: 20661154-20661154 on Build GRCh38
rs756728875 Chr.1: 20661283-20661283 on Build GRCh38
rs773982629 Chr.1: 20660910-20660910 on Build GRCh38
rs947926749 Chr.1: 20661138-20661138 on Build GRCh38
rs769669810 Chr.1: 20660986-20660991 on Build GRCh38
rs770770173 Chr.1: 20660894-20660894 on Build GRCh38
rs767417901 Chr.1: 20660932-20660932 on Build GRCh38
rs200838967 Chr.1: 20660933-20660933 on Build GRCh38
rs747449468 Chr.1: 20660978-20660978 on Build GRCh38
rs189424476 Chr.1: 20661163-20661163 on Build GRCh38
rs550280048 Chr.1: 20660961-20660961 on Build GRCh38
rs779763847 Chr.1: 20661288-20661288 on Build GRCh38
rs887468386 Chr.1: 20661145-20661145 on Build GRCh38
rs200589624 Chr.1: 20661269-20661269 on Build GRCh38
rs777772794 Chr.1: 20661082-20661082 on Build GRCh38
rs936904719 Chr.1: 20661233-20661233 on Build GRCh38
rs369325245 Chr.1: 20661019-20661019 on Build GRCh38
rs528257664 Chr.1: 20661268-20661268 on Build GRCh38
rs991973558 Chr.1: 20661091-20661091 on Build GRCh38
rs755896796 Chr.1: 20660836-20660836 on Build GRCh38
rs201767006 Chr.1: 20661280-20661280 on Build GRCh38
rs772573714 Chr.1: 20660864-20660864 on Build GRCh38
rs773029088 Chr.1: 20661244-20661244 on Build GRCh38
rs377206248 Chr.1: 20660854-20660854 on Build GRCh38
rs150936849 Chr.1: 20661242-20661242 on Build GRCh38
rs577577693 Chr.1: 20661143-20661143 on Build GRCh38
rs770645431 Chr.1: 20661014-20661014 on Build GRCh38
rs764152311 Chr.1: 20661029-20661029 on Build GRCh38
rs774147717 Chr.1: 20661018-20661018 on Build GRCh38
rs375334867 Chr.1: 20660916-20660916 on Build GRCh38
rs763767892 Chr.1: 20660937-20660937 on Build GRCh38
rs780562028 Chr.1: 20660977-20660977 on Build GRCh38
rs1804069 Chr.1: 20660943-20660943 on Build GRCh38
rs762661909 Chr.1: 20660893-20660893 on Build GRCh38
rs1034188838 Chr.1: 20661135-20661135 on Build GRCh38
rs912277276 Chr.1: 20661194-20661194 on Build GRCh38
rs781685198 Chr.1: 20661011-20661011 on Build GRCh38
rs753702642 Chr.1: 20660939-20660939 on Build GRCh38
rs912711217 Chr.1: 20660949-20660949 on Build GRCh38
rs897338524 Chr.1: 20660912-20660912 on Build GRCh38
rs140409217 Chr.1: 20661209-20661209 on Build GRCh38
rs372328091 Chr.1: 20660875-20660875 on Build GRCh38
rs745482314 Chr.1: 20661201-20661201 on Build GRCh38
rs532150969 Chr.1: 20660953-20660953 on Build GRCh38
rs765563614 Chr.1: 20661113-20661113 on Build GRCh38
rs373157475 Chr.1: 20661174-20661174 on Build GRCh38
rs979950154 Chr.1: 20661065-20661065 on Build GRCh38
rs759126376 Chr.1: 20660918-20660918 on Build GRCh38
rs1052427793 Chr.1: 20661060-20661060 on Build GRCh38
rs567066796 Chr.1: 20661088-20661088 on Build GRCh38
rs750665522 Chr.1: 20661155-20661155 on Build GRCh38
rs756575337 Chr.1: 20661188-20661188 on Build GRCh38
rs1011928396 Chr.1: 20660861-20660861 on Build GRCh38
rs761548809 Chr.1: 20661232-20661232 on Build GRCh38
rs780430960 Chr.1: 20660867-20660867 on Build GRCh38
rs778280288 Chr.1: 20661284-20661284 on Build GRCh38
rs150110141 Chr.1: 20661266-20661266 on Build GRCh38
rs778335267 Chr.1: 20661197-20661197 on Build GRCh38
rs1001750883 Chr.1: 20661134-20661134 on Build GRCh38
rs778007667 Chr.1: 20660838-20660838 on Build GRCh38
rs758439973 Chr.1: 20660972-20660972 on Build GRCh38
rs142391409 Chr.1: 20661066-20661067 on Build GRCh38
rs968009641 Chr.1: 20661057-20661057 on Build GRCh38
rs201557697 Chr.1: 20660936-20660936 on Build GRCh38
rs778961282 Chr.1: 20660852-20660852 on Build GRCh38
rs200373074 Chr.1: 20661178-20661178 on Build GRCh38
rs748578362 Chr.1: 20661012-20661012 on Build GRCh38
rs79243170 Chr.1: 20660872-20660872 on Build GRCh38
rs892509764 Chr.1: 20661066-20661066 on Build GRCh38
rs534524510 Chr.1: 20661108-20661108 on Build GRCh38
rs555185346 Chr.1: 20661077-20661077 on Build GRCh38
rs750435886 Chr.1: 20660964-20660964 on Build GRCh38
rs763079640 Chr.1: 20661256-20661256 on Build GRCh38
rs746007450 Chr.1: 20660859-20660859 on Build GRCh38
rs752475661 Chr.1: 20660831-20660831 on Build GRCh38
rs17837938 Chr.1: 20661285-20661285 on Build GRCh38
rs377211359 Chr.1: 20661159-20661159 on Build GRCh38
rs751727838 Chr.1: 20661164-20661164 on Build GRCh38
rs776581220 Chr.1: 20661228-20661228 on Build GRCh38
rs986993375 Chr.1: 20660920-20660920 on Build GRCh38
rs760528439 Chr.1: 20661026-20661026 on Build GRCh38
rs768515558 Chr.1: 20661213-20661213 on Build GRCh38
rs143421115 Chr.1: 20661203-20661203 on Build GRCh38
rs774852781 Chr.1: 20661204-20661204 on Build GRCh38
rs117238433 Chr.1: 20661022-20661022 on Build GRCh38
rs1018203357 Chr.1: 20661054-20661054 on Build GRCh38
rs752956398 Chr.1: 20661025-20661025 on Build GRCh38
rs755547426 Chr.1: 20661000-20661000 on Build GRCh38
rs573529962 Chr.1: 20661173-20661173 on Build GRCh38
rs559276348 Chr.1: 20661118-20661118 on Build GRCh38
rs1025260225 Chr.1: 20661162-20661162 on Build GRCh38
rs757514079 Chr.1: 20660841-20660841 on Build GRCh38
rs758642488 Chr.1: 20661030-20661030 on Build GRCh38

View Full Product Details