Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74750436-74750932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1A2
Entrez Gene ID
1544
Gene Name
cytochrome P450 family 1 subfamily A member 2
Gene Aliases
CP12, P3-450, P450(PA)
UniGene
Hs.1361
Gene Chromosome Location
Chr.15: 74748843-74756600 on Build GRCh38
Transcripts
NM_000761.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6242298 c.825_827delTGA p.D276delD Chr.15: 74750563-74750565 on Build GRCh38
4506602 c.724C>T p.L242F Chr.15: 74750462-74750462 on Build GRCh38
3503978 c.804C>T p.V268V Chr.15: 74750542-74750542 on Build GRCh38
6639035 c.792G>A p.L264L Chr.15: 74750530-74750530 on Build GRCh38
4773202 c.831+8C>A p.? Chr.15: 74750577-74750577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765330684 Chr.15: 74750548-74750548 on Build GRCh38
rs764657919 Chr.15: 74750811-74750811 on Build GRCh38
rs954408196 Chr.15: 74750726-74750726 on Build GRCh38
rs34182516 Chr.15: 74750632-74750632 on Build GRCh38
rs576092370 Chr.15: 74750578-74750578 on Build GRCh38
rs45533242 Chr.15: 74750607-74750607 on Build GRCh38
rs532415396 Chr.15: 74750669-74750669 on Build GRCh38
rs759538814 Chr.15: 74750608-74750608 on Build GRCh38
rs542879792 Chr.15: 74750479-74750479 on Build GRCh38
rs759584175 Chr.15: 74750538-74750538 on Build GRCh38
rs975610364 Chr.15: 74750897-74750897 on Build GRCh38
rs1020691866 Chr.15: 74750717-74750717 on Build GRCh38
rs989256995 Chr.15: 74750829-74750829 on Build GRCh38
rs756704382 Chr.15: 74750539-74750539 on Build GRCh38
rs560219459 Chr.15: 74750690-74750690 on Build GRCh38
rs777563957 Chr.15: 74750587-74750587 on Build GRCh38
rs775258473 Chr.15: 74750510-74750510 on Build GRCh38
rs1044494837 Chr.15: 74750875-74750875 on Build GRCh38
rs543761661 Chr.15: 74750586-74750586 on Build GRCh38
rs776619123 Chr.15: 74750531-74750531 on Build GRCh38
rs966814318 Chr.15: 74750718-74750718 on Build GRCh38
rs564886643 Chr.15: 74750634-74750634 on Build GRCh38
rs377683245 Chr.15: 74750494-74750494 on Build GRCh38
rs770736974 Chr.15: 74750600-74750600 on Build GRCh38
rs947452126 Chr.15: 74750842-74750842 on Build GRCh38
rs778744251 Chr.15: 74750473-74750473 on Build GRCh38
rs747838338 Chr.15: 74750514-74750514 on Build GRCh38
rs768186715 Chr.15: 74750466-74750466 on Build GRCh38
rs778361701 Chr.15: 74750577-74750577 on Build GRCh38
rs370942811 Chr.15: 74750564-74750564 on Build GRCh38
rs375308675 Chr.15: 74750590-74750590 on Build GRCh38
rs905819883 Chr.15: 74750891-74750891 on Build GRCh38
rs938683103 Chr.15: 74750909-74750909 on Build GRCh38
rs924913866 Chr.15: 74750819-74750819 on Build GRCh38
rs71541924 Chr.15: 74750628-74750629 on Build GRCh38
rs905891949 Chr.15: 74750574-74750574 on Build GRCh38
rs527607611 Chr.15: 74750838-74750838 on Build GRCh38
rs763174698 Chr.15: 74750551-74750551 on Build GRCh38
rs575035489 Chr.15: 74750484-74750484 on Build GRCh38
rs561167723 Chr.15: 74750507-74750507 on Build GRCh38
rs752723652 Chr.15: 74750550-74750550 on Build GRCh38
rs1002088448 Chr.15: 74750601-74750601 on Build GRCh38
rs750907355 Chr.15: 74750568-74750568 on Build GRCh38
rs374699009 Chr.15: 74750572-74750572 on Build GRCh38
rs769397646 Chr.15: 74750621-74750622 on Build GRCh38
rs1038333951 Chr.15: 74750569-74750569 on Build GRCh38
rs34954622 Chr.15: 74750588-74750588 on Build GRCh38
rs778592782 Chr.15: 74750581-74750581 on Build GRCh38
rs551640542 Chr.15: 74750670-74750670 on Build GRCh38
rs957629866 Chr.15: 74750836-74750836 on Build GRCh38
rs747609163 Chr.15: 74750583-74750583 on Build GRCh38
rs375045680 Chr.15: 74750465-74750465 on Build GRCh38
rs140421378 Chr.15: 74750554-74750554 on Build GRCh38
rs769661100 Chr.15: 74750619-74750619 on Build GRCh38
rs987307397 Chr.15: 74750764-74750764 on Build GRCh38
rs764233104 Chr.15: 74750552-74750552 on Build GRCh38
rs935281105 Chr.15: 74750822-74750822 on Build GRCh38
rs1005063025 Chr.15: 74750624-74750624 on Build GRCh38

View Full Product Details