Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75657428-75657935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX33
Entrez Gene ID
257364
Gene Name
sorting nexin 33
Gene Aliases
SH3PX3, SH3PXD3C, SNX30
UniGene
Hs.8705
Gene Chromosome Location
Chr.15: 75649007-75658627 on Build GRCh38
Transcripts
NM_001318146.1 NM_153271.1

SNP Detail

SNP ID Chromosome Location
rs1003303485 Chr.15: 75657488-75657488 on Build GRCh38
rs561624825 Chr.15: 75657595-75657595 on Build GRCh38
rs568418382 Chr.15: 75657874-75657874 on Build GRCh38
rs117906459 Chr.15: 75657580-75657580 on Build GRCh38
rs766317075 Chr.15: 75657845-75657845 on Build GRCh38
rs929705448 Chr.15: 75657690-75657690 on Build GRCh38
rs1037306455 Chr.15: 75657854-75657854 on Build GRCh38
rs947722707 Chr.15: 75657745-75657745 on Build GRCh38
rs532989681 Chr.15: 75657778-75657778 on Build GRCh38
rs528986701 Chr.15: 75657647-75657647 on Build GRCh38
rs11638663 Chr.15: 75657822-75657822 on Build GRCh38
rs1008218256 Chr.15: 75657619-75657619 on Build GRCh38
rs968599829 Chr.15: 75657732-75657732 on Build GRCh38
rs748411498 Chr.15: 75657592-75657592 on Build GRCh38
rs748497602 Chr.15: 75657620-75657620 on Build GRCh38
rs972211116 Chr.15: 75657657-75657657 on Build GRCh38
rs1022244074 Chr.15: 75657569-75657569 on Build GRCh38
rs906437472 Chr.15: 75657812-75657812 on Build GRCh38
rs956768472 Chr.15: 75657553-75657553 on Build GRCh38
rs115663243 Chr.15: 75657724-75657724 on Build GRCh38
rs940166828 Chr.15: 75657534-75657534 on Build GRCh38
rs959685670 Chr.15: 75657818-75657818 on Build GRCh38
rs770067102 Chr.15: 75657777-75657777 on Build GRCh38
rs1048141447 Chr.15: 75657692-75657692 on Build GRCh38
rs918380525 Chr.15: 75657668-75657668 on Build GRCh38
rs546539846 Chr.15: 75657471-75657471 on Build GRCh38
rs1022339433 Chr.15: 75657862-75657862 on Build GRCh38
rs528076402 Chr.15: 75657568-75657568 on Build GRCh38
rs1031075757 Chr.15: 75657532-75657532 on Build GRCh38
rs767495820 Chr.15: 75657781-75657782 on Build GRCh38
rs867278947 Chr.15: 75657615-75657615 on Build GRCh38
rs1050265508 Chr.15: 75657883-75657883 on Build GRCh38
rs995976616 Chr.15: 75657578-75657578 on Build GRCh38
rs111488993 Chr.15: 75657861-75657861 on Build GRCh38
rs564797292 Chr.15: 75657480-75657480 on Build GRCh38
rs181726098 Chr.15: 75657903-75657903 on Build GRCh38
rs550469350 Chr.15: 75657650-75657650 on Build GRCh38
rs534696911 Chr.15: 75657829-75657829 on Build GRCh38
rs1028412290 Chr.15: 75657583-75657583 on Build GRCh38
rs576427357 Chr.15: 75657856-75657856 on Build GRCh38
rs147725039 Chr.15: 75657793-75657793 on Build GRCh38

View Full Product Details