Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80172924-80173419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAH
Entrez Gene ID
2184
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
-
UniGene
Hs.73875
Gene Chromosome Location
Chr.15: 80152891-80186582 on Build GRCh38
Transcripts
NM_000137.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1374878 c.815T>C p.F272S Chr.15: 80173122-80173122 on Build GRCh38
4057234 c.834G>T p.K278N Chr.15: 80173141-80173141 on Build GRCh38
5530763 c.750C>T p.F250F Chr.15: 80173057-80173057 on Build GRCh38
3794450 c.807C>T p.L269L Chr.15: 80173114-80173114 on Build GRCh38
6657981 c.820G>A p.V274M Chr.15: 80173127-80173127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144228661 Chr.15: 80173022-80173022 on Build GRCh38
rs902678789 Chr.15: 80173297-80173297 on Build GRCh38
rs80338898 Chr.15: 80173089-80173089 on Build GRCh38
rs121965078 Chr.15: 80173143-80173143 on Build GRCh38
rs982400572 Chr.15: 80172960-80172960 on Build GRCh38
rs756267775 Chr.15: 80173075-80173075 on Build GRCh38
rs762398502 Chr.15: 80172979-80172979 on Build GRCh38
rs768365785 Chr.15: 80173196-80173196 on Build GRCh38
rs755673416 Chr.15: 80173181-80173181 on Build GRCh38
rs958151487 Chr.15: 80172949-80172949 on Build GRCh38
rs766663540 Chr.15: 80173079-80173079 on Build GRCh38
rs773134383 Chr.15: 80173140-80173140 on Build GRCh38
rs909723251 Chr.15: 80173076-80173076 on Build GRCh38
rs748945402 Chr.15: 80173185-80173185 on Build GRCh38
rs958099117 Chr.15: 80173209-80173209 on Build GRCh38
rs750572597 Chr.15: 80173066-80173066 on Build GRCh38
rs372980573 Chr.15: 80173138-80173138 on Build GRCh38
rs768315646 Chr.15: 80173254-80173254 on Build GRCh38
rs373850404 Chr.15: 80173182-80173182 on Build GRCh38
rs546505287 Chr.15: 80173378-80173378 on Build GRCh38
rs80338899 Chr.15: 80173093-80173093 on Build GRCh38
rs1019434195 Chr.15: 80173301-80173301 on Build GRCh38
rs757288883 Chr.15: 80173097-80173097 on Build GRCh38
rs369092208 Chr.15: 80173153-80173153 on Build GRCh38
rs959648657 Chr.15: 80173316-80173316 on Build GRCh38
rs749136942 Chr.15: 80173380-80173380 on Build GRCh38
rs1054732462 Chr.15: 80173201-80173201 on Build GRCh38
rs574779745 Chr.15: 80173137-80173137 on Build GRCh38
rs777880578 Chr.15: 80172975-80172975 on Build GRCh38
rs751432965 Chr.15: 80173171-80173171 on Build GRCh38
rs375834291 Chr.15: 80173144-80173144 on Build GRCh38
rs774861939 Chr.15: 80173049-80173049 on Build GRCh38
rs779284513 Chr.15: 80173002-80173004 on Build GRCh38
rs148153674 Chr.15: 80173134-80173134 on Build GRCh38
rs369462541 Chr.15: 80173241-80173241 on Build GRCh38
rs937697712 Chr.15: 80173038-80173038 on Build GRCh38
rs199995781 Chr.15: 80173030-80173030 on Build GRCh38
rs557467122 Chr.15: 80173254-80173264 on Build GRCh38
rs745871490 Chr.15: 80172959-80172959 on Build GRCh38
rs780555815 Chr.15: 80173106-80173106 on Build GRCh38
rs1057516368 Chr.15: 80173051-80173051 on Build GRCh38
rs769303398 Chr.15: 80173115-80173115 on Build GRCh38
rs183545173 Chr.15: 80173264-80173264 on Build GRCh38
rs750741137 Chr.15: 80173049-80173049 on Build GRCh38
rs149052294 Chr.15: 80173013-80173013 on Build GRCh38
rs564024247 Chr.15: 80173261-80173261 on Build GRCh38
rs920286758 Chr.15: 80173349-80173349 on Build GRCh38
rs990465858 Chr.15: 80173323-80173323 on Build GRCh38
rs928228617 Chr.15: 80173300-80173300 on Build GRCh38
rs748802314 Chr.15: 80173207-80173207 on Build GRCh38
rs745435849 Chr.15: 80173107-80173107 on Build GRCh38
rs775814901 Chr.15: 80173020-80173020 on Build GRCh38
rs147099660 Chr.15: 80173103-80173103 on Build GRCh38
rs770358271 Chr.15: 80172994-80172994 on Build GRCh38
rs865953786 Chr.15: 80173389-80173389 on Build GRCh38
rs774774261 Chr.15: 80173135-80173135 on Build GRCh38
rs766656032 Chr.15: 80173152-80173152 on Build GRCh38
rs982734739 Chr.15: 80173289-80173289 on Build GRCh38
rs202041382 Chr.15: 80173063-80173063 on Build GRCh38
rs3752693 Chr.15: 80172958-80172958 on Build GRCh38
rs775156172 Chr.15: 80173127-80173127 on Build GRCh38
rs749865213 Chr.15: 80173176-80173176 on Build GRCh38
rs757121915 Chr.15: 80173175-80173175 on Build GRCh38
rs886286986 Chr.15: 80173123-80173123 on Build GRCh38
rs769550316 Chr.15: 80173016-80173016 on Build GRCh38
rs748523671 Chr.15: 80173383-80173383 on Build GRCh38
rs746464745 Chr.15: 80172978-80172978 on Build GRCh38
rs763974108 Chr.15: 80173170-80173170 on Build GRCh38
rs145851627 Chr.15: 80173048-80173048 on Build GRCh38
rs367884280 Chr.15: 80172968-80172968 on Build GRCh38
rs1028372530 Chr.15: 80172945-80172945 on Build GRCh38
rs1057517436 Chr.15: 80173087-80173088 on Build GRCh38
rs901942273 Chr.15: 80173355-80173355 on Build GRCh38
rs777792671 Chr.15: 80173090-80173090 on Build GRCh38
rs191028698 Chr.15: 80172951-80172951 on Build GRCh38
rs138757552 Chr.15: 80173054-80173054 on Build GRCh38
rs752497048 Chr.15: 80173085-80173085 on Build GRCh38
rs1047529751 Chr.15: 80173393-80173393 on Build GRCh38

View Full Product Details