Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70857618-70858024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYDIN
Entrez Gene ID
54768
Gene Name
HYDIN, axonemal central pair apparatus protein
Gene Aliases
CILD5, HYDIN1, HYDIN2, PPP1R31
UniGene
Hs.351914
Gene Chromosome Location
Chr.16: 70802084-71230722 on Build GRCh38
Transcripts
NM_001270974.2 XM_006721206.2 XM_011523146.2 XM_011523147.1 XM_011523148.1 XM_011523151.1 XM_011523152.1 XM_011523155.2 XM_017023346.1 XM_017023347.1 XM_017023348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
704297 c.12144C>A p.L4048L Chr.16: 70857709-70857709 on Build GRCh38
704303 c.12146T>A p.I4049N Chr.16: 70857707-70857707 on Build GRCh38
704299 c.12145A>G p.I4049V Chr.16: 70857708-70857708 on Build GRCh38
3273607 c.12139C>T p.L4047F Chr.16: 70857714-70857714 on Build GRCh38
3772002 c.12102C>A p.L4034L Chr.16: 70857751-70857751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780905520 Chr.16: 70857672-70857672 on Build GRCh38
rs566274174 Chr.16: 70857644-70857644 on Build GRCh38
rs767092629 Chr.16: 70857772-70857772 on Build GRCh38
rs760429510 Chr.16: 70857774-70857774 on Build GRCh38
rs533701620 Chr.16: 70857652-70857652 on Build GRCh38
rs749284461 Chr.16: 70857695-70857695 on Build GRCh38
rs1002287109 Chr.16: 70857699-70857699 on Build GRCh38
rs750606517 Chr.16: 70857799-70857799 on Build GRCh38
rs779528201 Chr.16: 70857822-70857822 on Build GRCh38
rs750463411 Chr.16: 70857893-70857893 on Build GRCh38
rs16943483 Chr.16: 70857887-70857887 on Build GRCh38
rs757360651 Chr.16: 70857757-70857757 on Build GRCh38
rs766082071 Chr.16: 70857775-70857775 on Build GRCh38
rs368293743 Chr.16: 70857950-70857950 on Build GRCh38
rs777888096 Chr.16: 70857706-70857706 on Build GRCh38
rs761424172 Chr.16: 70857742-70857742 on Build GRCh38
rs954546656 Chr.16: 70857841-70857841 on Build GRCh38
rs750119921 Chr.16: 70857746-70857746 on Build GRCh38
rs374853224 Chr.16: 70857744-70857744 on Build GRCh38
rs775023271 Chr.16: 70857769-70857769 on Build GRCh38
rs770042798 Chr.16: 70857760-70857760 on Build GRCh38
rs760107477 Chr.16: 70857867-70857867 on Build GRCh38
rs757264577 Chr.16: 70857795-70857795 on Build GRCh38
rs757041280 Chr.16: 70857664-70857664 on Build GRCh38
rs781242720 Chr.16: 70857759-70857759 on Build GRCh38
rs755831136 Chr.16: 70857749-70857749 on Build GRCh38
rs781391522 Chr.16: 70857796-70857796 on Build GRCh38
rs374384918 Chr.16: 70857724-70857724 on Build GRCh38
rs771073080 Chr.16: 70857709-70857709 on Build GRCh38
rs112110309 Chr.16: 70857870-70857870 on Build GRCh38
rs945337805 Chr.16: 70857943-70857943 on Build GRCh38
rs936752437 Chr.16: 70857890-70857890 on Build GRCh38
rs556457810 Chr.16: 70857802-70857802 on Build GRCh38
rs536692475 Chr.16: 70857805-70857805 on Build GRCh38
rs367747522 Chr.16: 70857751-70857751 on Build GRCh38
rs779813455 Chr.16: 70857694-70857694 on Build GRCh38
rs769177222 Chr.16: 70857809-70857809 on Build GRCh38
rs368313340 Chr.16: 70857736-70857736 on Build GRCh38
rs1774417 Chr.16: 70857875-70857875 on Build GRCh38
rs770236045 Chr.16: 70857720-70857720 on Build GRCh38
rs758447006 Chr.16: 70857696-70857696 on Build GRCh38
rs575687858 Chr.16: 70857804-70857804 on Build GRCh38
rs1002338159 Chr.16: 70857972-70857972 on Build GRCh38
rs773935602 Chr.16: 70857739-70857739 on Build GRCh38
rs770557117 Chr.16: 70857868-70857868 on Build GRCh38
rs769230375 Chr.16: 70857762-70857762 on Build GRCh38
rs752672507 Chr.16: 70857793-70857793 on Build GRCh38
rs906767908 Chr.16: 70857964-70857964 on Build GRCh38
rs763014973 Chr.16: 70857889-70857889 on Build GRCh38
rs747031668 Chr.16: 70857756-70857756 on Build GRCh38
rs765109053 Chr.16: 70857786-70857786 on Build GRCh38
rs558906994 Chr.16: 70857923-70857923 on Build GRCh38
rs763613808 Chr.16: 70857725-70857725 on Build GRCh38
rs971997630 Chr.16: 70857792-70857792 on Build GRCh38
rs552091529 Chr.16: 70857687-70857687 on Build GRCh38
rs374761324 Chr.16: 70857651-70857651 on Build GRCh38
rs537767167 Chr.16: 70857753-70857753 on Build GRCh38
rs372726695 Chr.16: 70857708-70857708 on Build GRCh38
rs748709280 Chr.16: 70857823-70857823 on Build GRCh38
rs763946125 Chr.16: 70857892-70857892 on Build GRCh38
rs746180502 Chr.16: 70857715-70857715 on Build GRCh38
rs754779221 Chr.16: 70857784-70857784 on Build GRCh38
rs749679799 Chr.16: 70857761-70857761 on Build GRCh38
rs2487972 Chr.16: 70857915-70857915 on Build GRCh38
rs1774416 Chr.16: 70857737-70857737 on Build GRCh38
rs780385647 Chr.16: 70857922-70857922 on Build GRCh38
rs771465983 Chr.16: 70857852-70857852 on Build GRCh38
rs868484151 Chr.16: 70857846-70857846 on Build GRCh38
rs754974697 Chr.16: 70857755-70857755 on Build GRCh38
rs776924229 Chr.16: 70857712-70857712 on Build GRCh38
rs772845454 Chr.16: 70857861-70857861 on Build GRCh38
rs375063220 Chr.16: 70857884-70857884 on Build GRCh38
rs570699554 Chr.16: 70857688-70857688 on Build GRCh38
rs761364396 Chr.16: 70857771-70857771 on Build GRCh38
rs371312960 Chr.16: 70857878-70857878 on Build GRCh38

View Full Product Details