Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7443774-7444273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF11
Entrez Gene ID
2256
Gene Name
fibroblast growth factor 11
Gene Aliases
FGF-11, FHF-3, FHF3
UniGene
Hs.655193
Gene Chromosome Location
Chr.17: 7438273-7444932 on Build GRCh38
Transcripts
NM_001303460.1 NM_004112.3 NR_130156.1

SNP Detail

SNP ID Chromosome Location
rs756297137 Chr.17: 7443999-7443999 on Build GRCh38
rs916670333 Chr.17: 7444250-7444250 on Build GRCh38
rs922038388 Chr.17: 7443992-7443992 on Build GRCh38
rs939449580 Chr.17: 7444237-7444237 on Build GRCh38
rs573460710 Chr.17: 7444192-7444192 on Build GRCh38
rs766538534 Chr.17: 7443890-7443890 on Build GRCh38
rs920608813 Chr.17: 7443909-7443909 on Build GRCh38
rs755479221 Chr.17: 7443878-7443878 on Build GRCh38
rs954560666 Chr.17: 7443996-7443996 on Build GRCh38
rs910354683 Chr.17: 7443875-7443875 on Build GRCh38
rs1001211203 Chr.17: 7443810-7443810 on Build GRCh38
rs895590310 Chr.17: 7444096-7444096 on Build GRCh38
rs1005112138 Chr.17: 7444161-7444161 on Build GRCh38
rs112339226 Chr.17: 7444050-7444050 on Build GRCh38
rs950621752 Chr.17: 7444183-7444183 on Build GRCh38
rs954432678 Chr.17: 7443854-7443854 on Build GRCh38
rs141432540 Chr.17: 7443805-7443805 on Build GRCh38
rs147206656 Chr.17: 7444082-7444082 on Build GRCh38
rs983437882 Chr.17: 7444191-7444191 on Build GRCh38
rs558351801 Chr.17: 7444117-7444117 on Build GRCh38
rs4151134 Chr.17: 7443804-7443804 on Build GRCh38
rs3764433 Chr.17: 7444065-7444065 on Build GRCh38
rs949579283 Chr.17: 7444102-7444102 on Build GRCh38
rs184662875 Chr.17: 7443861-7443861 on Build GRCh38
rs749548990 Chr.17: 7444038-7444038 on Build GRCh38
rs998521894 Chr.17: 7443827-7443827 on Build GRCh38
rs929347496 Chr.17: 7443933-7443933 on Build GRCh38
rs895273892 Chr.17: 7444002-7444002 on Build GRCh38
rs928079695 Chr.17: 7443965-7443965 on Build GRCh38
rs1046965069 Chr.17: 7444107-7444107 on Build GRCh38
rs968752959 Chr.17: 7443826-7443826 on Build GRCh38
rs1047712027 Chr.17: 7443955-7443955 on Build GRCh38
rs1031325672 Chr.17: 7444045-7444045 on Build GRCh38
rs1011162357 Chr.17: 7444014-7444014 on Build GRCh38
rs111650828 Chr.17: 7443983-7443983 on Build GRCh38
rs1016242791 Chr.17: 7443938-7443938 on Build GRCh38
rs1017446037 Chr.17: 7444070-7444070 on Build GRCh38
rs148931304 Chr.17: 7443983-7443984 on Build GRCh38
rs1029074731 Chr.17: 7444195-7444195 on Build GRCh38
rs770860677 Chr.17: 7444194-7444194 on Build GRCh38
rs945921985 Chr.17: 7444034-7444034 on Build GRCh38
rs1021301932 Chr.17: 7443818-7443818 on Build GRCh38
rs754010042 Chr.17: 7443903-7443903 on Build GRCh38
rs58775098 Chr.17: 7444168-7444168 on Build GRCh38
rs1031461518 Chr.17: 7443840-7443840 on Build GRCh38
rs528334555 Chr.17: 7443913-7443913 on Build GRCh38
rs1044411510 Chr.17: 7444018-7444018 on Build GRCh38
rs987327609 Chr.17: 7443858-7443858 on Build GRCh38
rs150396695 Chr.17: 7443879-7443879 on Build GRCh38
rs920542026 Chr.17: 7444136-7444136 on Build GRCh38
rs568061444 Chr.17: 7444030-7444030 on Build GRCh38
rs539573268 Chr.17: 7444108-7444108 on Build GRCh38
rs991275972 Chr.17: 7444241-7444241 on Build GRCh38
rs575653662 Chr.17: 7443917-7443917 on Build GRCh38
rs967203454 Chr.17: 7444252-7444252 on Build GRCh38

View Full Product Details