Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36088438-36088938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL3
Entrez Gene ID
6348
Gene Name
C-C motif chemokine ligand 3
Gene Aliases
G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
UniGene
Hs.514107
Gene Chromosome Location
Chr.17: 36088256-36090160 on Build GRCh38
Transcripts
NM_002983.2

SNP Detail

SNP ID Chromosome Location
rs1049191 Chr.17: 36088473-36088473 on Build GRCh38
rs764051613 Chr.17: 36088771-36088771 on Build GRCh38
rs1055562989 Chr.17: 36088540-36088540 on Build GRCh38
rs925610395 Chr.17: 36088533-36088533 on Build GRCh38
rs757259437 Chr.17: 36088705-36088705 on Build GRCh38
rs775138296 Chr.17: 36088642-36088642 on Build GRCh38
rs570129817 Chr.17: 36088472-36088472 on Build GRCh38
rs753553385 Chr.17: 36088791-36088791 on Build GRCh38
rs41360951 Chr.17: 36088510-36088510 on Build GRCh38
rs768957090 Chr.17: 36088646-36088646 on Build GRCh38
rs548448001 Chr.17: 36088703-36088703 on Build GRCh38
rs755625112 Chr.17: 36088781-36088781 on Build GRCh38
rs769321709 Chr.17: 36088640-36088640 on Build GRCh38
rs897655273 Chr.17: 36088824-36088824 on Build GRCh38
rs1049114 Chr.17: 36088699-36088699 on Build GRCh38
rs569801296 Chr.17: 36088792-36088792 on Build GRCh38
rs776848484 Chr.17: 36088811-36088811 on Build GRCh38
rs373603935 Chr.17: 36088665-36088665 on Build GRCh38
rs778166572 Chr.17: 36088803-36088803 on Build GRCh38
rs770996135 Chr.17: 36088754-36088754 on Build GRCh38
rs1049121 Chr.17: 36088693-36088693 on Build GRCh38
rs572302960 Chr.17: 36088832-36088832 on Build GRCh38
rs913383840 Chr.17: 36088465-36088465 on Build GRCh38
rs556756996 Chr.17: 36088821-36088821 on Build GRCh38
rs187371717 Chr.17: 36088536-36088536 on Build GRCh38
rs758981861 Chr.17: 36088655-36088655 on Build GRCh38
rs767648000 Chr.17: 36088656-36088656 on Build GRCh38
rs540744686 Chr.17: 36088582-36088582 on Build GRCh38
rs772441681 Chr.17: 36088800-36088800 on Build GRCh38
rs774884627 Chr.17: 36088652-36088652 on Build GRCh38
rs182552517 Chr.17: 36088886-36088886 on Build GRCh38
rs532580689 Chr.17: 36088682-36088682 on Build GRCh38
rs753104405 Chr.17: 36088600-36088600 on Build GRCh38
rs748914581 Chr.17: 36088742-36088742 on Build GRCh38
rs751391118 Chr.17: 36088773-36088773 on Build GRCh38
rs201437905 Chr.17: 36088712-36088712 on Build GRCh38
rs3210157 Chr.17: 36088743-36088743 on Build GRCh38
rs967550713 Chr.17: 36088901-36088901 on Build GRCh38
rs34171309 Chr.17: 36088717-36088717 on Build GRCh38
rs1049188 Chr.17: 36088485-36088485 on Build GRCh38
rs555235636 Chr.17: 36088809-36088809 on Build GRCh38
rs779590579 Chr.17: 36088737-36088737 on Build GRCh38
rs749642132 Chr.17: 36088645-36088645 on Build GRCh38
rs749941734 Chr.17: 36088624-36088624 on Build GRCh38
rs780539298 Chr.17: 36088630-36088630 on Build GRCh38
rs1049176 Chr.17: 36088555-36088555 on Build GRCh38
rs759898496 Chr.17: 36088814-36088814 on Build GRCh38
rs578119387 Chr.17: 36088873-36088873 on Build GRCh38
rs574807543 Chr.17: 36088570-36088570 on Build GRCh38
rs540710731 Chr.17: 36088833-36088833 on Build GRCh38
rs369914435 Chr.17: 36088777-36088777 on Build GRCh38
rs147323689 Chr.17: 36088733-36088733 on Build GRCh38
rs779583503 Chr.17: 36088790-36088790 on Build GRCh38
rs554387528 Chr.17: 36088698-36088698 on Build GRCh38
rs1013986556 Chr.17: 36088578-36088578 on Build GRCh38
rs1719130 Chr.17: 36088900-36088900 on Build GRCh38
rs8070375 Chr.17: 36088483-36088483 on Build GRCh38
rs5029408 Chr.17: 36088746-36088746 on Build GRCh38
rs998687647 Chr.17: 36088895-36088895 on Build GRCh38
rs141182065 Chr.17: 36088692-36088692 on Build GRCh38
rs748710179 Chr.17: 36088744-36088744 on Build GRCh38
rs5029409 Chr.17: 36088749-36088749 on Build GRCh38
rs5029407 Chr.17: 36088639-36088639 on Build GRCh38
rs1018732965 Chr.17: 36088869-36088869 on Build GRCh38
rs5029410 Chr.17: 36088805-36088805 on Build GRCh38
rs542527118 Chr.17: 36088877-36088877 on Build GRCh38
rs897648438 Chr.17: 36088584-36088584 on Build GRCh38
rs554356555 Chr.17: 36088834-36088834 on Build GRCh38
rs545841233 Chr.17: 36088865-36088865 on Build GRCh38
rs776769566 Chr.17: 36088756-36088756 on Build GRCh38
rs748516138 Chr.17: 36088794-36088794 on Build GRCh38
rs562905594 Chr.17: 36088870-36088870 on Build GRCh38
rs1049131 Chr.17: 36088687-36088687 on Build GRCh38
rs557795937 Chr.17: 36088883-36088883 on Build GRCh38
rs773703371 Chr.17: 36088748-36088748 on Build GRCh38
rs377681850 Chr.17: 36088761-36088761 on Build GRCh38
rs556132749 Chr.17: 36088780-36088780 on Build GRCh38
rs577304178 Chr.17: 36088838-36088838 on Build GRCh38
rs760749398 Chr.17: 36088674-36088674 on Build GRCh38
rs775237622 Chr.17: 36088534-36088535 on Build GRCh38

View Full Product Details