Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63797927-63798428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX42
Entrez Gene ID
11325
Gene Name
DEAD-box helicase 42
Gene Aliases
DDX42P, RHELP, RNAHP, SF3B8, SF3b125
UniGene
Hs.702010
Gene Chromosome Location
Chr.17: 63773856-63819317 on Build GRCh38
Transcripts
NM_007372.3 NM_203499.2 XM_006721657.1 XM_017024111.1 XM_017024112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249903 c.433A>C p.K145Q Chr.17: 63798098-63798098 on Build GRCh38
2795329 c.428A>T p.N143I Chr.17: 63798093-63798093 on Build GRCh38
2795330 c.429C>T p.N143N Chr.17: 63798094-63798094 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753592017 Chr.17: 63798133-63798133 on Build GRCh38
rs939451116 Chr.17: 63798303-63798303 on Build GRCh38
rs752269401 Chr.17: 63798119-63798119 on Build GRCh38
rs893802954 Chr.17: 63797987-63797987 on Build GRCh38
rs1045114440 Chr.17: 63798261-63798261 on Build GRCh38
rs762002666 Chr.17: 63798033-63798033 on Build GRCh38
rs555863712 Chr.17: 63798143-63798143 on Build GRCh38
rs985116734 Chr.17: 63798174-63798174 on Build GRCh38
rs776325605 Chr.17: 63798207-63798207 on Build GRCh38
rs1017958519 Chr.17: 63798204-63798204 on Build GRCh38
rs768897858 Chr.17: 63798011-63798011 on Build GRCh38
rs758957649 Chr.17: 63798144-63798144 on Build GRCh38
rs548918823 Chr.17: 63798312-63798312 on Build GRCh38
rs561866066 Chr.17: 63798228-63798228 on Build GRCh38
rs776880001 Chr.17: 63798016-63798016 on Build GRCh38
rs934430394 Chr.17: 63798223-63798223 on Build GRCh38
rs1017969078 Chr.17: 63798386-63798389 on Build GRCh38
rs767303829 Chr.17: 63798111-63798111 on Build GRCh38
rs561447556 Chr.17: 63798241-63798241 on Build GRCh38
rs36003342 Chr.17: 63798353-63798354 on Build GRCh38
rs756955368 Chr.17: 63798142-63798142 on Build GRCh38
rs1043866600 Chr.17: 63797971-63797971 on Build GRCh38
rs541548442 Chr.17: 63798206-63798206 on Build GRCh38
rs994532476 Chr.17: 63798171-63798171 on Build GRCh38
rs905673342 Chr.17: 63798064-63798064 on Build GRCh38
rs147394847 Chr.17: 63798334-63798334 on Build GRCh38
rs905360432 Chr.17: 63797982-63797982 on Build GRCh38
rs965126365 Chr.17: 63798212-63798212 on Build GRCh38
rs938188019 Chr.17: 63798264-63798264 on Build GRCh38
rs757820506 Chr.17: 63798139-63798140 on Build GRCh38
rs747748010 Chr.17: 63798272-63798272 on Build GRCh38
rs145700448 Chr.17: 63798038-63798038 on Build GRCh38
rs748722661 Chr.17: 63798388-63798388 on Build GRCh38
rs1006131370 Chr.17: 63798374-63798374 on Build GRCh38
rs952393446 Chr.17: 63798191-63798191 on Build GRCh38
rs1006570484 Chr.17: 63798195-63798195 on Build GRCh38
rs760219361 Chr.17: 63798123-63798123 on Build GRCh38
rs750467291 Chr.17: 63798180-63798180 on Build GRCh38
rs780385921 Chr.17: 63798150-63798150 on Build GRCh38
rs111967757 Chr.17: 63798233-63798233 on Build GRCh38
rs545787369 Chr.17: 63797963-63797963 on Build GRCh38
rs61756260 Chr.17: 63798070-63798070 on Build GRCh38
rs112588937 Chr.17: 63798095-63798095 on Build GRCh38
rs975793271 Chr.17: 63798201-63798201 on Build GRCh38
rs185782867 Chr.17: 63798327-63798327 on Build GRCh38
rs568571501 Chr.17: 63798395-63798395 on Build GRCh38
rs1026277922 Chr.17: 63798320-63798321 on Build GRCh38
rs755357929 Chr.17: 63798235-63798235 on Build GRCh38
rs976879322 Chr.17: 63798221-63798221 on Build GRCh38
rs747011997 Chr.17: 63797992-63797992 on Build GRCh38
rs531976974 Chr.17: 63798299-63798299 on Build GRCh38
rs192408599 Chr.17: 63798162-63798162 on Build GRCh38
rs765513497 Chr.17: 63798140-63798141 on Build GRCh38
rs1026615568 Chr.17: 63798182-63798182 on Build GRCh38
rs758565121 Chr.17: 63798196-63798196 on Build GRCh38
rs948086947 Chr.17: 63797989-63797989 on Build GRCh38
rs375840253 Chr.17: 63798159-63798159 on Build GRCh38
rs759526274 Chr.17: 63798103-63798103 on Build GRCh38
rs774015735 Chr.17: 63798102-63798102 on Build GRCh38
rs990149277 Chr.17: 63798230-63798230 on Build GRCh38
rs74338263 Chr.17: 63798306-63798306 on Build GRCh38
rs763663088 Chr.17: 63798132-63798132 on Build GRCh38
rs888387707 Chr.17: 63798399-63798399 on Build GRCh38
rs564006609 Chr.17: 63798282-63798282 on Build GRCh38
rs548392221 Chr.17: 63798380-63798380 on Build GRCh38
rs780404319 Chr.17: 63798227-63798227 on Build GRCh38
rs951965164 Chr.17: 63798155-63798155 on Build GRCh38

View Full Product Details