Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75757774-75758271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALK1
Entrez Gene ID
2584
Gene Name
galactokinase 1
Gene Aliases
GALK, GK1, HEL-S-19
UniGene
Hs.407966
Gene Chromosome Location
Chr.17: 75757937-75765199 on Build GRCh38
Transcripts
NM_000154.1 NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2 XM_011524752.2

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
UniGene
Hs.632226
Gene Chromosome Location
Chr.17: 75721328-75757819 on Build GRCh38
Transcripts
NM_000154.1 NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2 XM_011524752.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147893 c.1144C>G p.Q382E Chr.17: 75758091-75758091 on Build GRCh38
5363788 c.1141_1144delTCTC p.S381fs*>11 Chr.17: 75758091-75758094 on Build GRCh38
3691820 c.1119C>T p.G373G Chr.17: 75758116-75758116 on Build GRCh38
7282219 c.1089C>T p.H363H Chr.17: 75758228-75758228 on Build GRCh38
6668934 c.1124C>T p.T375I Chr.17: 75758111-75758111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772736008 Chr.17: 75758020-75758020 on Build GRCh38
rs774118588 Chr.17: 75758027-75758027 on Build GRCh38
rs774296016 Chr.17: 75758175-75758175 on Build GRCh38
rs763195503 Chr.17: 75758119-75758119 on Build GRCh38
rs773416475 Chr.17: 75758118-75758118 on Build GRCh38
rs375257344 Chr.17: 75758105-75758105 on Build GRCh38
rs371917340 Chr.17: 75758096-75758096 on Build GRCh38
rs760593029 Chr.17: 75757920-75757920 on Build GRCh38
rs371755069 Chr.17: 75758060-75758060 on Build GRCh38
rs376516996 Chr.17: 75758176-75758176 on Build GRCh38
rs954124595 Chr.17: 75757834-75757834 on Build GRCh38
rs778419485 Chr.17: 75758153-75758153 on Build GRCh38
rs1035183432 Chr.17: 75757845-75757845 on Build GRCh38
rs113161372 Chr.17: 75758168-75758168 on Build GRCh38
rs200502537 Chr.17: 75758228-75758228 on Build GRCh38
rs955465338 Chr.17: 75757910-75757910 on Build GRCh38
rs80270402 Chr.17: 75757909-75757909 on Build GRCh38
rs968092383 Chr.17: 75757873-75757873 on Build GRCh38
rs931037398 Chr.17: 75757963-75757963 on Build GRCh38
rs1045339079 Chr.17: 75757801-75757801 on Build GRCh38
rs73997615 Chr.17: 75758116-75758116 on Build GRCh38
rs751070244 Chr.17: 75758002-75758002 on Build GRCh38
rs536478616 Chr.17: 75758221-75758221 on Build GRCh38
rs527987307 Chr.17: 75758035-75758035 on Build GRCh38
rs892578232 Chr.17: 75757861-75757861 on Build GRCh38
rs116845474 Chr.17: 75758024-75758024 on Build GRCh38
rs778420863 Chr.17: 75758188-75758188 on Build GRCh38
rs768625071 Chr.17: 75758203-75758203 on Build GRCh38
rs919574313 Chr.17: 75758141-75758141 on Build GRCh38
rs953866726 Chr.17: 75758093-75758093 on Build GRCh38
rs571466395 Chr.17: 75757870-75757870 on Build GRCh38
rs532337789 Chr.17: 75758148-75758148 on Build GRCh38
rs376050836 Chr.17: 75758240-75758240 on Build GRCh38
rs753012802 Chr.17: 75758142-75758142 on Build GRCh38
rs754208230 Chr.17: 75758239-75758239 on Build GRCh38
rs150586960 Chr.17: 75758068-75758068 on Build GRCh38
rs553689427 Chr.17: 75757886-75757886 on Build GRCh38
rs779841629 Chr.17: 75758164-75758164 on Build GRCh38
rs1034283634 Chr.17: 75757874-75757874 on Build GRCh38
rs533842408 Chr.17: 75758184-75758184 on Build GRCh38
rs771909437 Chr.17: 75758161-75758161 on Build GRCh38
rs753437577 Chr.17: 75758186-75758186 on Build GRCh38
rs373041566 Chr.17: 75758083-75758083 on Build GRCh38
rs573827457 Chr.17: 75757895-75757895 on Build GRCh38
rs770140211 Chr.17: 75758165-75758165 on Build GRCh38
rs1009641860 Chr.17: 75757867-75757867 on Build GRCh38
rs576088936 Chr.17: 75758000-75758000 on Build GRCh38
rs1056675284 Chr.17: 75758034-75758034 on Build GRCh38
rs743554 Chr.17: 75758167-75758167 on Build GRCh38
rs561312049 Chr.17: 75758044-75758044 on Build GRCh38
rs750536223 Chr.17: 75758053-75758053 on Build GRCh38
rs745780334 Chr.17: 75758154-75758154 on Build GRCh38
rs545046092 Chr.17: 75758023-75758023 on Build GRCh38
rs905357808 Chr.17: 75757985-75757985 on Build GRCh38
rs766965363 Chr.17: 75758125-75758125 on Build GRCh38
rs774310746 Chr.17: 75758089-75758089 on Build GRCh38
rs201811960 Chr.17: 75758115-75758115 on Build GRCh38
rs374566360 Chr.17: 75758227-75758227 on Build GRCh38
rs201663491 Chr.17: 75758205-75758205 on Build GRCh38
rs765368371 Chr.17: 75758050-75758050 on Build GRCh38
rs763207227 Chr.17: 75758201-75758201 on Build GRCh38
rs769296629 Chr.17: 75758016-75758016 on Build GRCh38
rs777202950 Chr.17: 75758082-75758082 on Build GRCh38
rs961142959 Chr.17: 75757970-75757970 on Build GRCh38
rs771092692 Chr.17: 75758173-75758173 on Build GRCh38
rs771852922 Chr.17: 75758109-75758109 on Build GRCh38
rs111033608 Chr.17: 75758091-75758091 on Build GRCh38
rs541755348 Chr.17: 75758036-75758036 on Build GRCh38
rs755750574 Chr.17: 75758147-75758147 on Build GRCh38
rs780967150 Chr.17: 75758007-75758007 on Build GRCh38
rs762512608 Chr.17: 75757980-75757981 on Build GRCh38
rs758343807 Chr.17: 75758192-75758192 on Build GRCh38
rs767851895 Chr.17: 75758140-75758140 on Build GRCh38
rs144570556 Chr.17: 75757877-75757877 on Build GRCh38
rs112611253 Chr.17: 75758185-75758185 on Build GRCh38
rs1003746670 Chr.17: 75757842-75757842 on Build GRCh38
rs370066494 Chr.17: 75758150-75758150 on Build GRCh38
rs996893691 Chr.17: 75758097-75758097 on Build GRCh38
rs755467791 Chr.17: 75758139-75758139 on Build GRCh38
rs747793390 Chr.17: 75758080-75758080 on Build GRCh38
rs549837719 Chr.17: 75758076-75758076 on Build GRCh38
rs771288908 Chr.17: 75758215-75758215 on Build GRCh38
rs747586542 Chr.17: 75758014-75758014 on Build GRCh38
rs939565358 Chr.17: 75758031-75758031 on Build GRCh38
rs776963220 Chr.17: 75758039-75758039 on Build GRCh38
rs542735255 Chr.17: 75757971-75757971 on Build GRCh38
rs755413378 Chr.17: 75758072-75758072 on Build GRCh38
rs1021009749 Chr.17: 75758028-75758028 on Build GRCh38
rs756936329 Chr.17: 75758187-75758187 on Build GRCh38
rs770850103 Chr.17: 75758088-75758088 on Build GRCh38
rs767055363 Chr.17: 75758063-75758063 on Build GRCh38
rs369493085 Chr.17: 75758198-75758198 on Build GRCh38
rs905404770 Chr.17: 75757813-75757813 on Build GRCh38
rs759440799 Chr.17: 75758029-75758029 on Build GRCh38
rs908367115 Chr.17: 75757966-75757966 on Build GRCh38
rs147878654 Chr.17: 75757990-75757990 on Build GRCh38
rs538714933 Chr.17: 75758220-75758220 on Build GRCh38
rs752159077 Chr.17: 75758138-75758138 on Build GRCh38
rs779926252 Chr.17: 75758196-75758196 on Build GRCh38
rs773036056 Chr.17: 75757919-75757919 on Build GRCh38
rs554076462 Chr.17: 75758204-75758204 on Build GRCh38
rs763105416 Chr.17: 75758170-75758170 on Build GRCh38

View Full Product Details