Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38303562-38304102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIF1B
Entrez Gene ID
90522
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases
FinGER8
UniGene
Hs.280741
Gene Chromosome Location
Chr.19: 38303560-38315976 on Build GRCh38
Transcripts
NM_001039671.2 NM_001039672.2 NM_001039673.2 NM_001145461.1 NM_001145462.1 NM_001145463.1

SNP Detail

SNP ID Chromosome Location
rs946289696 Chr.19: 38303936-38303936 on Build GRCh38
rs139850537 Chr.19: 38303983-38303983 on Build GRCh38
rs1018846281 Chr.19: 38303801-38303801 on Build GRCh38
rs983759367 Chr.19: 38303975-38303975 on Build GRCh38
rs150656456 Chr.19: 38303832-38303832 on Build GRCh38
rs946070436 Chr.19: 38303777-38303777 on Build GRCh38
rs35710536 Chr.19: 38303728-38303728 on Build GRCh38
rs114220322 Chr.19: 38303768-38303768 on Build GRCh38
rs906338106 Chr.19: 38303892-38303892 on Build GRCh38
rs1035675261 Chr.19: 38303693-38303693 on Build GRCh38
rs1010171789 Chr.19: 38303906-38303906 on Build GRCh38
rs1002897851 Chr.19: 38303679-38303679 on Build GRCh38
rs147886339 Chr.19: 38303930-38303930 on Build GRCh38
rs944151814 Chr.19: 38303817-38303817 on Build GRCh38
rs939150291 Chr.19: 38304063-38304063 on Build GRCh38
rs909776734 Chr.19: 38303842-38303842 on Build GRCh38
rs918291998 Chr.19: 38303822-38303822 on Build GRCh38
rs779932620 Chr.19: 38303922-38303922 on Build GRCh38
rs891793863 Chr.19: 38303722-38303722 on Build GRCh38
rs374419460 Chr.19: 38304047-38304047 on Build GRCh38
rs546686646 Chr.19: 38303860-38303860 on Build GRCh38
rs866504894 Chr.19: 38303959-38303959 on Build GRCh38
rs1041723051 Chr.19: 38303798-38303798 on Build GRCh38
rs1004716533 Chr.19: 38303881-38303881 on Build GRCh38
rs189444979 Chr.19: 38303819-38303819 on Build GRCh38
rs112065330 Chr.19: 38303879-38303879 on Build GRCh38
rs536849619 Chr.19: 38303783-38303783 on Build GRCh38
rs527530498 Chr.19: 38303927-38303927 on Build GRCh38
rs927496638 Chr.19: 38303709-38303709 on Build GRCh38
rs887690509 Chr.19: 38303872-38303872 on Build GRCh38
rs555344838 Chr.19: 38303803-38303803 on Build GRCh38
rs570884122 Chr.19: 38303699-38303699 on Build GRCh38
rs984040406 Chr.19: 38303839-38303839 on Build GRCh38
rs781366447 Chr.19: 38303737-38303737 on Build GRCh38
rs978049707 Chr.19: 38303940-38303940 on Build GRCh38
rs553792602 Chr.19: 38303760-38303760 on Build GRCh38
rs904801905 Chr.19: 38303919-38303919 on Build GRCh38
rs907655230 Chr.19: 38303613-38303613 on Build GRCh38
rs541168994 Chr.19: 38303864-38303864 on Build GRCh38
rs947874142 Chr.19: 38303647-38303647 on Build GRCh38
rs1025358149 Chr.19: 38303934-38303934 on Build GRCh38
rs542395907 Chr.19: 38303928-38303928 on Build GRCh38
rs981091283 Chr.19: 38304024-38304024 on Build GRCh38
rs1014598450 Chr.19: 38303896-38303896 on Build GRCh38
rs981563908 Chr.19: 38303675-38303675 on Build GRCh38
rs971171485 Chr.19: 38303665-38303665 on Build GRCh38
rs559791499 Chr.19: 38303906-38303906 on Build GRCh38
rs78421293 Chr.19: 38303984-38303984 on Build GRCh38
rs565354207 Chr.19: 38303829-38303829 on Build GRCh38
rs1057134680 Chr.19: 38303897-38303897 on Build GRCh38
rs905916529 Chr.19: 38303670-38303670 on Build GRCh38
rs951741039 Chr.19: 38303590-38303590 on Build GRCh38
rs971441552 Chr.19: 38303900-38303900 on Build GRCh38
rs145255633 Chr.19: 38304053-38304053 on Build GRCh38
rs577128420 Chr.19: 38303841-38303841 on Build GRCh38
rs981232443 Chr.19: 38303921-38303921 on Build GRCh38
rs745953225 Chr.19: 38303924-38303924 on Build GRCh38
rs985619771 Chr.19: 38303773-38303773 on Build GRCh38
rs983069456 Chr.19: 38303597-38303597 on Build GRCh38
rs912583875 Chr.19: 38303975-38303975 on Build GRCh38
rs899918070 Chr.19: 38304007-38304007 on Build GRCh38

View Full Product Details