Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52438307-52438854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF534
Entrez Gene ID
147658
Gene Name
zinc finger protein 534
Gene Aliases
KRBO3
UniGene
Hs.447583
Gene Chromosome Location
Chr.19: 52431414-52451939 on Build GRCh38
Transcripts
NM_001143938.1 NM_001143939.1 NM_001291368.1 NM_001291369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3893288 c.1249C>T p.H417Y Chr.19: 52438670-52438670 on Build GRCh38
7338148 c.1316C>T p.T439I Chr.19: 52438737-52438737 on Build GRCh38
1721080 c.1242G>A p.R414R Chr.19: 52438663-52438663 on Build GRCh38
324499 c.1385A>G p.H462R Chr.19: 52438806-52438806 on Build GRCh38
4464415 c.1332C>T p.I444I Chr.19: 52438753-52438753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750102210 Chr.19: 52438677-52438677 on Build GRCh38
rs761380531 Chr.19: 52438742-52438742 on Build GRCh38
rs373118749 Chr.19: 52438697-52438697 on Build GRCh38
rs113034684 Chr.19: 52438792-52438792 on Build GRCh38
rs988309496 Chr.19: 52438360-52438360 on Build GRCh38
rs766569940 Chr.19: 52438352-52438352 on Build GRCh38
rs752686460 Chr.19: 52438515-52438515 on Build GRCh38
rs758150444 Chr.19: 52438372-52438373 on Build GRCh38
rs767510488 Chr.19: 52438646-52438646 on Build GRCh38
rs540132566 Chr.19: 52438656-52438656 on Build GRCh38
rs780833231 Chr.19: 52438802-52438802 on Build GRCh38
rs748957324 Chr.19: 52438409-52438409 on Build GRCh38
rs267605635 Chr.19: 52438663-52438663 on Build GRCh38
rs768506004 Chr.19: 52438686-52438686 on Build GRCh38
rs777995810 Chr.19: 52438486-52438487 on Build GRCh38
rs568452042 Chr.19: 52438534-52438534 on Build GRCh38
rs763298428 Chr.19: 52438489-52438489 on Build GRCh38
rs199880338 Chr.19: 52438655-52438655 on Build GRCh38
rs376623315 Chr.19: 52438570-52438570 on Build GRCh38
rs1046595541 Chr.19: 52438545-52438545 on Build GRCh38
rs779179010 Chr.19: 52438826-52438826 on Build GRCh38
rs749045275 Chr.19: 52438553-52438553 on Build GRCh38
rs755965954 Chr.19: 52438399-52438399 on Build GRCh38
rs925844752 Chr.19: 52438781-52438781 on Build GRCh38
rs201506254 Chr.19: 52438717-52438717 on Build GRCh38
rs749804564 Chr.19: 52438708-52438708 on Build GRCh38
rs774501334 Chr.19: 52438609-52438609 on Build GRCh38
rs766462078 Chr.19: 52438504-52438504 on Build GRCh38
rs775052830 Chr.19: 52438577-52438577 on Build GRCh38
rs777553862 Chr.19: 52438540-52438540 on Build GRCh38
rs756928217 Chr.19: 52438554-52438554 on Build GRCh38
rs751552185 Chr.19: 52438355-52438355 on Build GRCh38
rs935879951 Chr.19: 52438784-52438784 on Build GRCh38
rs781038015 Chr.19: 52438367-52438367 on Build GRCh38
rs918999856 Chr.19: 52438524-52438524 on Build GRCh38
rs980956391 Chr.19: 52438368-52438368 on Build GRCh38
rs867851485 Chr.19: 52438362-52438362 on Build GRCh38
rs1025540965 Chr.19: 52438361-52438361 on Build GRCh38
rs773035527 Chr.19: 52438788-52438789 on Build GRCh38
rs1051620458 Chr.19: 52438542-52438542 on Build GRCh38
rs372365866 Chr.19: 52438392-52438392 on Build GRCh38
rs760641478 Chr.19: 52438653-52438653 on Build GRCh38
rs769117783 Chr.19: 52438787-52438788 on Build GRCh38
rs780644789 Chr.19: 52438659-52438660 on Build GRCh38
rs926292731 Chr.19: 52438372-52438372 on Build GRCh38
rs759787666 Chr.19: 52438625-52438625 on Build GRCh38
rs377150271 Chr.19: 52438619-52438619 on Build GRCh38
rs764128976 Chr.19: 52438735-52438735 on Build GRCh38
rs772300259 Chr.19: 52438723-52438723 on Build GRCh38
rs867435181 Chr.19: 52438813-52438813 on Build GRCh38
rs768257937 Chr.19: 52438695-52438695 on Build GRCh38
rs781131781 Chr.19: 52438497-52438498 on Build GRCh38
rs751781020 Chr.19: 52438506-52438506 on Build GRCh38
rs780014293 Chr.19: 52438647-52438647 on Build GRCh38
rs778766299 Chr.19: 52438666-52438666 on Build GRCh38
rs777720150 Chr.19: 52438536-52438536 on Build GRCh38
rs748668130 Chr.19: 52438578-52438578 on Build GRCh38
rs376234098 Chr.19: 52438827-52438827 on Build GRCh38
rs1366259 Chr.19: 52438594-52438594 on Build GRCh38
rs902366618 Chr.19: 52438628-52438628 on Build GRCh38
rs942136569 Chr.19: 52438465-52438465 on Build GRCh38
rs757399041 Chr.19: 52438494-52438495 on Build GRCh38
rs188181551 Chr.19: 52438349-52438349 on Build GRCh38
rs751758326 Chr.19: 52438490-52438490 on Build GRCh38
rs758006379 Chr.19: 52438790-52438790 on Build GRCh38
rs749770278 Chr.19: 52438817-52438817 on Build GRCh38
rs745314449 Chr.19: 52438447-52438447 on Build GRCh38
rs866689119 Chr.19: 52438786-52438786 on Build GRCh38
rs773946722 Chr.19: 52438427-52438427 on Build GRCh38
rs973690608 Chr.19: 52438681-52438681 on Build GRCh38
rs761798845 Chr.19: 52438758-52438758 on Build GRCh38
rs558022035 Chr.19: 52438774-52438774 on Build GRCh38
rs531814574 Chr.19: 52438507-52438507 on Build GRCh38
rs770219049 Chr.19: 52438611-52438612 on Build GRCh38
rs550315661 Chr.19: 52438531-52438531 on Build GRCh38
rs879234100 Chr.19: 52438600-52438600 on Build GRCh38
rs529331549 Chr.19: 52438580-52438580 on Build GRCh38
rs941696732 Chr.19: 52438378-52438378 on Build GRCh38
rs767626341 Chr.19: 52438514-52438514 on Build GRCh38
rs374462017 Chr.19: 52438822-52438822 on Build GRCh38
rs773645152 Chr.19: 52438473-52438473 on Build GRCh38
rs752790421 Chr.19: 52438744-52438746 on Build GRCh38
rs371048121 Chr.19: 52438793-52438793 on Build GRCh38
rs753839602 Chr.19: 52438736-52438736 on Build GRCh38
rs372171741 Chr.19: 52438459-52438459 on Build GRCh38
rs35035043 Chr.19: 52438579-52438580 on Build GRCh38
rs773733386 Chr.19: 52438587-52438587 on Build GRCh38
rs757042185 Chr.19: 52438657-52438657 on Build GRCh38
rs746449044 Chr.19: 52438499-52438500 on Build GRCh38
rs267605633 Chr.19: 52438334-52438334 on Build GRCh38
rs749690078 Chr.19: 52438746-52438746 on Build GRCh38
rs1042312603 Chr.19: 52438824-52438824 on Build GRCh38
rs1010038184 Chr.19: 52438623-52438623 on Build GRCh38
rs777535380 Chr.19: 52438403-52438403 on Build GRCh38
rs973160353 Chr.19: 52438491-52438491 on Build GRCh38
rs759684444 Chr.19: 52438722-52438722 on Build GRCh38
rs756302062 Chr.19: 52438522-52438522 on Build GRCh38
rs774717255 Chr.19: 52438719-52438719 on Build GRCh38
rs372600849 Chr.19: 52438438-52438438 on Build GRCh38
rs760796766 Chr.19: 52438726-52438726 on Build GRCh38
rs770506801 Chr.19: 52438423-52438423 on Build GRCh38
rs376112270 Chr.19: 52438725-52438725 on Build GRCh38
rs746468806 Chr.19: 52438694-52438694 on Build GRCh38
rs267605634 Chr.19: 52438479-52438479 on Build GRCh38
rs779900642 Chr.19: 52438791-52438791 on Build GRCh38
rs368394896 Chr.19: 52438690-52438690 on Build GRCh38
rs765709124 Chr.19: 52438471-52438471 on Build GRCh38
rs750191434 Chr.19: 52438776-52438776 on Build GRCh38
rs911683843 Chr.19: 52438546-52438546 on Build GRCh38
rs866921294 Chr.19: 52438685-52438685 on Build GRCh38
rs771292593 Chr.19: 52438595-52438595 on Build GRCh38
rs746199476 Chr.19: 52438828-52438828 on Build GRCh38
rs1366258 Chr.19: 52438365-52438365 on Build GRCh38
rs866552251 Chr.19: 52438412-52438412 on Build GRCh38
rs778481309 Chr.19: 52438557-52438557 on Build GRCh38
rs201395526 Chr.19: 52438574-52438574 on Build GRCh38
rs998935403 Chr.19: 52438641-52438641 on Build GRCh38
rs777044212 Chr.19: 52438467-52438467 on Build GRCh38
rs751992255 Chr.19: 52438340-52438340 on Build GRCh38
rs754598339 Chr.19: 52438798-52438798 on Build GRCh38
rs879115374 Chr.19: 52438618-52438618 on Build GRCh38

View Full Product Details