Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55075400-55075942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L1
Entrez Gene ID
54869
Gene Name
EPS8 like 1
Gene Aliases
DRC3, EPS8R1, PP10566
UniGene
Hs.438862
Gene Chromosome Location
Chr.19: 55075853-55087923 on Build GRCh38
Transcripts
NM_133180.2 XM_005259020.1 XM_011527051.2 XM_011527052.2

SNP Detail

SNP ID Chromosome Location
rs145300507 Chr.19: 55075770-55075770 on Build GRCh38
rs931025746 Chr.19: 55075864-55075864 on Build GRCh38
rs1023831931 Chr.19: 55075766-55075766 on Build GRCh38
rs960868947 Chr.19: 55075589-55075589 on Build GRCh38
rs370773431 Chr.19: 55075765-55075765 on Build GRCh38
rs1041071715 Chr.19: 55075702-55075702 on Build GRCh38
rs1008459085 Chr.19: 55075701-55075701 on Build GRCh38
rs928264209 Chr.19: 55075577-55075577 on Build GRCh38
rs964144336 Chr.19: 55075449-55075449 on Build GRCh38
rs913130110 Chr.19: 55075535-55075535 on Build GRCh38
rs62124193 Chr.19: 55075436-55075436 on Build GRCh38
rs886681354 Chr.19: 55075571-55075571 on Build GRCh38
rs892983801 Chr.19: 55075745-55075745 on Build GRCh38
rs1019595816 Chr.19: 55075444-55075444 on Build GRCh38
rs536980653 Chr.19: 55075686-55075686 on Build GRCh38
rs563393511 Chr.19: 55075481-55075481 on Build GRCh38
rs1020508349 Chr.19: 55075768-55075768 on Build GRCh38
rs967269146 Chr.19: 55075820-55075820 on Build GRCh38
rs529204534 Chr.19: 55075555-55075555 on Build GRCh38
rs551005190 Chr.19: 55075649-55075649 on Build GRCh38
rs746554401 Chr.19: 55075551-55075551 on Build GRCh38
rs12460661 Chr.19: 55075703-55075703 on Build GRCh38
rs982987473 Chr.19: 55075865-55075865 on Build GRCh38
rs1030641257 Chr.19: 55075814-55075814 on Build GRCh38
rs1056517269 Chr.19: 55075890-55075890 on Build GRCh38
rs1011396097 Chr.19: 55075756-55075756 on Build GRCh38
rs908672850 Chr.19: 55075891-55075891 on Build GRCh38
rs967808228 Chr.19: 55075769-55075769 on Build GRCh38
rs571665868 Chr.19: 55075608-55075608 on Build GRCh38
rs896774520 Chr.19: 55075759-55075759 on Build GRCh38
rs940679129 Chr.19: 55075887-55075887 on Build GRCh38
rs1028095327 Chr.19: 55075505-55075505 on Build GRCh38
rs1039552682 Chr.19: 55075691-55075691 on Build GRCh38
rs115839249 Chr.19: 55075480-55075480 on Build GRCh38
rs996970385 Chr.19: 55075729-55075729 on Build GRCh38
rs930722311 Chr.19: 55075859-55075859 on Build GRCh38
rs773771384 Chr.19: 55075590-55075590 on Build GRCh38
rs12460187 Chr.19: 55075422-55075422 on Build GRCh38
rs981288589 Chr.19: 55075823-55075823 on Build GRCh38
rs1029253422 Chr.19: 55075472-55075472 on Build GRCh38
rs182342096 Chr.19: 55075593-55075593 on Build GRCh38
rs1035352436 Chr.19: 55075825-55075825 on Build GRCh38
rs887379386 Chr.19: 55075672-55075672 on Build GRCh38
rs577262476 Chr.19: 55075462-55075462 on Build GRCh38
rs919670946 Chr.19: 55075849-55075849 on Build GRCh38
rs996896559 Chr.19: 55075457-55075457 on Build GRCh38
rs1047789588 Chr.19: 55075660-55075660 on Build GRCh38
rs557381609 Chr.19: 55075866-55075866 on Build GRCh38
rs991668456 Chr.19: 55075837-55075837 on Build GRCh38
rs981087158 Chr.19: 55075567-55075567 on Build GRCh38
rs534406868 Chr.19: 55075860-55075860 on Build GRCh38
rs900631388 Chr.19: 55075476-55075476 on Build GRCh38
rs975206030 Chr.19: 55075836-55075836 on Build GRCh38
rs990512292 Chr.19: 55075500-55075500 on Build GRCh38
rs920126678 Chr.19: 55075631-55075631 on Build GRCh38
rs942531430 Chr.19: 55075687-55075687 on Build GRCh38
rs80237352 Chr.19: 55075734-55075734 on Build GRCh38
rs967047627 Chr.19: 55075548-55075548 on Build GRCh38
rs577326281 Chr.19: 55075902-55075902 on Build GRCh38
rs187016607 Chr.19: 55075667-55075667 on Build GRCh38
rs370635508 Chr.19: 55075651-55075651 on Build GRCh38
rs530881745 Chr.19: 55075646-55075646 on Build GRCh38
rs1005097102 Chr.19: 55075719-55075719 on Build GRCh38
rs770155156 Chr.19: 55075616-55075616 on Build GRCh38
rs752584039 Chr.19: 55075817-55075818 on Build GRCh38
rs979101499 Chr.19: 55075610-55075610 on Build GRCh38
rs191088759 Chr.19: 55075582-55075582 on Build GRCh38
rs958361675 Chr.19: 55075826-55075826 on Build GRCh38
rs374040484 Chr.19: 55075699-55075699 on Build GRCh38
rs578096382 Chr.19: 55075642-55075642 on Build GRCh38
rs187830644 Chr.19: 55075568-55075568 on Build GRCh38
rs577890399 Chr.19: 55075703-55075706 on Build GRCh38
rs986465819 Chr.19: 55075824-55075824 on Build GRCh38
rs72402545 Chr.19: 55075812-55075817 on Build GRCh38
rs1049616707 Chr.19: 55075863-55075863 on Build GRCh38
rs150039139 Chr.19: 55075579-55075579 on Build GRCh38

View Full Product Details