Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55228998-55229497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM86B
Entrez Gene ID
255043
Gene Name
transmembrane protein 86B
Gene Aliases
-
UniGene
Hs.135215
Gene Chromosome Location
Chr.19: 55226634-55229264 on Build GRCh38
Transcripts
NM_173804.4

SNP Detail

SNP ID Chromosome Location
rs148786407 Chr.19: 55229153-55229153 on Build GRCh38
rs999691769 Chr.19: 55229277-55229277 on Build GRCh38
rs898512611 Chr.19: 55229051-55229051 on Build GRCh38
rs977126314 Chr.19: 55229138-55229139 on Build GRCh38
rs916703297 Chr.19: 55229136-55229136 on Build GRCh38
rs1013461896 Chr.19: 55229326-55229326 on Build GRCh38
rs796503006 Chr.19: 55229445-55229445 on Build GRCh38
rs369312543 Chr.19: 55229200-55229200 on Build GRCh38
rs569962685 Chr.19: 55229095-55229095 on Build GRCh38
rs749489372 Chr.19: 55229411-55229411 on Build GRCh38
rs984870954 Chr.19: 55229443-55229443 on Build GRCh38
rs556430411 Chr.19: 55229076-55229076 on Build GRCh38
rs920421860 Chr.19: 55229398-55229398 on Build GRCh38
rs961151895 Chr.19: 55229296-55229296 on Build GRCh38
rs943220876 Chr.19: 55229386-55229386 on Build GRCh38
rs928099315 Chr.19: 55229119-55229119 on Build GRCh38
rs907836183 Chr.19: 55229356-55229356 on Build GRCh38
rs896757138 Chr.19: 55229413-55229413 on Build GRCh38
rs901004687 Chr.19: 55229184-55229184 on Build GRCh38
rs991307419 Chr.19: 55229127-55229127 on Build GRCh38
rs931427522 Chr.19: 55229401-55229401 on Build GRCh38
rs1023775749 Chr.19: 55229434-55229434 on Build GRCh38
rs750007598 Chr.19: 55229121-55229121 on Build GRCh38
rs952138698 Chr.19: 55229330-55229330 on Build GRCh38
rs904672009 Chr.19: 55229458-55229458 on Build GRCh38
rs960861854 Chr.19: 55229122-55229122 on Build GRCh38
rs902193448 Chr.19: 55229268-55229268 on Build GRCh38
rs914291230 Chr.19: 55229101-55229101 on Build GRCh38
rs1010601495 Chr.19: 55229065-55229065 on Build GRCh38
rs3826885 Chr.19: 55229359-55229359 on Build GRCh38
rs1011093665 Chr.19: 55229237-55229237 on Build GRCh38
rs1035891127 Chr.19: 55229071-55229071 on Build GRCh38
rs1003528451 Chr.19: 55229066-55229066 on Build GRCh38
rs761914330 Chr.19: 55229390-55229390 on Build GRCh38
rs547464200 Chr.19: 55229388-55229388 on Build GRCh38
rs758265581 Chr.19: 55229185-55229185 on Build GRCh38
rs946451054 Chr.19: 55229389-55229389 on Build GRCh38
rs1022953879 Chr.19: 55229098-55229098 on Build GRCh38
rs776737586 Chr.19: 55229385-55229385 on Build GRCh38
rs922544380 Chr.19: 55229370-55229370 on Build GRCh38
rs978323740 Chr.19: 55229104-55229104 on Build GRCh38
rs76284829 Chr.19: 55229341-55229341 on Build GRCh38
rs546157525 Chr.19: 55229304-55229304 on Build GRCh38
rs527757165 Chr.19: 55229372-55229372 on Build GRCh38
rs1049084165 Chr.19: 55229138-55229138 on Build GRCh38
rs139324437 Chr.19: 55229315-55229315 on Build GRCh38
rs966912683 Chr.19: 55229100-55229100 on Build GRCh38
rs910429358 Chr.19: 55229384-55229384 on Build GRCh38
rs959108344 Chr.19: 55229075-55229075 on Build GRCh38
rs930625480 Chr.19: 55229137-55229137 on Build GRCh38
rs1039796506 Chr.19: 55229258-55229258 on Build GRCh38
rs902183710 Chr.19: 55229403-55229403 on Build GRCh38
rs968377372 Chr.19: 55229113-55229113 on Build GRCh38
rs1021797959 Chr.19: 55229331-55229331 on Build GRCh38
rs553833971 Chr.19: 55229252-55229252 on Build GRCh38
rs931396440 Chr.19: 55229283-55229283 on Build GRCh38
rs953837504 Chr.19: 55229057-55229057 on Build GRCh38
rs568203385 Chr.19: 55229118-55229118 on Build GRCh38
rs142295142 Chr.19: 55229225-55229225 on Build GRCh38
rs535998684 Chr.19: 55229097-55229097 on Build GRCh38
rs887097382 Chr.19: 55229289-55229289 on Build GRCh38
rs771243425 Chr.19: 55229417-55229417 on Build GRCh38
rs572373648 Chr.19: 55229465-55229465 on Build GRCh38
rs1049559850 Chr.19: 55229379-55229379 on Build GRCh38
rs779976640 Chr.19: 55229187-55229187 on Build GRCh38
rs539871518 Chr.19: 55229198-55229198 on Build GRCh38
rs972638740 Chr.19: 55229360-55229360 on Build GRCh38
rs760031087 Chr.19: 55229159-55229170 on Build GRCh38
rs562766073 Chr.19: 55229314-55229314 on Build GRCh38
rs189453932 Chr.19: 55229191-55229191 on Build GRCh38
rs939885099 Chr.19: 55229162-55229162 on Build GRCh38
rs894758261 Chr.19: 55229439-55229439 on Build GRCh38
rs553903019 Chr.19: 55229422-55229422 on Build GRCh38
rs151242238 Chr.19: 55229292-55229292 on Build GRCh38
rs58957726 Chr.19: 55229429-55229429 on Build GRCh38
rs999179810 Chr.19: 55229404-55229404 on Build GRCh38
rs1025853802 Chr.19: 55229063-55229063 on Build GRCh38
rs572454877 Chr.19: 55229312-55229312 on Build GRCh38
rs146568705 Chr.19: 55229441-55229441 on Build GRCh38
rs889569725 Chr.19: 55229192-55229192 on Build GRCh38
rs1043455926 Chr.19: 55229402-55229402 on Build GRCh38
rs1047087723 Chr.19: 55229286-55229286 on Build GRCh38
rs1036953961 Chr.19: 55229166-55229166 on Build GRCh38

View Full Product Details