Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46356638-46357150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPAS1
Entrez Gene ID
2034
Gene Name
endothelial PAS domain protein 1
Gene Aliases
ECYT4, HIF2A, HLF, MOP2, PASD2, bHLHe73
UniGene
Hs.468410
Gene Chromosome Location
Chr.2: 46297402-46386703 on Build GRCh38
Transcripts
NM_001430.4 XM_011532698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3799030 c.447C>G p.L149L Chr.2: 46356801-46356801 on Build GRCh38
3391744 c.454+5C>T p.? Chr.2: 46356813-46356813 on Build GRCh38
122853 c.430C>T p.R144C Chr.2: 46356784-46356784 on Build GRCh38
1021168 c.435G>T p.E145D Chr.2: 46356789-46356789 on Build GRCh38
5045132 c.376C>A p.L126I Chr.2: 46356730-46356730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998493494 Chr.2: 46356945-46356945 on Build GRCh38
rs1056934942 Chr.2: 46356976-46356976 on Build GRCh38
rs761056194 Chr.2: 46356843-46356843 on Build GRCh38
rs371776927 Chr.2: 46356874-46356874 on Build GRCh38
rs926748370 Chr.2: 46356964-46356964 on Build GRCh38
rs760887233 Chr.2: 46356688-46356688 on Build GRCh38
rs139599383 Chr.2: 46356682-46356682 on Build GRCh38
rs17035050 Chr.2: 46356951-46356951 on Build GRCh38
rs763646291 Chr.2: 46356742-46356742 on Build GRCh38
rs1005443589 Chr.2: 46356689-46356689 on Build GRCh38
rs897941068 Chr.2: 46357042-46357042 on Build GRCh38
rs542793806 Chr.2: 46356952-46356952 on Build GRCh38
rs570779627 Chr.2: 46356875-46356875 on Build GRCh38
rs745690526 Chr.2: 46356773-46356773 on Build GRCh38
rs909788435 Chr.2: 46357064-46357064 on Build GRCh38
rs563755754 Chr.2: 46356940-46356940 on Build GRCh38
rs941184834 Chr.2: 46356871-46356871 on Build GRCh38
rs773238476 Chr.2: 46356839-46356839 on Build GRCh38
rs976800461 Chr.2: 46356989-46356989 on Build GRCh38
rs946926515 Chr.2: 46356949-46356949 on Build GRCh38
rs368194877 Chr.2: 46356834-46356834 on Build GRCh38
rs994003553 Chr.2: 46357068-46357068 on Build GRCh38
rs766872843 Chr.2: 46356702-46356702 on Build GRCh38
rs374636224 Chr.2: 46356768-46356768 on Build GRCh38
rs749287823 Chr.2: 46356812-46356812 on Build GRCh38
rs780172672 Chr.2: 46356804-46356804 on Build GRCh38
rs768682583 Chr.2: 46356823-46356823 on Build GRCh38
rs548278993 Chr.2: 46356988-46356988 on Build GRCh38
rs1049028581 Chr.2: 46357097-46357097 on Build GRCh38
rs887991380 Chr.2: 46356747-46356748 on Build GRCh38
rs371978750 Chr.2: 46356855-46356855 on Build GRCh38
rs949765147 Chr.2: 46356931-46356931 on Build GRCh38
rs922559254 Chr.2: 46356703-46356703 on Build GRCh38
rs200773236 Chr.2: 46356745-46356745 on Build GRCh38
rs983211947 Chr.2: 46357032-46357032 on Build GRCh38
rs902827681 Chr.2: 46356942-46356942 on Build GRCh38
rs544567363 Chr.2: 46356756-46356756 on Build GRCh38
rs755906196 Chr.2: 46356777-46356777 on Build GRCh38
rs774555938 Chr.2: 46356831-46356831 on Build GRCh38
rs773681234 Chr.2: 46356683-46356683 on Build GRCh38
rs754298535 Chr.2: 46356715-46356715 on Build GRCh38
rs34136947 Chr.2: 46356920-46356920 on Build GRCh38
rs951337085 Chr.2: 46357020-46357020 on Build GRCh38
rs757929699 Chr.2: 46356716-46356716 on Build GRCh38
rs939576510 Chr.2: 46356969-46356969 on Build GRCh38
rs772349615 Chr.2: 46356835-46356835 on Build GRCh38
rs1029767431 Chr.2: 46356996-46356996 on Build GRCh38
rs766676935 Chr.2: 46356853-46356853 on Build GRCh38
rs757004008 Chr.2: 46356767-46356767 on Build GRCh38
rs868804269 Chr.2: 46357092-46357092 on Build GRCh38
rs544076595 Chr.2: 46356937-46356937 on Build GRCh38
rs976749570 Chr.2: 46356667-46356667 on Build GRCh38
rs956555374 Chr.2: 46356895-46356895 on Build GRCh38
rs964212202 Chr.2: 46356985-46356985 on Build GRCh38

View Full Product Details