Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46616892-46617396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIPT
Entrez Gene ID
9419
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Gene Aliases
HSPC139, SSMDF
UniGene
Hs.133998
Gene Chromosome Location
Chr.2: 46617172-46630176 on Build GRCh38
Transcripts
NM_014171.5 NM_002643.3 NM_173074.2 XM_005264369.2 XM_011532908.2

Gene Symbol
PIGF
Entrez Gene ID
5281
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Gene Aliases
-
UniGene
Hs.468415
Gene Chromosome Location
Chr.2: 46581274-46617112 on Build GRCh38
Transcripts
NM_014171.5 NM_002643.3 NM_173074.2 XM_005264369.2 XM_011532908.2

SNP Detail

SNP ID Chromosome Location
rs947278261 Chr.2: 46617017-46617017 on Build GRCh38
rs768610241 Chr.2: 46617327-46617327 on Build GRCh38
rs957216644 Chr.2: 46616933-46616933 on Build GRCh38
rs1051243330 Chr.2: 46617137-46617137 on Build GRCh38
rs970915260 Chr.2: 46617235-46617235 on Build GRCh38
rs545395277 Chr.2: 46616970-46616970 on Build GRCh38
rs556588511 Chr.2: 46616930-46616930 on Build GRCh38
rs987805803 Chr.2: 46616946-46616946 on Build GRCh38
rs778036285 Chr.2: 46617304-46617304 on Build GRCh38
rs77297964 Chr.2: 46617213-46617213 on Build GRCh38
rs187894233 Chr.2: 46617247-46617247 on Build GRCh38
rs192177022 Chr.2: 46617254-46617254 on Build GRCh38
rs373698985 Chr.2: 46617349-46617349 on Build GRCh38
rs1001854402 Chr.2: 46617197-46617197 on Build GRCh38
rs909128856 Chr.2: 46617234-46617234 on Build GRCh38
rs528159996 Chr.2: 46617032-46617032 on Build GRCh38
rs760198395 Chr.2: 46617041-46617041 on Build GRCh38
rs184385563 Chr.2: 46617351-46617351 on Build GRCh38
rs758198081 Chr.2: 46617235-46617237 on Build GRCh38
rs745445025 Chr.2: 46616974-46616974 on Build GRCh38
rs975370340 Chr.2: 46617253-46617253 on Build GRCh38
rs967714111 Chr.2: 46617066-46617066 on Build GRCh38
rs747818861 Chr.2: 46617253-46617254 on Build GRCh38
rs867071154 Chr.2: 46617127-46617127 on Build GRCh38
rs749422145 Chr.2: 46617322-46617322 on Build GRCh38
rs1038997346 Chr.2: 46617028-46617028 on Build GRCh38
rs928375683 Chr.2: 46616988-46616988 on Build GRCh38
rs960417518 Chr.2: 46617209-46617209 on Build GRCh38
rs909850003 Chr.2: 46617110-46617110 on Build GRCh38
rs888461823 Chr.2: 46617009-46617009 on Build GRCh38
rs1022406718 Chr.2: 46616950-46616950 on Build GRCh38
rs780983285 Chr.2: 46617136-46617136 on Build GRCh38
rs543580517 Chr.2: 46616962-46616962 on Build GRCh38
rs892630840 Chr.2: 46617149-46617149 on Build GRCh38
rs939159743 Chr.2: 46616999-46616999 on Build GRCh38
rs397934088 Chr.2: 46617081-46617082 on Build GRCh38
rs866531117 Chr.2: 46617194-46617194 on Build GRCh38
rs5830913 Chr.2: 46616943-46616944 on Build GRCh38
rs369643223 Chr.2: 46617252-46617252 on Build GRCh38
rs981092150 Chr.2: 46616958-46616958 on Build GRCh38
rs919164730 Chr.2: 46617001-46617001 on Build GRCh38
rs757078301 Chr.2: 46617290-46617290 on Build GRCh38
rs765057433 Chr.2: 46617190-46617190 on Build GRCh38
rs562634136 Chr.2: 46617078-46617078 on Build GRCh38
rs751165866 Chr.2: 46617281-46617281 on Build GRCh38
rs1016427589 Chr.2: 46617047-46617047 on Build GRCh38
rs921815424 Chr.2: 46617155-46617155 on Build GRCh38
rs941355325 Chr.2: 46617114-46617114 on Build GRCh38
rs533305710 Chr.2: 46617093-46617093 on Build GRCh38
rs762347127 Chr.2: 46617262-46617262 on Build GRCh38
rs567807980 Chr.2: 46617191-46617191 on Build GRCh38
rs537008712 Chr.2: 46617234-46617235 on Build GRCh38
rs974537445 Chr.2: 46617257-46617257 on Build GRCh38
rs549516850 Chr.2: 46617158-46617158 on Build GRCh38
rs201326006 Chr.2: 46616941-46616942 on Build GRCh38
rs921329108 Chr.2: 46617124-46617124 on Build GRCh38
rs900484537 Chr.2: 46617031-46617031 on Build GRCh38
rs376513299 Chr.2: 46617256-46617256 on Build GRCh38
rs1004957947 Chr.2: 46617204-46617204 on Build GRCh38
rs113423551 Chr.2: 46617108-46617108 on Build GRCh38
rs374562794 Chr.2: 46617007-46617007 on Build GRCh38
rs753835708 Chr.2: 46617041-46617041 on Build GRCh38
rs746893919 Chr.2: 46617242-46617253 on Build GRCh38
rs775762100 Chr.2: 46617012-46617019 on Build GRCh38
rs773862266 Chr.2: 46617243-46617243 on Build GRCh38
rs1009789423 Chr.2: 46617183-46617183 on Build GRCh38
rs886489763 Chr.2: 46617040-46617040 on Build GRCh38
rs563782489 Chr.2: 46617008-46617008 on Build GRCh38
rs904093924 Chr.2: 46617361-46617361 on Build GRCh38
rs375640576 Chr.2: 46616942-46616943 on Build GRCh38
rs539969299 Chr.2: 46617034-46617034 on Build GRCh38
rs1051727498 Chr.2: 46617192-46617192 on Build GRCh38
rs534257406 Chr.2: 46617115-46617115 on Build GRCh38
rs779932571 Chr.2: 46617321-46617321 on Build GRCh38
rs970967685 Chr.2: 46617059-46617059 on Build GRCh38
rs926411451 Chr.2: 46616987-46616987 on Build GRCh38
rs780790471 Chr.2: 46617297-46617297 on Build GRCh38
rs1010252728 Chr.2: 46617057-46617057 on Build GRCh38
rs907853515 Chr.2: 46617313-46617313 on Build GRCh38
rs111261981 Chr.2: 46617255-46617255 on Build GRCh38
rs1051624903 Chr.2: 46617315-46617315 on Build GRCh38
rs192666754 Chr.2: 46616919-46616919 on Build GRCh38
rs574227348 Chr.2: 46617252-46617253 on Build GRCh38
rs561884991 Chr.2: 46617035-46617035 on Build GRCh38
rs575130522 Chr.2: 46616960-46616960 on Build GRCh38
rs550802716 Chr.2: 46617050-46617050 on Build GRCh38
rs67893203 Chr.2: 46616944-46616945 on Build GRCh38
rs756000762 Chr.2: 46617311-46617311 on Build GRCh38
rs771386179 Chr.2: 46616944-46616944 on Build GRCh38
rs914023155 Chr.2: 46617326-46617326 on Build GRCh38
rs1018372101 Chr.2: 46616915-46616915 on Build GRCh38
rs6761704 Chr.2: 46617330-46617330 on Build GRCh38
rs950612655 Chr.2: 46617221-46617221 on Build GRCh38
rs1043926539 Chr.2: 46617353-46617353 on Build GRCh38
rs900011418 Chr.2: 46616917-46616917 on Build GRCh38
rs964621661 Chr.2: 46617251-46617251 on Build GRCh38
rs960470070 Chr.2: 46617080-46617080 on Build GRCh38
rs984672459 Chr.2: 46617232-46617232 on Build GRCh38
rs1001905103 Chr.2: 46617073-46617073 on Build GRCh38
rs748254908 Chr.2: 46617348-46617348 on Build GRCh38
rs763865311 Chr.2: 46617264-46617264 on Build GRCh38
rs377229602 Chr.2: 46617184-46617184 on Build GRCh38
rs572519181 Chr.2: 46617370-46617370 on Build GRCh38
rs141311698 Chr.2: 46617096-46617096 on Build GRCh38
rs772642330 Chr.2: 46617000-46617000 on Build GRCh38
rs540887526 Chr.2: 46617182-46617182 on Build GRCh38
rs1033377258 Chr.2: 46617206-46617206 on Build GRCh38
rs1021924124 Chr.2: 46617064-46617064 on Build GRCh38
rs538058682 Chr.2: 46617212-46617212 on Build GRCh38
rs950638243 Chr.2: 46617103-46617103 on Build GRCh38
rs5830914 Chr.2: 46617080-46617081 on Build GRCh38
rs185728643 Chr.2: 46617048-46617048 on Build GRCh38
rs377630390 Chr.2: 46617248-46617248 on Build GRCh38

View Full Product Details