Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46624353-46624857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIPT
Entrez Gene ID
9419
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Gene Aliases
HSPC139, SSMDF
UniGene
Hs.133998
Gene Chromosome Location
Chr.2: 46617172-46630176 on Build GRCh38
Transcripts
NM_014171.5

SNP Detail

SNP ID Chromosome Location
rs925334290 Chr.2: 46624582-46624582 on Build GRCh38
rs767704653 Chr.2: 46624828-46624828 on Build GRCh38
rs897095256 Chr.2: 46624824-46624824 on Build GRCh38
rs911318002 Chr.2: 46624608-46624608 on Build GRCh38
rs910778507 Chr.2: 46624394-46624394 on Build GRCh38
rs897418855 Chr.2: 46624412-46624412 on Build GRCh38
rs986329309 Chr.2: 46624390-46624390 on Build GRCh38
rs375508912 Chr.2: 46624747-46624747 on Build GRCh38
rs772588221 Chr.2: 46624668-46624668 on Build GRCh38
rs958033539 Chr.2: 46624818-46624818 on Build GRCh38
rs748622797 Chr.2: 46624645-46624647 on Build GRCh38
rs879771844 Chr.2: 46624772-46624775 on Build GRCh38
rs951874736 Chr.2: 46624463-46624463 on Build GRCh38
rs769349078 Chr.2: 46624493-46624493 on Build GRCh38
rs1002751943 Chr.2: 46624571-46624571 on Build GRCh38
rs1006313605 Chr.2: 46624684-46624684 on Build GRCh38
rs543462673 Chr.2: 46624651-46624651 on Build GRCh38
rs897409226 Chr.2: 46624631-46624631 on Build GRCh38
rs555599326 Chr.2: 46624666-46624666 on Build GRCh38
rs1036892517 Chr.2: 46624816-46624816 on Build GRCh38
rs1015501471 Chr.2: 46624585-46624585 on Build GRCh38
rs978492767 Chr.2: 46624569-46624569 on Build GRCh38
rs966251718 Chr.2: 46624649-46624649 on Build GRCh38
rs13392119 Chr.2: 46624497-46624497 on Build GRCh38
rs938558489 Chr.2: 46624795-46624795 on Build GRCh38
rs887428518 Chr.2: 46624674-46624674 on Build GRCh38
rs8390 Chr.2: 46624728-46624728 on Build GRCh38
rs894900420 Chr.2: 46624693-46624693 on Build GRCh38
rs1034697056 Chr.2: 46624688-46624688 on Build GRCh38
rs576963767 Chr.2: 46624723-46624723 on Build GRCh38
rs553726248 Chr.2: 46624466-46624466 on Build GRCh38
rs1044225966 Chr.2: 46624506-46624506 on Build GRCh38
rs952241231 Chr.2: 46624616-46624616 on Build GRCh38
rs1033449216 Chr.2: 46624481-46624481 on Build GRCh38
rs1008367077 Chr.2: 46624389-46624389 on Build GRCh38
rs948550039 Chr.2: 46624503-46624503 on Build GRCh38
rs897825056 Chr.2: 46624587-46624587 on Build GRCh38
rs924880281 Chr.2: 46624667-46624667 on Build GRCh38
rs1013424825 Chr.2: 46624698-46624698 on Build GRCh38
rs936731324 Chr.2: 46624584-46624584 on Build GRCh38
rs538825013 Chr.2: 46624422-46624422 on Build GRCh38
rs924264816 Chr.2: 46624483-46624483 on Build GRCh38
rs533752673 Chr.2: 46624602-46624602 on Build GRCh38
rs1017789758 Chr.2: 46624633-46624633 on Build GRCh38
rs907036060 Chr.2: 46624788-46624788 on Build GRCh38
rs960926295 Chr.2: 46624692-46624692 on Build GRCh38
rs1041252293 Chr.2: 46624393-46624393 on Build GRCh38
rs975707770 Chr.2: 46624472-46624472 on Build GRCh38
rs576034461 Chr.2: 46624601-46624601 on Build GRCh38
rs536185049 Chr.2: 46624543-46624543 on Build GRCh38
rs1026018336 Chr.2: 46624448-46624448 on Build GRCh38
rs948474127 Chr.2: 46624735-46624735 on Build GRCh38
rs554517471 Chr.2: 46624590-46624590 on Build GRCh38
rs990236015 Chr.2: 46624682-46624682 on Build GRCh38
rs985433893 Chr.2: 46624823-46624823 on Build GRCh38
rs192854805 Chr.2: 46624783-46624783 on Build GRCh38
rs1033397073 Chr.2: 46624686-46624686 on Build GRCh38
rs986275724 Chr.2: 46624618-46624618 on Build GRCh38
rs993531327 Chr.2: 46624413-46624413 on Build GRCh38
rs530390498 Chr.2: 46624799-46624799 on Build GRCh38
rs774982146 Chr.2: 46624499-46624499 on Build GRCh38
rs1041198240 Chr.2: 46624619-46624619 on Build GRCh38
rs944176029 Chr.2: 46624609-46624609 on Build GRCh38
rs934331337 Chr.2: 46624675-46624675 on Build GRCh38
rs992002910 Chr.2: 46624484-46624484 on Build GRCh38
rs967013994 Chr.2: 46624737-46624737 on Build GRCh38

View Full Product Details