Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46624541-46625058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIPT
Entrez Gene ID
9419
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Gene Aliases
HSPC139, SSMDF
UniGene
Hs.133998
Gene Chromosome Location
Chr.2: 46617172-46630176 on Build GRCh38
Transcripts
NM_014171.5

SNP Detail

SNP ID Chromosome Location
rs925334290 Chr.2: 46624582-46624582 on Build GRCh38
rs993486188 Chr.2: 46624870-46624870 on Build GRCh38
rs767704653 Chr.2: 46624828-46624828 on Build GRCh38
rs139691368 Chr.2: 46624955-46624955 on Build GRCh38
rs897095256 Chr.2: 46624824-46624824 on Build GRCh38
rs911318002 Chr.2: 46624608-46624608 on Build GRCh38
rs112560095 Chr.2: 46624937-46624937 on Build GRCh38
rs3087822 Chr.2: 46624894-46624894 on Build GRCh38
rs149746896 Chr.2: 46625017-46625017 on Build GRCh38
rs375508912 Chr.2: 46624747-46624747 on Build GRCh38
rs772588221 Chr.2: 46624668-46624668 on Build GRCh38
rs958033539 Chr.2: 46624818-46624818 on Build GRCh38
rs748622797 Chr.2: 46624645-46624647 on Build GRCh38
rs879771844 Chr.2: 46624772-46624775 on Build GRCh38
rs1002751943 Chr.2: 46624571-46624571 on Build GRCh38
rs888704995 Chr.2: 46624954-46624954 on Build GRCh38
rs1006313605 Chr.2: 46624684-46624684 on Build GRCh38
rs543462673 Chr.2: 46624651-46624651 on Build GRCh38
rs897409226 Chr.2: 46624631-46624631 on Build GRCh38
rs555599326 Chr.2: 46624666-46624666 on Build GRCh38
rs1036892517 Chr.2: 46624816-46624816 on Build GRCh38
rs1015501471 Chr.2: 46624585-46624585 on Build GRCh38
rs978492767 Chr.2: 46624569-46624569 on Build GRCh38
rs966251718 Chr.2: 46624649-46624649 on Build GRCh38
rs938558489 Chr.2: 46624795-46624795 on Build GRCh38
rs531337119 Chr.2: 46624909-46624909 on Build GRCh38
rs887428518 Chr.2: 46624674-46624674 on Build GRCh38
rs894900420 Chr.2: 46624693-46624693 on Build GRCh38
rs1034697056 Chr.2: 46624688-46624688 on Build GRCh38
rs8390 Chr.2: 46624728-46624728 on Build GRCh38
rs1028088590 Chr.2: 46624872-46624872 on Build GRCh38
rs576963767 Chr.2: 46624723-46624723 on Build GRCh38
rs952241231 Chr.2: 46624616-46624616 on Build GRCh38
rs897825056 Chr.2: 46624587-46624587 on Build GRCh38
rs924880281 Chr.2: 46624667-46624667 on Build GRCh38
rs1013424825 Chr.2: 46624698-46624698 on Build GRCh38
rs936731324 Chr.2: 46624584-46624584 on Build GRCh38
rs1048612295 Chr.2: 46624944-46624944 on Build GRCh38
rs533752673 Chr.2: 46624602-46624602 on Build GRCh38
rs1017789758 Chr.2: 46624633-46624633 on Build GRCh38
rs907036060 Chr.2: 46624788-46624788 on Build GRCh38
rs960926295 Chr.2: 46624692-46624692 on Build GRCh38
rs576034461 Chr.2: 46624601-46624601 on Build GRCh38
rs78613586 Chr.2: 46625013-46625013 on Build GRCh38
rs948474127 Chr.2: 46624735-46624735 on Build GRCh38
rs554517471 Chr.2: 46624590-46624590 on Build GRCh38
rs542174111 Chr.2: 46624855-46624855 on Build GRCh38
rs990236015 Chr.2: 46624682-46624682 on Build GRCh38
rs985433893 Chr.2: 46624823-46624823 on Build GRCh38
rs192854805 Chr.2: 46624783-46624783 on Build GRCh38
rs985014242 Chr.2: 46625023-46625023 on Build GRCh38
rs986275724 Chr.2: 46624618-46624618 on Build GRCh38
rs1033397073 Chr.2: 46624686-46624686 on Build GRCh38
rs530390498 Chr.2: 46624799-46624799 on Build GRCh38
rs1041198240 Chr.2: 46624619-46624619 on Build GRCh38
rs944176029 Chr.2: 46624609-46624609 on Build GRCh38
rs558717604 Chr.2: 46624982-46624982 on Build GRCh38
rs183906288 Chr.2: 46624903-46624903 on Build GRCh38
rs934331337 Chr.2: 46624675-46624675 on Build GRCh38
rs59798877 Chr.2: 46624999-46625000 on Build GRCh38
rs188293300 Chr.2: 46624927-46624927 on Build GRCh38
rs10200194 Chr.2: 46624950-46624950 on Build GRCh38
rs967013994 Chr.2: 46624737-46624737 on Build GRCh38
rs894890606 Chr.2: 46625019-46625019 on Build GRCh38
rs547550761 Chr.2: 46624976-46624976 on Build GRCh38

View Full Product Details