Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 109543653-109544153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_001321513.1 NM_001321514.1 NM_001321515.1 NM_144710.4 NM_178584.3 XM_011510698.2 XM_011510699.2 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510703.2 XM_011510704.2

Gene Symbol
SEPT10
Entrez Gene ID
151011
Gene Name
septin 10
Gene Aliases
-
UniGene
Hs.469615
Gene Chromosome Location
Chr.2: 109542797-109614247 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_001321513.1 NM_001321514.1 NM_001321515.1 NM_144710.4 NM_178584.3 XM_011510698.2 XM_011510699.2 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510703.2 XM_011510704.2

SNP Detail

SNP ID Chromosome Location
rs148818283 Chr.2: 109543934-109543934 on Build GRCh38
rs771024780 Chr.2: 109544095-109544095 on Build GRCh38
rs952262107 Chr.2: 109543952-109543952 on Build GRCh38
rs892563439 Chr.2: 109543693-109543693 on Build GRCh38
rs779559888 Chr.2: 109543734-109543734 on Build GRCh38
rs145530743 Chr.2: 109543895-109543895 on Build GRCh38
rs3886070 Chr.2: 109543830-109543830 on Build GRCh38
rs536438498 Chr.2: 109543875-109543875 on Build GRCh38
rs1009648375 Chr.2: 109543709-109543709 on Build GRCh38
rs1043804903 Chr.2: 109544113-109544113 on Build GRCh38
rs904338519 Chr.2: 109543795-109543795 on Build GRCh38
rs913902687 Chr.2: 109544019-109544019 on Build GRCh38
rs1034288545 Chr.2: 109543843-109543843 on Build GRCh38
rs769557142 Chr.2: 109544124-109544124 on Build GRCh38
rs557346450 Chr.2: 109543930-109543930 on Build GRCh38
rs867355696 Chr.2: 109543833-109543833 on Build GRCh38
rs577132216 Chr.2: 109543967-109543967 on Build GRCh38
rs776477675 Chr.2: 109544096-109544096 on Build GRCh38
rs571735691 Chr.2: 109543807-109543807 on Build GRCh38
rs146923692 Chr.2: 109544015-109544015 on Build GRCh38
rs745817496 Chr.2: 109544117-109544117 on Build GRCh38
rs1023917026 Chr.2: 109543886-109543886 on Build GRCh38
rs748651458 Chr.2: 109544028-109544028 on Build GRCh38
rs1040361722 Chr.2: 109543689-109543689 on Build GRCh38
rs775265178 Chr.2: 109544127-109544127 on Build GRCh38
rs780767320 Chr.2: 109543777-109543777 on Build GRCh38
rs986683133 Chr.2: 109543962-109543962 on Build GRCh38
rs556486925 Chr.2: 109543876-109543876 on Build GRCh38
rs945467461 Chr.2: 109544057-109544057 on Build GRCh38
rs576308594 Chr.2: 109543882-109543882 on Build GRCh38
rs377505192 Chr.2: 109543732-109543732 on Build GRCh38
rs536593756 Chr.2: 109544055-109544055 on Build GRCh38
rs920787687 Chr.2: 109543950-109543950 on Build GRCh38

View Full Product Details