Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 109574136-109574670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_001321513.1 NM_001321514.1 NM_001321515.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510698.2 XM_011510699.2 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510703.2 XM_011510704.2

Gene Symbol
SEPT10
Entrez Gene ID
151011
Gene Name
septin 10
Gene Aliases
-
UniGene
Hs.469615
Gene Chromosome Location
Chr.2: 109542797-109614247 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_001321513.1 NM_001321514.1 NM_001321515.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510698.2 XM_011510699.2 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510703.2 XM_011510704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1153473 c.548G>A p.G183D Chr.2: 109574633-109574633 on Build GRCh38
6339305 c.456T>C p.C152C Chr.2: 109574656-109574656 on Build GRCh38
6775183 c.522G>A p.V174V Chr.2: 109574659-109574659 on Build GRCh38
1005332 c.479G>A p.G160D Chr.2: 109574633-109574633 on Build GRCh38
6338766 c.518A>G p.H173R Chr.2: 109574663-109574663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368933601 Chr.2: 109574642-109574642 on Build GRCh38
rs4953784 Chr.2: 109574198-109574198 on Build GRCh38
rs1053372150 Chr.2: 109574467-109574467 on Build GRCh38
rs4953729 Chr.2: 109574310-109574310 on Build GRCh38
rs888517669 Chr.2: 109574602-109574602 on Build GRCh38
rs1044139300 Chr.2: 109574466-109574466 on Build GRCh38
rs556752518 Chr.2: 109574204-109574204 on Build GRCh38
rs904066485 Chr.2: 109574313-109574313 on Build GRCh38
rs917734216 Chr.2: 109574391-109574391 on Build GRCh38
rs1016024941 Chr.2: 109574506-109574506 on Build GRCh38
rs901306236 Chr.2: 109574549-109574549 on Build GRCh38
rs757137990 Chr.2: 109574426-109574427 on Build GRCh38
rs540248161 Chr.2: 109574612-109574612 on Build GRCh38
rs191123260 Chr.2: 109574638-109574638 on Build GRCh38
rs529277914 Chr.2: 109574465-109574466 on Build GRCh38
rs1041334258 Chr.2: 109574543-109574543 on Build GRCh38
rs576142127 Chr.2: 109574396-109574396 on Build GRCh38
rs907054480 Chr.2: 109574487-109574487 on Build GRCh38
rs541139072 Chr.2: 109574444-109574446 on Build GRCh38
rs546875931 Chr.2: 109574196-109574196 on Build GRCh38
rs747433058 Chr.2: 109574446-109574447 on Build GRCh38
rs961936043 Chr.2: 109574373-109574373 on Build GRCh38
rs892156862 Chr.2: 109574480-109574480 on Build GRCh38
rs4953730 Chr.2: 109574553-109574553 on Build GRCh38
rs746412959 Chr.2: 109574554-109574555 on Build GRCh38
rs751344445 Chr.2: 109574555-109574555 on Build GRCh38
rs373295966 Chr.2: 109574444-109574445 on Build GRCh38
rs747221233 Chr.2: 109574625-109574626 on Build GRCh38
rs774321778 Chr.2: 109574627-109574627 on Build GRCh38
rs984132222 Chr.2: 109574361-109574361 on Build GRCh38
rs767423727 Chr.2: 109574443-109574444 on Build GRCh38
rs765123886 Chr.2: 109574535-109574535 on Build GRCh38
rs185846561 Chr.2: 109574646-109574646 on Build GRCh38
rs778123198 Chr.2: 109574601-109574601 on Build GRCh38
rs972043540 Chr.2: 109574388-109574388 on Build GRCh38
rs572054765 Chr.2: 109574492-109574492 on Build GRCh38
rs528627152 Chr.2: 109574173-109574173 on Build GRCh38
rs762985012 Chr.2: 109574617-109574617 on Build GRCh38
rs1009851647 Chr.2: 109574212-109574212 on Build GRCh38
rs377574367 Chr.2: 109574639-109574639 on Build GRCh38
rs945148316 Chr.2: 109574276-109574276 on Build GRCh38
rs55980206 Chr.2: 109574464-109574466 on Build GRCh38
rs926392564 Chr.2: 109574448-109574448 on Build GRCh38
rs144722410 Chr.2: 109574265-109574265 on Build GRCh38
rs944607458 Chr.2: 109574206-109574206 on Build GRCh38
rs775354135 Chr.2: 109574616-109574616 on Build GRCh38
rs758760040 Chr.2: 109574603-109574609 on Build GRCh38
rs936313005 Chr.2: 109574461-109574461 on Build GRCh38
rs28372574 Chr.2: 109574565-109574565 on Build GRCh38
rs201043038 Chr.2: 109574546-109574546 on Build GRCh38
rs776879717 Chr.2: 109574613-109574613 on Build GRCh38
rs780349316 Chr.2: 109574624-109574625 on Build GRCh38
rs1028421120 Chr.2: 109574284-109574284 on Build GRCh38
rs772502792 Chr.2: 109574596-109574596 on Build GRCh38
rs756994652 Chr.2: 109574557-109574557 on Build GRCh38
rs997333804 Chr.2: 109574244-109574244 on Build GRCh38
rs535072095 Chr.2: 109574179-109574179 on Build GRCh38
rs558499022 Chr.2: 109574474-109574474 on Build GRCh38
rs779346744 Chr.2: 109574584-109574584 on Build GRCh38
rs543344099 Chr.2: 109574574-109574574 on Build GRCh38
rs1020183892 Chr.2: 109574220-109574220 on Build GRCh38
rs544694926 Chr.2: 109574404-109574404 on Build GRCh38
rs965615410 Chr.2: 109574222-109574222 on Build GRCh38
rs749945195 Chr.2: 109574582-109574582 on Build GRCh38
rs995614382 Chr.2: 109574560-109574560 on Build GRCh38
rs375381556 Chr.2: 109574372-109574372 on Build GRCh38
rs531075178 Chr.2: 109574466-109574466 on Build GRCh38
rs540391413 Chr.2: 109574444-109574444 on Build GRCh38
rs980890273 Chr.2: 109574416-109574416 on Build GRCh38
rs74700664 Chr.2: 109574275-109574275 on Build GRCh38
rs986085962 Chr.2: 109574178-109574178 on Build GRCh38
rs369971346 Chr.2: 109574595-109574595 on Build GRCh38
rs985971019 Chr.2: 109574608-109574608 on Build GRCh38
rs775944684 Chr.2: 109574445-109574446 on Build GRCh38
rs751942653 Chr.2: 109574508-109574508 on Build GRCh38
rs761673341 Chr.2: 109574633-109574633 on Build GRCh38
rs1013815351 Chr.2: 109574481-109574481 on Build GRCh38
rs1055184563 Chr.2: 109574374-109574374 on Build GRCh38
rs3829701 Chr.2: 109574615-109574615 on Build GRCh38
rs1053940185 Chr.2: 109574166-109574166 on Build GRCh38

View Full Product Details