Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17084532-17085041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17RA
Entrez Gene ID
23765
Gene Name
interleukin 17 receptor A
Gene Aliases
CANDF5, CD217, CDw217, IL-17RA, IL17R, hIL-17R
UniGene
Hs.48353
Gene Chromosome Location
Chr.22: 17084959-17115694 on Build GRCh38
Transcripts
NM_001289905.1 NM_014339.6

SNP Detail

SNP ID Chromosome Location
rs942969410 Chr.22: 17084934-17084936 on Build GRCh38
rs539657742 Chr.22: 17084685-17084685 on Build GRCh38
rs559764929 Chr.22: 17085005-17085005 on Build GRCh38
rs776374859 Chr.22: 17084809-17084830 on Build GRCh38
rs767639812 Chr.22: 17084615-17084618 on Build GRCh38
rs930198397 Chr.22: 17084753-17084753 on Build GRCh38
rs569831094 Chr.22: 17084838-17084838 on Build GRCh38
rs980116362 Chr.22: 17084697-17084697 on Build GRCh38
rs376996486 Chr.22: 17084660-17084660 on Build GRCh38
rs1039883779 Chr.22: 17084936-17084936 on Build GRCh38
rs922791146 Chr.22: 17084955-17084955 on Build GRCh38
rs185181993 Chr.22: 17084659-17084659 on Build GRCh38
rs1022213204 Chr.22: 17084846-17084846 on Build GRCh38
rs960147437 Chr.22: 17084712-17084712 on Build GRCh38
rs969749029 Chr.22: 17084852-17084852 on Build GRCh38
rs546067797 Chr.22: 17084928-17084928 on Build GRCh38
rs951421797 Chr.22: 17084920-17084920 on Build GRCh38
rs892085069 Chr.22: 17084971-17084971 on Build GRCh38
rs1002099232 Chr.22: 17085008-17085008 on Build GRCh38
rs554059605 Chr.22: 17084893-17084893 on Build GRCh38
rs761649275 Chr.22: 17084964-17084964 on Build GRCh38
rs992774091 Chr.22: 17084716-17084716 on Build GRCh38
rs1048654657 Chr.22: 17084761-17084761 on Build GRCh38
rs1030709032 Chr.22: 17084622-17084622 on Build GRCh38
rs956813994 Chr.22: 17084623-17084623 on Build GRCh38
rs759661735 Chr.22: 17084759-17084759 on Build GRCh38
rs1018044838 Chr.22: 17084556-17084556 on Build GRCh38
rs910134712 Chr.22: 17084932-17084932 on Build GRCh38
rs868044162 Chr.22: 17084833-17084833 on Build GRCh38
rs918620407 Chr.22: 17084747-17084747 on Build GRCh38
rs774572394 Chr.22: 17084739-17084739 on Build GRCh38
rs564384700 Chr.22: 17084956-17084956 on Build GRCh38
rs942850613 Chr.22: 17084781-17084781 on Build GRCh38
rs562165706 Chr.22: 17084966-17084966 on Build GRCh38
rs551634574 Chr.22: 17084818-17084818 on Build GRCh38
rs918555491 Chr.22: 17084901-17084901 on Build GRCh38
rs997966403 Chr.22: 17084822-17084822 on Build GRCh38
rs1010840292 Chr.22: 17084630-17084630 on Build GRCh38
rs900916677 Chr.22: 17084819-17084819 on Build GRCh38
rs1052075915 Chr.22: 17084823-17084823 on Build GRCh38
rs529290465 Chr.22: 17084997-17084997 on Build GRCh38
rs910007153 Chr.22: 17084768-17084768 on Build GRCh38
rs1022516602 Chr.22: 17084679-17084679 on Build GRCh38
rs969255028 Chr.22: 17084693-17084693 on Build GRCh38
rs58106132 Chr.22: 17084665-17084665 on Build GRCh38
rs892142683 Chr.22: 17084835-17084835 on Build GRCh38
rs13053694 Chr.22: 17084998-17084998 on Build GRCh38
rs576568869 Chr.22: 17084958-17084958 on Build GRCh38
rs116718351 Chr.22: 17084886-17084886 on Build GRCh38
rs1034793747 Chr.22: 17085013-17085013 on Build GRCh38
rs117794241 Chr.22: 17084595-17084595 on Build GRCh38
rs572440976 Chr.22: 17084917-17084917 on Build GRCh38
rs546636380 Chr.22: 17084943-17084944 on Build GRCh38
rs543561697 Chr.22: 17084959-17084959 on Build GRCh38
rs900802101 Chr.22: 17084577-17084577 on Build GRCh38
rs140831326 Chr.22: 17084840-17084840 on Build GRCh38
rs190077844 Chr.22: 17084890-17084890 on Build GRCh38
rs573561759 Chr.22: 17084887-17084887 on Build GRCh38
rs865935570 Chr.22: 17084590-17084590 on Build GRCh38
rs992892022 Chr.22: 17084896-17084896 on Build GRCh38
rs12330050 Chr.22: 17084858-17084858 on Build GRCh38
rs772400784 Chr.22: 17084802-17084802 on Build GRCh38

View Full Product Details