Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17735148-17735642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BID
Entrez Gene ID
637
Gene Name
BH3 interacting domain death agonist
Gene Aliases
FP497
UniGene
Hs.517145
Gene Chromosome Location
Chr.22: 17734140-17774665 on Build GRCh38
Transcripts
NM_001196.3 NM_001244567.1 NM_001244569.1 NM_001244570.1 NM_001244572.1 NM_197966.2 NM_197967.2

SNP Detail

SNP ID Chromosome Location
rs1003613466 Chr.22: 17735259-17735259 on Build GRCh38
rs8190354 Chr.22: 17735545-17735545 on Build GRCh38
rs1047526411 Chr.22: 17735249-17735249 on Build GRCh38
rs928760794 Chr.22: 17735460-17735460 on Build GRCh38
rs1061200 Chr.22: 17735414-17735414 on Build GRCh38
rs776421618 Chr.22: 17735201-17735202 on Build GRCh38
rs779513207 Chr.22: 17735521-17735521 on Build GRCh38
rs900383501 Chr.22: 17735306-17735306 on Build GRCh38
rs758207864 Chr.22: 17735544-17735544 on Build GRCh38
rs760766944 Chr.22: 17735601-17735601 on Build GRCh38
rs148682143 Chr.22: 17735578-17735578 on Build GRCh38
rs766770581 Chr.22: 17735534-17735534 on Build GRCh38
rs774410572 Chr.22: 17735554-17735575 on Build GRCh38
rs886218724 Chr.22: 17735260-17735260 on Build GRCh38
rs923941123 Chr.22: 17735334-17735334 on Build GRCh38
rs760965422 Chr.22: 17735529-17735529 on Build GRCh38
rs1044551485 Chr.22: 17735219-17735219 on Build GRCh38
rs768970992 Chr.22: 17735589-17735589 on Build GRCh38
rs757193291 Chr.22: 17735549-17735549 on Build GRCh38
rs201491655 Chr.22: 17735202-17735203 on Build GRCh38
rs1027124264 Chr.22: 17735274-17735274 on Build GRCh38
rs544426630 Chr.22: 17735413-17735413 on Build GRCh38
rs186394629 Chr.22: 17735458-17735458 on Build GRCh38
rs748664557 Chr.22: 17735592-17735592 on Build GRCh38
rs566629518 Chr.22: 17735507-17735507 on Build GRCh38
rs970297485 Chr.22: 17735449-17735449 on Build GRCh38
rs774617687 Chr.22: 17735591-17735591 on Build GRCh38
rs146895750 Chr.22: 17735202-17735202 on Build GRCh38
rs954555499 Chr.22: 17735337-17735337 on Build GRCh38
rs766576105 Chr.22: 17735607-17735607 on Build GRCh38
rs369074192 Chr.22: 17735506-17735506 on Build GRCh38
rs181869684 Chr.22: 17735305-17735305 on Build GRCh38
rs368957466 Chr.22: 17735176-17735176 on Build GRCh38
rs1036458651 Chr.22: 17735513-17735513 on Build GRCh38
rs372534120 Chr.22: 17735526-17735526 on Build GRCh38
rs373153872 Chr.22: 17735376-17735376 on Build GRCh38
rs938388909 Chr.22: 17735246-17735246 on Build GRCh38
rs113283699 Chr.22: 17735318-17735318 on Build GRCh38
rs1031696399 Chr.22: 17735367-17735367 on Build GRCh38
rs755229729 Chr.22: 17735542-17735542 on Build GRCh38
rs745440984 Chr.22: 17735577-17735577 on Build GRCh38
rs977921585 Chr.22: 17735364-17735364 on Build GRCh38
rs779623768 Chr.22: 17735581-17735581 on Build GRCh38
rs540967803 Chr.22: 17735261-17735261 on Build GRCh38
rs772513255 Chr.22: 17735594-17735594 on Build GRCh38
rs907374422 Chr.22: 17735226-17735226 on Build GRCh38
rs1012201043 Chr.22: 17735219-17735220 on Build GRCh38
rs749940968 Chr.22: 17735604-17735606 on Build GRCh38
rs935414596 Chr.22: 17735344-17735344 on Build GRCh38
rs1053636037 Chr.22: 17735203-17735203 on Build GRCh38
rs544643787 Chr.22: 17735352-17735352 on Build GRCh38
rs952862097 Chr.22: 17735301-17735301 on Build GRCh38
rs368291803 Chr.22: 17735541-17735541 on Build GRCh38
rs996497573 Chr.22: 17735310-17735310 on Build GRCh38
rs199693822 Chr.22: 17735599-17735599 on Build GRCh38
rs566764146 Chr.22: 17735441-17735442 on Build GRCh38
rs769273022 Chr.22: 17735508-17735508 on Build GRCh38
rs575412458 Chr.22: 17735211-17735212 on Build GRCh38
rs375225896 Chr.22: 17735504-17735504 on Build GRCh38
rs2305001 Chr.22: 17735444-17735444 on Build GRCh38
rs758789061 Chr.22: 17735312-17735312 on Build GRCh38
rs537020099 Chr.22: 17735265-17735265 on Build GRCh38
rs59225839 Chr.22: 17735587-17735587 on Build GRCh38
rs1802194 Chr.22: 17735429-17735429 on Build GRCh38
rs973104817 Chr.22: 17735270-17735271 on Build GRCh38
rs565001172 Chr.22: 17735398-17735398 on Build GRCh38
rs780744301 Chr.22: 17735564-17735564 on Build GRCh38
rs990161187 Chr.22: 17735440-17735440 on Build GRCh38

View Full Product Details