Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19820486-19820992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1L
Entrez Gene ID
54584
Gene Name
G protein subunit beta 1 like
Gene Aliases
DGCRK3, FKSG1, GY2, WDR14, WDVCF
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19788411-19854939 on Build GRCh38
Transcripts
NM_053004.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931312 c.236A>T p.Q79L Chr.22: 19820616-19820616 on Build GRCh38
4687433 c.200G>A p.G67D Chr.22: 19820652-19820652 on Build GRCh38
4687435 c.177G>A p.A59A Chr.22: 19820675-19820675 on Build GRCh38
1130505 c.236A>C p.Q79P Chr.22: 19820616-19820616 on Build GRCh38
3964096 c.190G>C p.D64H Chr.22: 19820662-19820662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779414365 Chr.22: 19820561-19820561 on Build GRCh38
rs765354937 Chr.22: 19820560-19820560 on Build GRCh38
rs761056471 Chr.22: 19820552-19820552 on Build GRCh38
rs544373606 Chr.22: 19820775-19820775 on Build GRCh38
rs150444079 Chr.22: 19820682-19820682 on Build GRCh38
rs887438563 Chr.22: 19820914-19820914 on Build GRCh38
rs766709794 Chr.22: 19820556-19820556 on Build GRCh38
rs765606321 Chr.22: 19820679-19820679 on Build GRCh38
rs930243073 Chr.22: 19820844-19820844 on Build GRCh38
rs532014981 Chr.22: 19820912-19820912 on Build GRCh38
rs377544200 Chr.22: 19820734-19820734 on Build GRCh38
rs1028775831 Chr.22: 19820967-19820967 on Build GRCh38
rs780057321 Chr.22: 19820700-19820700 on Build GRCh38
rs551847730 Chr.22: 19820969-19820969 on Build GRCh38
rs202097453 Chr.22: 19820611-19820611 on Build GRCh38
rs749304429 Chr.22: 19820584-19820584 on Build GRCh38
rs997168248 Chr.22: 19820956-19820956 on Build GRCh38
rs746545339 Chr.22: 19820750-19820750 on Build GRCh38
rs140221967 Chr.22: 19820614-19820614 on Build GRCh38
rs747204856 Chr.22: 19820573-19820573 on Build GRCh38
rs758450071 Chr.22: 19820630-19820630 on Build GRCh38
rs750615518 Chr.22: 19820707-19820707 on Build GRCh38
rs765685532 Chr.22: 19820625-19820625 on Build GRCh38
rs777629866 Chr.22: 19820747-19820747 on Build GRCh38
rs769759118 Chr.22: 19820576-19820576 on Build GRCh38
rs554963616 Chr.22: 19820537-19820537 on Build GRCh38
rs138999281 Chr.22: 19820651-19820651 on Build GRCh38
rs575033326 Chr.22: 19820615-19820615 on Build GRCh38
rs777976433 Chr.22: 19820641-19820641 on Build GRCh38
rs757547791 Chr.22: 19820645-19820645 on Build GRCh38
rs777888950 Chr.22: 19820571-19820571 on Build GRCh38
rs188043218 Chr.22: 19820802-19820802 on Build GRCh38
rs1045093335 Chr.22: 19820540-19820540 on Build GRCh38
rs1038779090 Chr.22: 19820937-19820937 on Build GRCh38
rs538092991 Chr.22: 19820527-19820527 on Build GRCh38
rs34770066 Chr.22: 19820648-19820649 on Build GRCh38
rs746959940 Chr.22: 19820704-19820704 on Build GRCh38
rs35357220 Chr.22: 19820679-19820680 on Build GRCh38
rs745514816 Chr.22: 19820773-19820773 on Build GRCh38
rs905266910 Chr.22: 19820746-19820746 on Build GRCh38
rs771543366 Chr.22: 19820661-19820661 on Build GRCh38
rs918869732 Chr.22: 19820814-19820814 on Build GRCh38
rs770588430 Chr.22: 19820673-19820673 on Build GRCh38
rs868112038 Chr.22: 19820913-19820913 on Build GRCh38
rs949357486 Chr.22: 19820702-19820702 on Build GRCh38
rs138225912 Chr.22: 19820683-19820683 on Build GRCh38
rs781497461 Chr.22: 19820649-19820649 on Build GRCh38
rs916758067 Chr.22: 19820778-19820778 on Build GRCh38
rs778745424 Chr.22: 19820713-19820713 on Build GRCh38
rs1019956982 Chr.22: 19820623-19820623 on Build GRCh38
rs111628424 Chr.22: 19820706-19820706 on Build GRCh38
rs201313303 Chr.22: 19820608-19820608 on Build GRCh38
rs746453433 Chr.22: 19820670-19820670 on Build GRCh38
rs772707311 Chr.22: 19820668-19820668 on Build GRCh38
rs765939928 Chr.22: 19820752-19820752 on Build GRCh38
rs560072011 Chr.22: 19820812-19820812 on Build GRCh38
rs776308828 Chr.22: 19820757-19820757 on Build GRCh38
rs949517821 Chr.22: 19820781-19820781 on Build GRCh38
rs752732745 Chr.22: 19820564-19820564 on Build GRCh38
rs901529824 Chr.22: 19820929-19820929 on Build GRCh38
rs757354984 Chr.22: 19820694-19820694 on Build GRCh38
rs560935682 Chr.22: 19820777-19820777 on Build GRCh38
rs776216882 Chr.22: 19820675-19820675 on Build GRCh38
rs764335737 Chr.22: 19820684-19820684 on Build GRCh38
rs771568536 Chr.22: 19820606-19820606 on Build GRCh38
rs768481898 Chr.22: 19820587-19820587 on Build GRCh38
rs5748447 Chr.22: 19820517-19820517 on Build GRCh38
rs958137088 Chr.22: 19820719-19820719 on Build GRCh38
rs149599532 Chr.22: 19820685-19820685 on Build GRCh38
rs753942180 Chr.22: 19820753-19820753 on Build GRCh38
rs761247710 Chr.22: 19820598-19820598 on Build GRCh38
rs202068306 Chr.22: 19820597-19820597 on Build GRCh38
rs141534596 Chr.22: 19820676-19820676 on Build GRCh38
rs941773209 Chr.22: 19820921-19820921 on Build GRCh38
rs755584139 Chr.22: 19820555-19820556 on Build GRCh38
rs758338248 Chr.22: 19820739-19820739 on Build GRCh38
rs554106644 Chr.22: 19820653-19820653 on Build GRCh38
rs184782658 Chr.22: 19820654-19820654 on Build GRCh38
rs142592326 Chr.22: 19820644-19820644 on Build GRCh38
rs546600035 Chr.22: 19820807-19820807 on Build GRCh38
rs375080541 Chr.22: 19820569-19820569 on Build GRCh38
rs370557688 Chr.22: 19820557-19820557 on Build GRCh38
rs531891810 Chr.22: 19820969-19820970 on Build GRCh38
rs900339108 Chr.22: 19820960-19820960 on Build GRCh38
rs368294430 Chr.22: 19820624-19820624 on Build GRCh38
rs563248037 Chr.22: 19820705-19820705 on Build GRCh38
rs754767322 Chr.22: 19820711-19820711 on Build GRCh38

View Full Product Details