Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50176788-50177289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PANX2
Entrez Gene ID
56666
Gene Name
pannexin 2
Gene Aliases
PX2, hPANX2
UniGene
Hs.440092
Gene Chromosome Location
Chr.22: 50170731-50180295 on Build GRCh38
Transcripts
NM_001160300.1 NM_052839.3 NR_027691.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6737314 c.325G>A p.G109S Chr.22: 50177037-50177037 on Build GRCh38
1417013 c.379G>A p.A127T Chr.22: 50177091-50177091 on Build GRCh38
4952212 c.422G>T p.G141V Chr.22: 50177134-50177134 on Build GRCh38
6737326 c.267C>T p.R89R Chr.22: 50176979-50176979 on Build GRCh38
6737322 c.555G>A p.P185P Chr.22: 50177267-50177267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780499384 Chr.22: 50176917-50176917 on Build GRCh38
rs764530663 Chr.22: 50177186-50177186 on Build GRCh38
rs985929509 Chr.22: 50176943-50176943 on Build GRCh38
rs868711215 Chr.22: 50176862-50176862 on Build GRCh38
rs930213427 Chr.22: 50177109-50177109 on Build GRCh38
rs762320788 Chr.22: 50177024-50177024 on Build GRCh38
rs753938610 Chr.22: 50177188-50177188 on Build GRCh38
rs370425844 Chr.22: 50177270-50177270 on Build GRCh38
rs200374068 Chr.22: 50177157-50177157 on Build GRCh38
rs113728144 Chr.22: 50177100-50177100 on Build GRCh38
rs953531964 Chr.22: 50177097-50177097 on Build GRCh38
rs775516298 Chr.22: 50176964-50176964 on Build GRCh38
rs749293056 Chr.22: 50176961-50176961 on Build GRCh38
rs377363933 Chr.22: 50177263-50177263 on Build GRCh38
rs750122109 Chr.22: 50177022-50177022 on Build GRCh38
rs904893014 Chr.22: 50176851-50176851 on Build GRCh38
rs769899093 Chr.22: 50176946-50176946 on Build GRCh38
rs768497136 Chr.22: 50176987-50176987 on Build GRCh38
rs747857897 Chr.22: 50177123-50177123 on Build GRCh38
rs984788807 Chr.22: 50177122-50177122 on Build GRCh38
rs143921868 Chr.22: 50177171-50177171 on Build GRCh38
rs909188214 Chr.22: 50177258-50177258 on Build GRCh38
rs149462540 Chr.22: 50177113-50177113 on Build GRCh38
rs555858722 Chr.22: 50177153-50177153 on Build GRCh38
rs757547764 Chr.22: 50177195-50177195 on Build GRCh38
rs768075230 Chr.22: 50177042-50177042 on Build GRCh38
rs1053654979 Chr.22: 50176861-50176861 on Build GRCh38
rs895508131 Chr.22: 50176945-50176945 on Build GRCh38
rs779334671 Chr.22: 50177196-50177196 on Build GRCh38
rs751999520 Chr.22: 50177037-50177037 on Build GRCh38
rs780943862 Chr.22: 50177211-50177211 on Build GRCh38
rs267606285 Chr.22: 50177144-50177144 on Build GRCh38
rs377135639 Chr.22: 50177093-50177093 on Build GRCh38
rs369117934 Chr.22: 50177159-50177159 on Build GRCh38
rs758589956 Chr.22: 50177202-50177202 on Build GRCh38
rs760099816 Chr.22: 50177084-50177084 on Build GRCh38
rs753152454 Chr.22: 50177205-50177206 on Build GRCh38
rs1044820701 Chr.22: 50176850-50176850 on Build GRCh38
rs748131000 Chr.22: 50177223-50177223 on Build GRCh38
rs766091010 Chr.22: 50177150-50177150 on Build GRCh38
rs770551956 Chr.22: 50177265-50177265 on Build GRCh38
rs752986002 Chr.22: 50177046-50177046 on Build GRCh38
rs376958028 Chr.22: 50176900-50176900 on Build GRCh38
rs781545216 Chr.22: 50176931-50176931 on Build GRCh38
rs765646490 Chr.22: 50177172-50177172 on Build GRCh38
rs949164206 Chr.22: 50176844-50176844 on Build GRCh38
rs150278474 Chr.22: 50176988-50176988 on Build GRCh38
rs755167170 Chr.22: 50177158-50177158 on Build GRCh38
rs149618200 Chr.22: 50176860-50176860 on Build GRCh38
rs753273558 Chr.22: 50177081-50177082 on Build GRCh38
rs941735789 Chr.22: 50177152-50177152 on Build GRCh38
rs753274945 Chr.22: 50177160-50177160 on Build GRCh38
rs137878715 Chr.22: 50177076-50177076 on Build GRCh38
rs755473922 Chr.22: 50177040-50177040 on Build GRCh38
rs1009411525 Chr.22: 50176883-50176883 on Build GRCh38
rs771162262 Chr.22: 50176962-50176962 on Build GRCh38
rs998048101 Chr.22: 50176995-50176995 on Build GRCh38
rs757345103 Chr.22: 50177085-50177085 on Build GRCh38
rs765007738 Chr.22: 50176899-50176899 on Build GRCh38
rs1018342641 Chr.22: 50176985-50176985 on Build GRCh38
rs778109852 Chr.22: 50176948-50176948 on Build GRCh38
rs1002675982 Chr.22: 50176845-50176845 on Build GRCh38
rs761725284 Chr.22: 50177000-50177000 on Build GRCh38
rs765889070 Chr.22: 50177033-50177033 on Build GRCh38
rs746009911 Chr.22: 50177096-50177096 on Build GRCh38
rs565139391 Chr.22: 50176859-50176859 on Build GRCh38
rs769411522 Chr.22: 50177246-50177246 on Build GRCh38
rs755278990 Chr.22: 50176919-50176919 on Build GRCh38
rs748615506 Chr.22: 50176940-50176940 on Build GRCh38
rs762599245 Chr.22: 50177147-50177147 on Build GRCh38
rs777814098 Chr.22: 50177072-50177072 on Build GRCh38
rs756538935 Chr.22: 50177060-50177060 on Build GRCh38
rs760698271 Chr.22: 50176976-50176976 on Build GRCh38
rs767150673 Chr.22: 50177168-50177168 on Build GRCh38
rs577626429 Chr.22: 50177178-50177178 on Build GRCh38
rs769652627 Chr.22: 50177126-50177126 on Build GRCh38
rs765884570 Chr.22: 50177033-50177033 on Build GRCh38
rs757967849 Chr.22: 50176908-50176908 on Build GRCh38
rs776678689 Chr.22: 50177120-50177120 on Build GRCh38
rs764773897 Chr.22: 50177015-50177015 on Build GRCh38
rs566902203 Chr.22: 50176982-50176982 on Build GRCh38

View Full Product Details