Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3150239-3150747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2 XM_011533778.2

Gene Symbol
TRNT1
Entrez Gene ID
51095
Gene Name
tRNA nucleotidyl transferase 1
Gene Aliases
CCA1, CGI-47, MtCCA, RPEM, SIFD
UniGene
Hs.732725
Gene Chromosome Location
Chr.3: 3126916-3153435 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2 XM_011533778.2

SNP Detail

SNP ID Chromosome Location
rs923898945 Chr.3: 3150384-3150384 on Build GRCh38
rs138344480 Chr.3: 3150295-3150295 on Build GRCh38
rs772362692 Chr.3: 3150571-3150571 on Build GRCh38
rs1018465066 Chr.3: 3150664-3150664 on Build GRCh38
rs766040851 Chr.3: 3150717-3150718 on Build GRCh38
rs866700159 Chr.3: 3150576-3150576 on Build GRCh38
rs150489297 Chr.3: 3150284-3150284 on Build GRCh38
rs948018969 Chr.3: 3150319-3150319 on Build GRCh38
rs138718854 Chr.3: 3150481-3150481 on Build GRCh38
rs771322827 Chr.3: 3150494-3150494 on Build GRCh38
rs551497808 Chr.3: 3150562-3150562 on Build GRCh38
rs375499139 Chr.3: 3150320-3150321 on Build GRCh38
rs538385669 Chr.3: 3150509-3150509 on Build GRCh38
rs373628610 Chr.3: 3150665-3150665 on Build GRCh38
rs1011051866 Chr.3: 3150658-3150658 on Build GRCh38
rs907695406 Chr.3: 3150493-3150493 on Build GRCh38
rs1021379485 Chr.3: 3150614-3150614 on Build GRCh38
rs1021414400 Chr.3: 3150415-3150415 on Build GRCh38
rs750438512 Chr.3: 3150456-3150457 on Build GRCh38
rs567121143 Chr.3: 3150444-3150444 on Build GRCh38
rs76212525 Chr.3: 3150272-3150272 on Build GRCh38
rs550252032 Chr.3: 3150362-3150362 on Build GRCh38
rs577119206 Chr.3: 3150555-3150555 on Build GRCh38
rs760616455 Chr.3: 3150392-3150393 on Build GRCh38
rs961018989 Chr.3: 3150689-3150690 on Build GRCh38
rs3933095 Chr.3: 3150398-3150398 on Build GRCh38
rs1014573060 Chr.3: 3150306-3150306 on Build GRCh38
rs398091236 Chr.3: 3150532-3150532 on Build GRCh38
rs894020443 Chr.3: 3150394-3150394 on Build GRCh38
rs907747634 Chr.3: 3150275-3150275 on Build GRCh38
rs900215425 Chr.3: 3150666-3150666 on Build GRCh38
rs923153529 Chr.3: 3150270-3150270 on Build GRCh38
rs888012434 Chr.3: 3150297-3150297 on Build GRCh38
rs774741285 Chr.3: 3150558-3150558 on Build GRCh38
rs941895158 Chr.3: 3150503-3150503 on Build GRCh38
rs893968395 Chr.3: 3150640-3150640 on Build GRCh38
rs777148020 Chr.3: 3150438-3150438 on Build GRCh38
rs111409245 Chr.3: 3150352-3150352 on Build GRCh38
rs201102811 Chr.3: 3150519-3150520 on Build GRCh38
rs995972921 Chr.3: 3150682-3150682 on Build GRCh38
rs376695741 Chr.3: 3150317-3150317 on Build GRCh38
rs1000967373 Chr.3: 3150268-3150279 on Build GRCh38
rs143047659 Chr.3: 3150425-3150425 on Build GRCh38
rs948166954 Chr.3: 3150557-3150557 on Build GRCh38
rs916626788 Chr.3: 3150554-3150554 on Build GRCh38
rs549091759 Chr.3: 3150322-3150323 on Build GRCh38
rs935566819 Chr.3: 3150618-3150618 on Build GRCh38
rs983370791 Chr.3: 3150462-3150462 on Build GRCh38
rs1047518704 Chr.3: 3150273-3150273 on Build GRCh38
rs901545407 Chr.3: 3150520-3150520 on Build GRCh38
rs942117489 Chr.3: 3150277-3150277 on Build GRCh38
rs965832072 Chr.3: 3150312-3150312 on Build GRCh38
rs977346084 Chr.3: 3150668-3150668 on Build GRCh38
rs200243789 Chr.3: 3150277-3150278 on Build GRCh38
rs1004570850 Chr.3: 3150299-3150299 on Build GRCh38
rs558233875 Chr.3: 3150517-3150517 on Build GRCh38
rs747745731 Chr.3: 3150378-3150378 on Build GRCh38
rs751366924 Chr.3: 3150333-3150333 on Build GRCh38
rs953132055 Chr.3: 3150534-3150534 on Build GRCh38
rs1029189578 Chr.3: 3150532-3150533 on Build GRCh38
rs574983347 Chr.3: 3150451-3150451 on Build GRCh38
rs117137374 Chr.3: 3150459-3150459 on Build GRCh38
rs916672440 Chr.3: 3150692-3150693 on Build GRCh38
rs1036034293 Chr.3: 3150507-3150515 on Build GRCh38
rs11320492 Chr.3: 3150519-3150519 on Build GRCh38
rs1052713717 Chr.3: 3150633-3150633 on Build GRCh38
rs967269260 Chr.3: 3150645-3150645 on Build GRCh38
rs866991210 Chr.3: 3150345-3150345 on Build GRCh38
rs559409007 Chr.3: 3150570-3150570 on Build GRCh38
rs562453087 Chr.3: 3150323-3150323 on Build GRCh38
rs1030406673 Chr.3: 3150685-3150685 on Build GRCh38
rs534342778 Chr.3: 3150315-3150316 on Build GRCh38
rs79231110 Chr.3: 3150712-3150712 on Build GRCh38
rs928559318 Chr.3: 3150674-3150674 on Build GRCh38
rs63413460 Chr.3: 3150532-3150532 on Build GRCh38
rs967238007 Chr.3: 3150355-3150355 on Build GRCh38
rs1031169281 Chr.3: 3150391-3150391 on Build GRCh38
rs954423886 Chr.3: 3150698-3150698 on Build GRCh38
rs568242010 Chr.3: 3150458-3150458 on Build GRCh38
rs539598986 Chr.3: 3150424-3150424 on Build GRCh38
rs989929414 Chr.3: 3150337-3150340 on Build GRCh38

View Full Product Details