Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 4652019-4652517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
UniGene
Hs.567295
Gene Chromosome Location
Chr.3: 4493348-4847840 on Build GRCh38
Transcripts
NM_001099952.2 NM_001168272.1 NM_002222.5 XM_005265109.2 XM_005265110.2 XM_006713131.2 XM_011533681.1 XM_011533682.2 XM_011533683.2 XM_011533684.1 XM_011533685.1 XM_011533686.1 XM_011533687.1 XM_011533688.1 XM_011533690.1 XM_011533691.1 XM_011533692.2 XM_017006357.1 XM_017006358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4442500 c.911G>A p.R304H Chr.3: 4652178-4652178 on Build GRCh38
1211127 c.877C>T p.R293W Chr.3: 4652144-4652144 on Build GRCh38
1211129 c.877C>T p.R293W Chr.3: 4652144-4652144 on Build GRCh38
3916155 c.910C>T p.R304C Chr.3: 4652177-4652177 on Build GRCh38
1211128 c.877C>T p.R293W Chr.3: 4652144-4652144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977302186 Chr.3: 4652375-4652375 on Build GRCh38
rs28562557 Chr.3: 4652293-4652293 on Build GRCh38
rs755408791 Chr.3: 4652086-4652086 on Build GRCh38
rs959308992 Chr.3: 4652378-4652378 on Build GRCh38
rs747960676 Chr.3: 4652261-4652261 on Build GRCh38
rs532555370 Chr.3: 4652334-4652334 on Build GRCh38
rs559076826 Chr.3: 4652472-4652472 on Build GRCh38
rs762948180 Chr.3: 4652183-4652183 on Build GRCh38
rs764508786 Chr.3: 4652245-4652245 on Build GRCh38
rs377049416 Chr.3: 4652144-4652144 on Build GRCh38
rs779720395 Chr.3: 4652266-4652266 on Build GRCh38
rs374940696 Chr.3: 4652169-4652169 on Build GRCh38
rs755456504 Chr.3: 4652066-4652066 on Build GRCh38
rs1026083894 Chr.3: 4652322-4652322 on Build GRCh38
rs767971666 Chr.3: 4652226-4652226 on Build GRCh38
rs201693219 Chr.3: 4652196-4652196 on Build GRCh38
rs924747472 Chr.3: 4652457-4652457 on Build GRCh38
rs768096391 Chr.3: 4652098-4652098 on Build GRCh38
rs1002751445 Chr.3: 4652090-4652090 on Build GRCh38
rs772285603 Chr.3: 4652267-4652267 on Build GRCh38
rs28592010 Chr.3: 4652269-4652269 on Build GRCh38
rs753222753 Chr.3: 4652116-4652116 on Build GRCh38
rs746403201 Chr.3: 4652242-4652243 on Build GRCh38
rs778918958 Chr.3: 4652079-4652079 on Build GRCh38
rs769678088 Chr.3: 4652177-4652177 on Build GRCh38
rs908773665 Chr.3: 4652344-4652344 on Build GRCh38
rs779644839 Chr.3: 4652252-4652252 on Build GRCh38
rs768473868 Chr.3: 4652158-4652158 on Build GRCh38
rs56896093 Chr.3: 4652327-4652327 on Build GRCh38
rs542044311 Chr.3: 4652296-4652296 on Build GRCh38
rs766101615 Chr.3: 4652439-4652439 on Build GRCh38
rs1042025075 Chr.3: 4652287-4652287 on Build GRCh38
rs74656548 Chr.3: 4652486-4652486 on Build GRCh38
rs6442896 Chr.3: 4652326-4652326 on Build GRCh38
rs28724276 Chr.3: 4652259-4652259 on Build GRCh38
rs768429769 Chr.3: 4652283-4652283 on Build GRCh38
rs753169557 Chr.3: 4652233-4652233 on Build GRCh38
rs758662687 Chr.3: 4652268-4652268 on Build GRCh38
rs931686675 Chr.3: 4652430-4652430 on Build GRCh38
rs944185117 Chr.3: 4652230-4652230 on Build GRCh38
rs187915431 Chr.3: 4652299-4652299 on Build GRCh38
rs534892139 Chr.3: 4652044-4652044 on Build GRCh38
rs949266375 Chr.3: 4652444-4652444 on Build GRCh38
rs28565126 Chr.3: 4652197-4652197 on Build GRCh38
rs914366404 Chr.3: 4652466-4652466 on Build GRCh38
rs761236661 Chr.3: 4652238-4652238 on Build GRCh38
rs144440393 Chr.3: 4652454-4652454 on Build GRCh38
rs773337674 Chr.3: 4652178-4652178 on Build GRCh38
rs779400434 Chr.3: 4652224-4652225 on Build GRCh38
rs749888837 Chr.3: 4652249-4652249 on Build GRCh38
rs754678962 Chr.3: 4652258-4652258 on Build GRCh38
rs746760261 Chr.3: 4652154-4652154 on Build GRCh38
rs535024064 Chr.3: 4652478-4652478 on Build GRCh38
rs965670389 Chr.3: 4652358-4652358 on Build GRCh38
rs372668740 Chr.3: 4652131-4652131 on Build GRCh38
rs139601218 Chr.3: 4652043-4652043 on Build GRCh38
rs6442895 Chr.3: 4652253-4652253 on Build GRCh38
rs914567664 Chr.3: 4652493-4652493 on Build GRCh38
rs917797864 Chr.3: 4652386-4652386 on Build GRCh38
rs1012960913 Chr.3: 4652307-4652307 on Build GRCh38
rs745432988 Chr.3: 4652145-4652145 on Build GRCh38
rs779584031 Chr.3: 4652150-4652150 on Build GRCh38
rs1017763016 Chr.3: 4652292-4652292 on Build GRCh38
rs370893047 Chr.3: 4652348-4652348 on Build GRCh38
rs777587240 Chr.3: 4652265-4652265 on Build GRCh38
rs773798944 Chr.3: 4652471-4652471 on Build GRCh38
rs574185072 Chr.3: 4652171-4652171 on Build GRCh38
rs6442897 Chr.3: 4652384-4652384 on Build GRCh38
rs28461825 Chr.3: 4652275-4652275 on Build GRCh38
rs772671230 Chr.3: 4652246-4652246 on Build GRCh38
rs908832824 Chr.3: 4652227-4652227 on Build GRCh38
rs370157764 Chr.3: 4652149-4652149 on Build GRCh38
rs747182538 Chr.3: 4652282-4652282 on Build GRCh38
rs374618053 Chr.3: 4652200-4652200 on Build GRCh38
rs776454785 Chr.3: 4652170-4652170 on Build GRCh38
rs937891703 Chr.3: 4652067-4652067 on Build GRCh38
rs192965932 Chr.3: 4652479-4652479 on Build GRCh38

View Full Product Details