Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 5207938-5208445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDEM1
Entrez Gene ID
9695
Gene Name
ER degradation enhancing alpha-mannosidase like protein 1
Gene Aliases
EDEM
UniGene
Hs.224616
Gene Chromosome Location
Chr.3: 5187674-5219965 on Build GRCh38
Transcripts
NM_014674.2 XM_011534271.2 XM_011534272.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6033398 c.1444G>A p.A482T Chr.3: 5208198-5208198 on Build GRCh38
731103 c.1483G>T p.V495L Chr.3: 5208237-5208237 on Build GRCh38
3596079 c.1468C>T p.L490L Chr.3: 5208222-5208222 on Build GRCh38
6651570 c.1428T>C p.Y476Y Chr.3: 5208182-5208182 on Build GRCh38
5615300 c.1366A>G p.I456V Chr.3: 5208120-5208120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944894376 Chr.3: 5207999-5207999 on Build GRCh38
rs778150624 Chr.3: 5208277-5208278 on Build GRCh38
rs775419224 Chr.3: 5208290-5208290 on Build GRCh38
rs111425301 Chr.3: 5208300-5208300 on Build GRCh38
rs766152499 Chr.3: 5208045-5208045 on Build GRCh38
rs772770420 Chr.3: 5208157-5208157 on Build GRCh38
rs756108531 Chr.3: 5208069-5208069 on Build GRCh38
rs182996028 Chr.3: 5208197-5208197 on Build GRCh38
rs561737277 Chr.3: 5208395-5208395 on Build GRCh38
rs940687596 Chr.3: 5208392-5208392 on Build GRCh38
rs373755060 Chr.3: 5208278-5208278 on Build GRCh38
rs773838240 Chr.3: 5208280-5208280 on Build GRCh38
rs922074439 Chr.3: 5208278-5208278 on Build GRCh38
rs778578550 Chr.3: 5208278-5208280 on Build GRCh38
rs201299932 Chr.3: 5208156-5208156 on Build GRCh38
rs751521483 Chr.3: 5208010-5208010 on Build GRCh38
rs747459729 Chr.3: 5208248-5208248 on Build GRCh38
rs1018706268 Chr.3: 5208273-5208273 on Build GRCh38
rs749843599 Chr.3: 5208278-5208279 on Build GRCh38
rs983111387 Chr.3: 5208262-5208262 on Build GRCh38
rs754097657 Chr.3: 5208080-5208080 on Build GRCh38
rs374360841 Chr.3: 5208270-5208270 on Build GRCh38
rs759203902 Chr.3: 5208194-5208194 on Build GRCh38
rs757619200 Chr.3: 5208083-5208083 on Build GRCh38
rs765365530 Chr.3: 5208304-5208304 on Build GRCh38
rs149973887 Chr.3: 5208060-5208060 on Build GRCh38
rs376401602 Chr.3: 5208064-5208064 on Build GRCh38
rs928735277 Chr.3: 5208413-5208413 on Build GRCh38
rs750551379 Chr.3: 5208309-5208309 on Build GRCh38
rs761944774 Chr.3: 5208301-5208301 on Build GRCh38
rs950466878 Chr.3: 5208260-5208260 on Build GRCh38
rs1003338133 Chr.3: 5208166-5208166 on Build GRCh38
rs770453897 Chr.3: 5208279-5208279 on Build GRCh38
rs965754102 Chr.3: 5208276-5208276 on Build GRCh38
rs767266941 Chr.3: 5208182-5208182 on Build GRCh38
rs113256642 Chr.3: 5208275-5208276 on Build GRCh38
rs747552775 Chr.3: 5208147-5208147 on Build GRCh38
rs139257563 Chr.3: 5208000-5208000 on Build GRCh38
rs899031805 Chr.3: 5208021-5208021 on Build GRCh38
rs371280594 Chr.3: 5208160-5208160 on Build GRCh38
rs765507493 Chr.3: 5208222-5208222 on Build GRCh38
rs1051717487 Chr.3: 5208370-5208370 on Build GRCh38
rs757527194 Chr.3: 5208219-5208219 on Build GRCh38
rs745541490 Chr.3: 5208282-5208282 on Build GRCh38
rs974921969 Chr.3: 5208277-5208277 on Build GRCh38
rs760654800 Chr.3: 5208291-5208291 on Build GRCh38
rs779389826 Chr.3: 5208085-5208085 on Build GRCh38
rs777235425 Chr.3: 5208255-5208255 on Build GRCh38
rs995074555 Chr.3: 5208025-5208025 on Build GRCh38
rs922054007 Chr.3: 5208359-5208359 on Build GRCh38
rs541830835 Chr.3: 5208389-5208389 on Build GRCh38
rs764172403 Chr.3: 5208292-5208292 on Build GRCh38
rs201054930 Chr.3: 5208224-5208224 on Build GRCh38
rs772504341 Chr.3: 5208102-5208102 on Build GRCh38
rs1003230855 Chr.3: 5208073-5208073 on Build GRCh38
rs781333235 Chr.3: 5208078-5208078 on Build GRCh38
rs201858816 Chr.3: 5208088-5208088 on Build GRCh38
rs755548077 Chr.3: 5208250-5208250 on Build GRCh38
rs577740480 Chr.3: 5208180-5208180 on Build GRCh38
rs191056239 Chr.3: 5208052-5208052 on Build GRCh38
rs150607992 Chr.3: 5208206-5208206 on Build GRCh38
rs778007196 Chr.3: 5208078-5208078 on Build GRCh38
rs760720751 Chr.3: 5208207-5208207 on Build GRCh38
rs771709061 Chr.3: 5208284-5208284 on Build GRCh38
rs780381956 Chr.3: 5208138-5208138 on Build GRCh38
rs867423282 Chr.3: 5208133-5208133 on Build GRCh38
rs1027557454 Chr.3: 5208256-5208256 on Build GRCh38
rs372559080 Chr.3: 5208043-5208043 on Build GRCh38
rs555353136 Chr.3: 5208033-5208033 on Build GRCh38
rs891815462 Chr.3: 5208039-5208039 on Build GRCh38
rs139708132 Chr.3: 5208231-5208231 on Build GRCh38
rs752929657 Chr.3: 5208083-5208084 on Build GRCh38
rs377014505 Chr.3: 5208003-5208003 on Build GRCh38
rs757501917 Chr.3: 5208364-5208364 on Build GRCh38
rs922188081 Chr.3: 5208005-5208005 on Build GRCh38
rs759489888 Chr.3: 5208053-5208053 on Build GRCh38
rs571876128 Chr.3: 5207982-5207982 on Build GRCh38
rs745430123 Chr.3: 5208291-5208291 on Build GRCh38
rs911634591 Chr.3: 5207989-5207989 on Build GRCh38
rs774174558 Chr.3: 5208177-5208177 on Build GRCh38

View Full Product Details