Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 5209938-5210466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDEM1
Entrez Gene ID
9695
Gene Name
ER degradation enhancing alpha-mannosidase like protein 1
Gene Aliases
EDEM
UniGene
Hs.224616
Gene Chromosome Location
Chr.3: 5187674-5219965 on Build GRCh38
Transcripts
NM_014674.2 XM_011534271.2 XM_011534272.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
420360 c.1527C>G p.F509L Chr.3: 5210192-5210192 on Build GRCh38
3683160 c.1578A>C p.K526N Chr.3: 5210243-5210243 on Build GRCh38
6098267 c.1581C>G p.V527V Chr.3: 5210246-5210246 on Build GRCh38
7365846 c.1541G>C p.G514A Chr.3: 5210206-5210206 on Build GRCh38
6651575 c.1550T>G p.I517S Chr.3: 5210215-5210215 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs910066237 Chr.3: 5210204-5210204 on Build GRCh38
rs1042787860 Chr.3: 5210315-5210315 on Build GRCh38
rs796881503 Chr.3: 5210262-5210262 on Build GRCh38
rs938338538 Chr.3: 5209979-5209979 on Build GRCh38
rs965075370 Chr.3: 5210121-5210121 on Build GRCh38
rs746748633 Chr.3: 5210169-5210169 on Build GRCh38
rs771609449 Chr.3: 5210252-5210252 on Build GRCh38
rs758706824 Chr.3: 5210124-5210124 on Build GRCh38
rs1049007555 Chr.3: 5210050-5210050 on Build GRCh38
rs901385812 Chr.3: 5210312-5210312 on Build GRCh38
rs569941341 Chr.3: 5210157-5210157 on Build GRCh38
rs747660062 Chr.3: 5210024-5210024 on Build GRCh38
rs200565135 Chr.3: 5210135-5210135 on Build GRCh38
rs778502254 Chr.3: 5210153-5210153 on Build GRCh38
rs531352986 Chr.3: 5210399-5210399 on Build GRCh38
rs757865177 Chr.3: 5210293-5210293 on Build GRCh38
rs938189542 Chr.3: 5210194-5210194 on Build GRCh38
rs985249290 Chr.3: 5210100-5210100 on Build GRCh38
rs773409453 Chr.3: 5210202-5210202 on Build GRCh38
rs112319379 Chr.3: 5210136-5210136 on Build GRCh38
rs767876978 Chr.3: 5210248-5210248 on Build GRCh38
rs536066834 Chr.3: 5210053-5210053 on Build GRCh38
rs778469623 Chr.3: 5209962-5209962 on Build GRCh38
rs1050161342 Chr.3: 5210358-5210358 on Build GRCh38
rs759858483 Chr.3: 5210244-5210244 on Build GRCh38
rs918022492 Chr.3: 5210201-5210201 on Build GRCh38
rs998183392 Chr.3: 5210087-5210087 on Build GRCh38
rs751802972 Chr.3: 5210243-5210243 on Build GRCh38
rs1037272703 Chr.3: 5210080-5210080 on Build GRCh38
rs552019857 Chr.3: 5210263-5210263 on Build GRCh38
rs62255914 Chr.3: 5210120-5210120 on Build GRCh38
rs1006699546 Chr.3: 5210424-5210424 on Build GRCh38
rs1055271039 Chr.3: 5210200-5210200 on Build GRCh38
rs1028413113 Chr.3: 5210095-5210095 on Build GRCh38
rs888719314 Chr.3: 5210061-5210061 on Build GRCh38
rs763055131 Chr.3: 5210208-5210208 on Build GRCh38
rs570601981 Chr.3: 5210428-5210428 on Build GRCh38
rs748531324 Chr.3: 5210075-5210075 on Build GRCh38
rs756693328 Chr.3: 5210151-5210151 on Build GRCh38
rs761854881 Chr.3: 5210196-5210196 on Build GRCh38
rs915645227 Chr.3: 5210014-5210014 on Build GRCh38
rs781631315 Chr.3: 5210139-5210139 on Build GRCh38
rs545657306 Chr.3: 5210099-5210099 on Build GRCh38
rs184281153 Chr.3: 5210394-5210394 on Build GRCh38
rs372929013 Chr.3: 5210162-5210162 on Build GRCh38
rs896558907 Chr.3: 5210001-5210001 on Build GRCh38
rs749995398 Chr.3: 5210292-5210292 on Build GRCh38
rs901207501 Chr.3: 5210085-5210085 on Build GRCh38
rs139759614 Chr.3: 5210185-5210185 on Build GRCh38
rs775122598 Chr.3: 5210164-5210164 on Build GRCh38
rs184359596 Chr.3: 5209972-5209972 on Build GRCh38
rs527300151 Chr.3: 5210438-5210438 on Build GRCh38
rs766650628 Chr.3: 5210127-5210127 on Build GRCh38
rs773766672 Chr.3: 5210432-5210432 on Build GRCh38
rs189315538 Chr.3: 5210098-5210098 on Build GRCh38
rs147376764 Chr.3: 5210403-5210403 on Build GRCh38
rs759037616 Chr.3: 5210439-5210439 on Build GRCh38
rs748689473 Chr.3: 5210148-5210148 on Build GRCh38
rs574358486 Chr.3: 5210344-5210344 on Build GRCh38
rs557911036 Chr.3: 5210161-5210161 on Build GRCh38
rs1032664464 Chr.3: 5210076-5210076 on Build GRCh38
rs117396008 Chr.3: 5210000-5210000 on Build GRCh38
rs144357818 Chr.3: 5210186-5210186 on Build GRCh38
rs192670492 Chr.3: 5210382-5210382 on Build GRCh38
rs756960288 Chr.3: 5210288-5210292 on Build GRCh38
rs753148546 Chr.3: 5210261-5210261 on Build GRCh38
rs1020929037 Chr.3: 5210043-5210043 on Build GRCh38
rs554520105 Chr.3: 5210270-5210270 on Build GRCh38
rs540961499 Chr.3: 5210234-5210234 on Build GRCh38
rs775045162 Chr.3: 5210297-5210297 on Build GRCh38

View Full Product Details