Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 112071762-112072262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBE1
Entrez Gene ID
51175
Gene Name
tubulin epsilon 1
Gene Aliases
TUBE, dJ142L7.2
UniGene
Hs.34851
Gene Chromosome Location
Chr.6: 112070657-112087548 on Build GRCh38
Transcripts
NM_016262.4 XM_011535875.2 XM_017010918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372023 c.1121C>A p.S374Y Chr.6: 112072050-112072050 on Build GRCh38
6418832 c.1124G>A p.W375* Chr.6: 112072047-112072047 on Build GRCh38
6420672 c.1235T>C p.L412P Chr.6: 112071936-112071936 on Build GRCh38
5692964 c.1110A>G p.L370L Chr.6: 112072061-112072061 on Build GRCh38
1439864 c.1158C>T p.S386S Chr.6: 112072013-112072013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142963380 Chr.6: 112072000-112072000 on Build GRCh38
rs139268676 Chr.6: 112072067-112072067 on Build GRCh38
rs782054921 Chr.6: 112071986-112071986 on Build GRCh38
rs782076588 Chr.6: 112071960-112071960 on Build GRCh38
rs782432076 Chr.6: 112071948-112071948 on Build GRCh38
rs1020827238 Chr.6: 112071793-112071793 on Build GRCh38
rs782195572 Chr.6: 112071953-112071953 on Build GRCh38
rs369915083 Chr.6: 112072085-112072085 on Build GRCh38
rs782394567 Chr.6: 112071967-112071967 on Build GRCh38
rs782020620 Chr.6: 112072032-112072032 on Build GRCh38
rs782532496 Chr.6: 112071893-112071893 on Build GRCh38
rs922441860 Chr.6: 112071843-112071843 on Build GRCh38
rs368208394 Chr.6: 112072093-112072093 on Build GRCh38
rs782121517 Chr.6: 112071886-112071886 on Build GRCh38
rs140683532 Chr.6: 112071952-112071952 on Build GRCh38
rs373883720 Chr.6: 112071850-112071850 on Build GRCh38
rs267600764 Chr.6: 112072013-112072013 on Build GRCh38
rs782149773 Chr.6: 112072092-112072092 on Build GRCh38
rs587687098 Chr.6: 112072150-112072150 on Build GRCh38
rs782310319 Chr.6: 112072054-112072054 on Build GRCh38
rs367934898 Chr.6: 112072003-112072003 on Build GRCh38
rs782240280 Chr.6: 112071947-112071947 on Build GRCh38
rs782800700 Chr.6: 112072236-112072236 on Build GRCh38
rs782545436 Chr.6: 112072120-112072121 on Build GRCh38
rs587607874 Chr.6: 112071794-112071794 on Build GRCh38
rs967480887 Chr.6: 112071828-112071828 on Build GRCh38
rs373864216 Chr.6: 112071877-112071877 on Build GRCh38
rs781992655 Chr.6: 112071970-112071970 on Build GRCh38
rs782752409 Chr.6: 112071863-112071863 on Build GRCh38
rs782488978 Chr.6: 112071914-112071914 on Build GRCh38
rs781810588 Chr.6: 112071982-112071982 on Build GRCh38
rs587669016 Chr.6: 112072215-112072215 on Build GRCh38
rs368923928 Chr.6: 112072237-112072237 on Build GRCh38
rs781967693 Chr.6: 112072083-112072083 on Build GRCh38
rs782308027 Chr.6: 112071957-112071957 on Build GRCh38
rs782035248 Chr.6: 112072090-112072090 on Build GRCh38
rs781811074 Chr.6: 112072101-112072101 on Build GRCh38
rs782168940 Chr.6: 112071892-112071893 on Build GRCh38
rs782673538 Chr.6: 112071997-112071997 on Build GRCh38
rs781885303 Chr.6: 112071999-112071999 on Build GRCh38
rs911123450 Chr.6: 112071874-112071874 on Build GRCh38
rs782761393 Chr.6: 112072023-112072024 on Build GRCh38
rs781787433 Chr.6: 112072113-112072113 on Build GRCh38
rs144370746 Chr.6: 112071987-112071987 on Build GRCh38
rs781792462 Chr.6: 112071908-112071908 on Build GRCh38
rs200707249 Chr.6: 112071892-112071892 on Build GRCh38
rs985373606 Chr.6: 112071857-112071857 on Build GRCh38
rs782198413 Chr.6: 112072046-112072046 on Build GRCh38
rs782780222 Chr.6: 112072086-112072086 on Build GRCh38
rs782166625 Chr.6: 112071972-112071972 on Build GRCh38
rs17073277 Chr.6: 112072190-112072190 on Build GRCh38
rs117382527 Chr.6: 112072129-112072129 on Build GRCh38
rs782101729 Chr.6: 112072084-112072085 on Build GRCh38
rs147313864 Chr.6: 112071922-112071922 on Build GRCh38
rs782057928 Chr.6: 112071853-112071853 on Build GRCh38
rs116779782 Chr.6: 112071830-112071830 on Build GRCh38
rs782811840 Chr.6: 112071973-112071973 on Build GRCh38
rs782581351 Chr.6: 112071898-112071898 on Build GRCh38
rs782439730 Chr.6: 112072025-112072025 on Build GRCh38
rs201034590 Chr.6: 112071888-112071888 on Build GRCh38
rs781966204 Chr.6: 112071958-112071958 on Build GRCh38
rs373944628 Chr.6: 112071963-112071963 on Build GRCh38

View Full Product Details