Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38901239-38901786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHA2
Entrez Gene ID
59339
Gene Name
pleckstrin homology domain containing A2
Gene Aliases
TAPP2
UniGene
Hs.369123
Gene Chromosome Location
Chr.8: 38901235-38973912 on Build GRCh38
Transcripts
NM_021623.1 XM_017013718.1

SNP Detail

SNP ID Chromosome Location
rs956402434 Chr.8: 38901732-38901732 on Build GRCh38
rs910988254 Chr.8: 38901311-38901311 on Build GRCh38
rs571500002 Chr.8: 38901618-38901618 on Build GRCh38
rs549368506 Chr.8: 38901524-38901524 on Build GRCh38
rs867212797 Chr.8: 38901419-38901419 on Build GRCh38
rs886451098 Chr.8: 38901678-38901678 on Build GRCh38
rs71216693 Chr.8: 38901446-38901447 on Build GRCh38
rs1051035084 Chr.8: 38901563-38901563 on Build GRCh38
rs188847176 Chr.8: 38901293-38901293 on Build GRCh38
rs991037929 Chr.8: 38901717-38901717 on Build GRCh38
rs1039528449 Chr.8: 38901574-38901574 on Build GRCh38
rs942421319 Chr.8: 38901342-38901342 on Build GRCh38
rs560634672 Chr.8: 38901296-38901296 on Build GRCh38
rs968677548 Chr.8: 38901539-38901539 on Build GRCh38
rs978800186 Chr.8: 38901308-38901308 on Build GRCh38
rs923611012 Chr.8: 38901548-38901548 on Build GRCh38
rs990955450 Chr.8: 38901483-38901483 on Build GRCh38
rs527726152 Chr.8: 38901361-38901361 on Build GRCh38
rs1009699826 Chr.8: 38901533-38901533 on Build GRCh38
rs561233419 Chr.8: 38901718-38901718 on Build GRCh38
rs935226473 Chr.8: 38901552-38901552 on Build GRCh38
rs762207197 Chr.8: 38901443-38901444 on Build GRCh38
rs976454391 Chr.8: 38901359-38901359 on Build GRCh38
rs879827294 Chr.8: 38901755-38901755 on Build GRCh38
rs968416252 Chr.8: 38901307-38901307 on Build GRCh38
rs567501977 Chr.8: 38901558-38901558 on Build GRCh38
rs982548240 Chr.8: 38901681-38901681 on Build GRCh38
rs991087542 Chr.8: 38901395-38901413 on Build GRCh38
rs1004063739 Chr.8: 38901479-38901479 on Build GRCh38
rs916372542 Chr.8: 38901496-38901496 on Build GRCh38
rs538160186 Chr.8: 38901559-38901559 on Build GRCh38
rs923581183 Chr.8: 38901731-38901731 on Build GRCh38
rs987104349 Chr.8: 38901739-38901739 on Build GRCh38
rs1035209623 Chr.8: 38901269-38901269 on Build GRCh38
rs954437739 Chr.8: 38901520-38901520 on Build GRCh38
rs1045447770 Chr.8: 38901431-38901431 on Build GRCh38
rs1046966037 Chr.8: 38901387-38901387 on Build GRCh38
rs979170562 Chr.8: 38901724-38901724 on Build GRCh38
rs1027632194 Chr.8: 38901652-38901652 on Build GRCh38
rs888328649 Chr.8: 38901388-38901388 on Build GRCh38
rs117064964 Chr.8: 38901710-38901710 on Build GRCh38
rs890941423 Chr.8: 38901568-38901568 on Build GRCh38
rs956844446 Chr.8: 38901276-38901276 on Build GRCh38
rs979031592 Chr.8: 38901537-38901537 on Build GRCh38
rs550065429 Chr.8: 38901611-38901611 on Build GRCh38
rs939330072 Chr.8: 38901731-38901731 on Build GRCh38
rs549667461 Chr.8: 38901730-38901731 on Build GRCh38
rs1005530021 Chr.8: 38901696-38901696 on Build GRCh38
rs967730843 Chr.8: 38901720-38901720 on Build GRCh38
rs942575011 Chr.8: 38901572-38901572 on Build GRCh38
rs76319743 Chr.8: 38901439-38901440 on Build GRCh38
rs988425110 Chr.8: 38901300-38901300 on Build GRCh38
rs1022551915 Chr.8: 38901301-38901301 on Build GRCh38
rs1035299444 Chr.8: 38901702-38901702 on Build GRCh38
rs1003730798 Chr.8: 38901267-38901267 on Build GRCh38
rs900623929 Chr.8: 38901590-38901590 on Build GRCh38
rs960931633 Chr.8: 38901464-38901464 on Build GRCh38
rs929805548 Chr.8: 38901371-38901371 on Build GRCh38
rs28419696 Chr.8: 38901745-38901745 on Build GRCh38
rs922355369 Chr.8: 38901639-38901639 on Build GRCh38

View Full Product Details