Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38946041-38946546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHA2
Entrez Gene ID
59339
Gene Name
pleckstrin homology domain containing A2
Gene Aliases
TAPP2
UniGene
Hs.369123
Gene Chromosome Location
Chr.8: 38901235-38973912 on Build GRCh38
Transcripts
NM_021623.1 XM_011544605.2 XM_011544606.2 XM_011544607.2 XM_017013718.1 XM_017013719.1 XM_017013720.1

SNP Detail

SNP ID Chromosome Location
rs757735976 Chr.8: 38946156-38946156 on Build GRCh38
rs756580424 Chr.8: 38946306-38946306 on Build GRCh38
rs923121827 Chr.8: 38946312-38946312 on Build GRCh38
rs13259004 Chr.8: 38946375-38946375 on Build GRCh38
rs769721872 Chr.8: 38946238-38946238 on Build GRCh38
rs758554669 Chr.8: 38946221-38946221 on Build GRCh38
rs975949374 Chr.8: 38946296-38946296 on Build GRCh38
rs182005092 Chr.8: 38946362-38946362 on Build GRCh38
rs1019106413 Chr.8: 38946067-38946067 on Build GRCh38
rs770668181 Chr.8: 38946256-38946256 on Build GRCh38
rs1034472761 Chr.8: 38946230-38946230 on Build GRCh38
rs753187006 Chr.8: 38946078-38946078 on Build GRCh38
rs774048628 Chr.8: 38946260-38946260 on Build GRCh38
rs907369194 Chr.8: 38946434-38946434 on Build GRCh38
rs773781800 Chr.8: 38946352-38946352 on Build GRCh38
rs997113002 Chr.8: 38946317-38946317 on Build GRCh38
rs1002146812 Chr.8: 38946475-38946475 on Build GRCh38
rs533443912 Chr.8: 38946504-38946504 on Build GRCh38
rs528797074 Chr.8: 38946336-38946336 on Build GRCh38
rs183766814 Chr.8: 38946471-38946471 on Build GRCh38
rs568293721 Chr.8: 38946282-38946282 on Build GRCh38
rs1024495605 Chr.8: 38946146-38946146 on Build GRCh38
rs752150198 Chr.8: 38946130-38946130 on Build GRCh38
rs746064052 Chr.8: 38946197-38946197 on Build GRCh38
rs770314502 Chr.8: 38946314-38946314 on Build GRCh38
rs941440110 Chr.8: 38946361-38946361 on Build GRCh38
rs528038194 Chr.8: 38946477-38946477 on Build GRCh38
rs764754859 Chr.8: 38946149-38946149 on Build GRCh38
rs1050399258 Chr.8: 38946334-38946334 on Build GRCh38
rs79105253 Chr.8: 38946120-38946120 on Build GRCh38
rs993184186 Chr.8: 38946442-38946442 on Build GRCh38
rs529484454 Chr.8: 38946376-38946376 on Build GRCh38
rs897439055 Chr.8: 38946121-38946121 on Build GRCh38
rs369160683 Chr.8: 38946254-38946254 on Build GRCh38
rs934844509 Chr.8: 38946285-38946285 on Build GRCh38
rs1045980566 Chr.8: 38946387-38946387 on Build GRCh38
rs961874124 Chr.8: 38946093-38946093 on Build GRCh38
rs994433293 Chr.8: 38946378-38946378 on Build GRCh38
rs759164184 Chr.8: 38946269-38946269 on Build GRCh38
rs989811465 Chr.8: 38946252-38946252 on Build GRCh38
rs780126710 Chr.8: 38946222-38946222 on Build GRCh38
rs931774199 Chr.8: 38946320-38946320 on Build GRCh38
rs559712063 Chr.8: 38946136-38946136 on Build GRCh38
rs773178151 Chr.8: 38946248-38946248 on Build GRCh38
rs544641217 Chr.8: 38946135-38946135 on Build GRCh38
rs374130438 Chr.8: 38946115-38946115 on Build GRCh38
rs911335493 Chr.8: 38946350-38946350 on Build GRCh38
rs115233051 Chr.8: 38946487-38946487 on Build GRCh38
rs888825654 Chr.8: 38946340-38946340 on Build GRCh38
rs920399118 Chr.8: 38946150-38946150 on Build GRCh38
rs893287860 Chr.8: 38946297-38946297 on Build GRCh38
rs967292165 Chr.8: 38946266-38946266 on Build GRCh38
rs779442333 Chr.8: 38946163-38946163 on Build GRCh38
rs377082190 Chr.8: 38946237-38946237 on Build GRCh38
rs754249074 Chr.8: 38946154-38946154 on Build GRCh38
rs991776179 Chr.8: 38946214-38946214 on Build GRCh38

View Full Product Details