Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38952394-38952892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHA2
Entrez Gene ID
59339
Gene Name
pleckstrin homology domain containing A2
Gene Aliases
TAPP2
UniGene
Hs.369123
Gene Chromosome Location
Chr.8: 38901235-38973912 on Build GRCh38
Transcripts
NM_021623.1 XM_011544605.2 XM_011544606.2 XM_011544607.2 XM_017013718.1 XM_017013719.1 XM_017013720.1

SNP Detail

SNP ID Chromosome Location
rs1044318056 Chr.8: 38952807-38952808 on Build GRCh38
rs551525193 Chr.8: 38952825-38952825 on Build GRCh38
rs933261697 Chr.8: 38952533-38952533 on Build GRCh38
rs748331352 Chr.8: 38952587-38952587 on Build GRCh38
rs577287566 Chr.8: 38952467-38952467 on Build GRCh38
rs902820844 Chr.8: 38952522-38952522 on Build GRCh38
rs114381450 Chr.8: 38952534-38952534 on Build GRCh38
rs749263592 Chr.8: 38952654-38952654 on Build GRCh38
rs948646340 Chr.8: 38952801-38952801 on Build GRCh38
rs781439408 Chr.8: 38952586-38952586 on Build GRCh38
rs1003391725 Chr.8: 38952514-38952514 on Build GRCh38
rs761397362 Chr.8: 38952652-38952652 on Build GRCh38
rs764936304 Chr.8: 38952662-38952662 on Build GRCh38
rs757951835 Chr.8: 38952685-38952685 on Build GRCh38
rs929008774 Chr.8: 38952780-38952780 on Build GRCh38
rs779449444 Chr.8: 38952688-38952688 on Build GRCh38
rs779873407 Chr.8: 38952581-38952581 on Build GRCh38
rs754904497 Chr.8: 38952655-38952655 on Build GRCh38
rs796491069 Chr.8: 38952670-38952670 on Build GRCh38
rs560534261 Chr.8: 38952839-38952839 on Build GRCh38
rs78456410 Chr.8: 38952603-38952603 on Build GRCh38
rs758158621 Chr.8: 38952521-38952521 on Build GRCh38
rs1012048746 Chr.8: 38952868-38952868 on Build GRCh38
rs1032053460 Chr.8: 38952857-38952857 on Build GRCh38
rs781558994 Chr.8: 38952734-38952734 on Build GRCh38
rs781265670 Chr.8: 38952821-38952821 on Build GRCh38
rs778183843 Chr.8: 38952753-38952753 on Build GRCh38
rs181724553 Chr.8: 38952791-38952791 on Build GRCh38
rs773416359 Chr.8: 38952602-38952602 on Build GRCh38
rs990339657 Chr.8: 38952535-38952535 on Build GRCh38
rs919022448 Chr.8: 38952770-38952770 on Build GRCh38
rs889028026 Chr.8: 38952541-38952541 on Build GRCh38
rs947171378 Chr.8: 38952838-38952838 on Build GRCh38
rs573506202 Chr.8: 38952606-38952606 on Build GRCh38
rs770140365 Chr.8: 38952750-38952750 on Build GRCh38
rs1048905444 Chr.8: 38952792-38952792 on Build GRCh38
rs749901840 Chr.8: 38952663-38952663 on Build GRCh38
rs763912195 Chr.8: 38952641-38952641 on Build GRCh38
rs779771412 Chr.8: 38952621-38952622 on Build GRCh38
rs186349633 Chr.8: 38952795-38952795 on Build GRCh38
rs981406501 Chr.8: 38952761-38952761 on Build GRCh38
rs1016085143 Chr.8: 38952569-38952569 on Build GRCh38
rs544791895 Chr.8: 38952755-38952755 on Build GRCh38
rs981624223 Chr.8: 38952439-38952439 on Build GRCh38
rs777817218 Chr.8: 38952490-38952490 on Build GRCh38
rs540927071 Chr.8: 38952651-38952651 on Build GRCh38
rs879065103 Chr.8: 38952570-38952570 on Build GRCh38
rs937551106 Chr.8: 38952482-38952482 on Build GRCh38
rs562204382 Chr.8: 38952720-38952720 on Build GRCh38
rs954309603 Chr.8: 38952626-38952626 on Build GRCh38
rs991643133 Chr.8: 38952813-38952813 on Build GRCh38
rs914357410 Chr.8: 38952492-38952492 on Build GRCh38
rs1012816150 Chr.8: 38952427-38952427 on Build GRCh38
rs189569674 Chr.8: 38952822-38952822 on Build GRCh38
rs1031751780 Chr.8: 38952517-38952517 on Build GRCh38
rs891879678 Chr.8: 38952867-38952867 on Build GRCh38
rs770150579 Chr.8: 38952600-38952600 on Build GRCh38
rs985666167 Chr.8: 38952636-38952636 on Build GRCh38
rs1035937692 Chr.8: 38952778-38952778 on Build GRCh38
rs544239011 Chr.8: 38952497-38952497 on Build GRCh38
rs760553172 Chr.8: 38952633-38952633 on Build GRCh38
rs188984628 Chr.8: 38952740-38952740 on Build GRCh38
rs1055881168 Chr.8: 38952490-38952490 on Build GRCh38
rs147372019 Chr.8: 38952419-38952420 on Build GRCh38
rs1007425533 Chr.8: 38952546-38952546 on Build GRCh38
rs369737302 Chr.8: 38952632-38952632 on Build GRCh38

View Full Product Details