Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41929760-41930304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT6A
Entrez Gene ID
7994
Gene Name
lysine acetyltransferase 6A
Gene Aliases
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220
UniGene
Hs.491577
Gene Chromosome Location
Chr.8: 41929479-42052026 on Build GRCh38
Transcripts
NM_006766.4 XM_011544656.2 XM_011544657.2 XM_011544658.2 XM_011544659.2 XM_017013863.1 XM_017013864.1

SNP Detail

SNP ID Chromosome Location
rs894296569 Chr.8: 41930137-41930137 on Build GRCh38
rs569739100 Chr.8: 41929983-41929983 on Build GRCh38
rs1040799649 Chr.8: 41930003-41930003 on Build GRCh38
rs770883409 Chr.8: 41929913-41929918 on Build GRCh38
rs982886678 Chr.8: 41930033-41930034 on Build GRCh38
rs1039148755 Chr.8: 41930158-41930158 on Build GRCh38
rs3832594 Chr.8: 41930191-41930195 on Build GRCh38
rs1030926822 Chr.8: 41930217-41930217 on Build GRCh38
rs886727961 Chr.8: 41930236-41930236 on Build GRCh38
rs764285935 Chr.8: 41930190-41930194 on Build GRCh38
rs1030571018 Chr.8: 41930232-41930232 on Build GRCh38
rs978034506 Chr.8: 41929813-41929813 on Build GRCh38
rs997749291 Chr.8: 41929974-41929975 on Build GRCh38
rs958493290 Chr.8: 41929921-41929921 on Build GRCh38
rs565364161 Chr.8: 41930064-41930064 on Build GRCh38
rs772389285 Chr.8: 41930119-41930119 on Build GRCh38
rs779094898 Chr.8: 41930115-41930115 on Build GRCh38
rs532044726 Chr.8: 41930020-41930020 on Build GRCh38
rs775515458 Chr.8: 41930157-41930157 on Build GRCh38
rs1005845718 Chr.8: 41930120-41930120 on Build GRCh38
rs1010002145 Chr.8: 41929931-41929934 on Build GRCh38
rs1046738749 Chr.8: 41930111-41930111 on Build GRCh38
rs561013203 Chr.8: 41929801-41929801 on Build GRCh38
rs576924246 Chr.8: 41929941-41929943 on Build GRCh38
rs903857083 Chr.8: 41930148-41930148 on Build GRCh38
rs145487556 Chr.8: 41929786-41929786 on Build GRCh38
rs999568960 Chr.8: 41930183-41930183 on Build GRCh38
rs955047978 Chr.8: 41930012-41930012 on Build GRCh38
rs748358221 Chr.8: 41930187-41930188 on Build GRCh38
rs958099107 Chr.8: 41930252-41930252 on Build GRCh38
rs183367970 Chr.8: 41930117-41930117 on Build GRCh38
rs903973030 Chr.8: 41929935-41929935 on Build GRCh38
rs941186833 Chr.8: 41929785-41929785 on Build GRCh38
rs549659896 Chr.8: 41929929-41929929 on Build GRCh38
rs1038202314 Chr.8: 41929796-41929796 on Build GRCh38
rs970762850 Chr.8: 41930174-41930174 on Build GRCh38
rs923961882 Chr.8: 41929867-41929867 on Build GRCh38
rs529944897 Chr.8: 41929805-41929805 on Build GRCh38
rs551778037 Chr.8: 41930025-41930025 on Build GRCh38
rs911504317 Chr.8: 41930278-41930278 on Build GRCh38
rs964881287 Chr.8: 41929972-41929973 on Build GRCh38
rs66535265 Chr.8: 41930188-41930192 on Build GRCh38
rs926426151 Chr.8: 41930065-41930065 on Build GRCh38
rs1056793829 Chr.8: 41929806-41929806 on Build GRCh38
rs748921483 Chr.8: 41929918-41929918 on Build GRCh38
rs911373149 Chr.8: 41929959-41929959 on Build GRCh38
rs1042482571 Chr.8: 41929933-41929934 on Build GRCh38
rs962476036 Chr.8: 41930056-41930056 on Build GRCh38
rs1014065004 Chr.8: 41930141-41930141 on Build GRCh38
rs138464098 Chr.8: 41930142-41930143 on Build GRCh38
rs989757146 Chr.8: 41929923-41929923 on Build GRCh38
rs942923097 Chr.8: 41929982-41929982 on Build GRCh38
rs1015292157 Chr.8: 41930040-41930040 on Build GRCh38
rs1003833920 Chr.8: 41930258-41930258 on Build GRCh38
rs990137994 Chr.8: 41930257-41930257 on Build GRCh38
rs776077155 Chr.8: 41930133-41930133 on Build GRCh38
rs937856651 Chr.8: 41930098-41930098 on Build GRCh38
rs900643647 Chr.8: 41930168-41930168 on Build GRCh38
rs922377615 Chr.8: 41930046-41930046 on Build GRCh38

View Full Product Details